Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 28240823-28241371 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JAZF1-AS1
Entrez Gene ID
100128081
Gene Name
JAZF1 antisense RNA 1
Gene Aliases
-
UniGene
Hs.434307
Gene Chromosome Location
Chr.7: 28180457-28241377 on Build GRCh38
Transcripts
NR_034097.1

SNP Detail

SNP ID Chromosome Location
rs571769411 Chr.7: 28240881-28240881 on Build GRCh38
rs939948195 Chr.7: 28241060-28241060 on Build GRCh38
rs930498209 Chr.7: 28241230-28241230 on Build GRCh38
rs534795184 Chr.7: 28240875-28240875 on Build GRCh38
rs376744356 Chr.7: 28240903-28240903 on Build GRCh38
rs960460917 Chr.7: 28240964-28240964 on Build GRCh38
rs907651756 Chr.7: 28241251-28241253 on Build GRCh38
rs1044635847 Chr.7: 28241335-28241335 on Build GRCh38
rs10486580 Chr.7: 28240886-28240886 on Build GRCh38
rs557284705 Chr.7: 28240909-28240909 on Build GRCh38
rs796922958 Chr.7: 28240983-28240983 on Build GRCh38
rs889199494 Chr.7: 28241098-28241098 on Build GRCh38
rs563267780 Chr.7: 28241172-28241172 on Build GRCh38
rs878973530 Chr.7: 28241079-28241079 on Build GRCh38
rs999127255 Chr.7: 28241174-28241174 on Build GRCh38
rs901028793 Chr.7: 28241066-28241066 on Build GRCh38
rs540423518 Chr.7: 28241007-28241007 on Build GRCh38
rs756960738 Chr.7: 28240913-28240913 on Build GRCh38
rs904498443 Chr.7: 28241154-28241154 on Build GRCh38
rs984189000 Chr.7: 28241013-28241013 on Build GRCh38
rs74680487 Chr.7: 28241248-28241248 on Build GRCh38
rs565178681 Chr.7: 28241021-28241021 on Build GRCh38
rs974673278 Chr.7: 28241196-28241196 on Build GRCh38
rs1023065941 Chr.7: 28241028-28241028 on Build GRCh38
rs115768341 Chr.7: 28240946-28240946 on Build GRCh38
rs181404688 Chr.7: 28240950-28240950 on Build GRCh38
rs939896165 Chr.7: 28240884-28240884 on Build GRCh38
rs900979472 Chr.7: 28240887-28240887 on Build GRCh38
rs745420444 Chr.7: 28240974-28240974 on Build GRCh38
rs1051884046 Chr.7: 28241078-28241078 on Build GRCh38
rs537231071 Chr.7: 28241002-28241002 on Build GRCh38
rs553234741 Chr.7: 28240880-28240880 on Build GRCh38
rs57522539 Chr.7: 28240980-28240980 on Build GRCh38
rs151280500 Chr.7: 28241212-28241212 on Build GRCh38
rs1018858640 Chr.7: 28241163-28241163 on Build GRCh38
rs189710563 Chr.7: 28240997-28240997 on Build GRCh38
rs553011743 Chr.7: 28241263-28241263 on Build GRCh38
rs80141853 Chr.7: 28241327-28241327 on Build GRCh38
rs767733323 Chr.7: 28240864-28240864 on Build GRCh38
rs1007604328 Chr.7: 28241108-28241108 on Build GRCh38
rs375230840 Chr.7: 28240914-28240914 on Build GRCh38
rs751057959 Chr.7: 28240876-28240876 on Build GRCh38
rs888355056 Chr.7: 28240848-28240848 on Build GRCh38
rs909512989 Chr.7: 28241030-28241030 on Build GRCh38
rs534817251 Chr.7: 28240976-28240976 on Build GRCh38
rs186513920 Chr.7: 28240988-28240988 on Build GRCh38
rs532822224 Chr.7: 28241065-28241065 on Build GRCh38
rs944513780 Chr.7: 28240910-28240910 on Build GRCh38
rs543268801 Chr.7: 28240947-28240947 on Build GRCh38
rs369603172 Chr.7: 28241147-28241147 on Build GRCh38
rs567547837 Chr.7: 28241306-28241306 on Build GRCh38
rs1030806042 Chr.7: 28240912-28240912 on Build GRCh38
rs1014830103 Chr.7: 28241272-28241272 on Build GRCh38
rs778531937 Chr.7: 28240944-28240944 on Build GRCh38
rs758422212 Chr.7: 28240897-28240901 on Build GRCh38
rs187553127 Chr.7: 28241153-28241153 on Build GRCh38
rs905986489 Chr.7: 28241338-28241339 on Build GRCh38
rs896174352 Chr.7: 28241319-28241319 on Build GRCh38
rs143273278 Chr.7: 28241074-28241074 on Build GRCh38

View Full Product Details