Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 29650965-29651458 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC646762
Entrez Gene ID
646762
Gene Name
uncharacterized LOC646762
Gene Aliases
-
UniGene
Hs.30579
Gene Chromosome Location
Chr.7: 29645922-29685138 on Build GRCh38
Transcripts
NR_024278.1

SNP Detail

SNP ID Chromosome Location
rs867091615 Chr.7: 29651232-29651232 on Build GRCh38
rs941609056 Chr.7: 29651299-29651300 on Build GRCh38
rs573915938 Chr.7: 29651162-29651162 on Build GRCh38
rs886839318 Chr.7: 29650998-29650998 on Build GRCh38
rs535635498 Chr.7: 29651056-29651056 on Build GRCh38
rs1728537 Chr.7: 29651042-29651042 on Build GRCh38
rs1049404966 Chr.7: 29651277-29651277 on Build GRCh38
rs990832393 Chr.7: 29651136-29651136 on Build GRCh38
rs117145942 Chr.7: 29651061-29651061 on Build GRCh38
rs879117538 Chr.7: 29651031-29651031 on Build GRCh38
rs185306757 Chr.7: 29651126-29651126 on Build GRCh38
rs781131330 Chr.7: 29651378-29651378 on Build GRCh38
rs752948642 Chr.7: 29651215-29651215 on Build GRCh38
rs1014168050 Chr.7: 29651362-29651363 on Build GRCh38
rs983603883 Chr.7: 29651176-29651176 on Build GRCh38
rs764821706 Chr.7: 29651036-29651036 on Build GRCh38
rs181325522 Chr.7: 29651283-29651283 on Build GRCh38
rs376795542 Chr.7: 29651240-29651240 on Build GRCh38
rs143767573 Chr.7: 29651299-29651299 on Build GRCh38
rs996917496 Chr.7: 29651043-29651043 on Build GRCh38
rs777315545 Chr.7: 29650991-29650991 on Build GRCh38
rs568996362 Chr.7: 29651109-29651109 on Build GRCh38
rs1007993669 Chr.7: 29651057-29651057 on Build GRCh38
rs577585924 Chr.7: 29651231-29651231 on Build GRCh38
rs181619842 Chr.7: 29651004-29651004 on Build GRCh38
rs959738727 Chr.7: 29651391-29651391 on Build GRCh38
rs970535860 Chr.7: 29651065-29651065 on Build GRCh38
rs1041263475 Chr.7: 29650997-29650997 on Build GRCh38
rs894496994 Chr.7: 29651347-29651347 on Build GRCh38
rs903269782 Chr.7: 29651368-29651368 on Build GRCh38
rs1032398890 Chr.7: 29651379-29651379 on Build GRCh38
rs1047224044 Chr.7: 29651297-29651297 on Build GRCh38
rs28591682 Chr.7: 29651195-29651195 on Build GRCh38
rs563022369 Chr.7: 29651415-29651415 on Build GRCh38
rs949638510 Chr.7: 29651174-29651174 on Build GRCh38
rs1038435750 Chr.7: 29651023-29651023 on Build GRCh38
rs939761246 Chr.7: 29651194-29651194 on Build GRCh38
rs919680420 Chr.7: 29651209-29651209 on Build GRCh38
rs567987643 Chr.7: 29650992-29650992 on Build GRCh38
rs189420205 Chr.7: 29651173-29651173 on Build GRCh38
rs930106242 Chr.7: 29651018-29651019 on Build GRCh38
rs959367222 Chr.7: 29651135-29651135 on Build GRCh38
rs867545214 Chr.7: 29651233-29651233 on Build GRCh38
rs543284154 Chr.7: 29651392-29651392 on Build GRCh38
rs908067952 Chr.7: 29651190-29651190 on Build GRCh38
rs750174204 Chr.7: 29651014-29651014 on Build GRCh38
rs115671734 Chr.7: 29651427-29651427 on Build GRCh38
rs939813314 Chr.7: 29651000-29651000 on Build GRCh38
rs1001110796 Chr.7: 29651326-29651326 on Build GRCh38
rs911003943 Chr.7: 29650986-29650986 on Build GRCh38
rs1045799831 Chr.7: 29651363-29651363 on Build GRCh38
rs781173369 Chr.7: 29651139-29651139 on Build GRCh38
rs778071533 Chr.7: 29651344-29651344 on Build GRCh38
rs536876843 Chr.7: 29651110-29651110 on Build GRCh38
rs757177785 Chr.7: 29651285-29651285 on Build GRCh38
rs902384079 Chr.7: 29651320-29651320 on Build GRCh38
rs1641211 Chr.7: 29651089-29651089 on Build GRCh38
rs1008129227 Chr.7: 29651289-29651289 on Build GRCh38
rs371863502 Chr.7: 29651054-29651054 on Build GRCh38
rs1030817981 Chr.7: 29651424-29651424 on Build GRCh38
rs1056420575 Chr.7: 29651321-29651321 on Build GRCh38
rs891026127 Chr.7: 29651281-29651281 on Build GRCh38

View Full Product Details