Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 66395900-66396433 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00174
Entrez Gene ID
285908
Gene Name
long intergenic non-protein coding RNA 174
Gene Aliases
NCRNA00174
UniGene
Hs.50755
Gene Chromosome Location
Chr.7: 66376044-66400408 on Build GRCh38
Transcripts
NR_026873.1

SNP Detail

SNP ID Chromosome Location
rs869120833 Chr.7: 66396406-66396407 on Build GRCh38
rs774941001 Chr.7: 66396198-66396198 on Build GRCh38
rs775910749 Chr.7: 66396280-66396280 on Build GRCh38
rs544278597 Chr.7: 66396186-66396186 on Build GRCh38
rs966141735 Chr.7: 66396385-66396386 on Build GRCh38
rs940228136 Chr.7: 66396045-66396045 on Build GRCh38
rs374860744 Chr.7: 66396353-66396353 on Build GRCh38
rs528103247 Chr.7: 66396383-66396383 on Build GRCh38
rs772272254 Chr.7: 66396274-66396274 on Build GRCh38
rs1012043121 Chr.7: 66396351-66396351 on Build GRCh38
rs559395926 Chr.7: 66396350-66396350 on Build GRCh38
rs951817851 Chr.7: 66396370-66396371 on Build GRCh38
rs778705 Chr.7: 66396128-66396128 on Build GRCh38
rs184163613 Chr.7: 66396019-66396019 on Build GRCh38
rs763833505 Chr.7: 66396371-66396371 on Build GRCh38
rs983082773 Chr.7: 66396170-66396170 on Build GRCh38
rs1042424142 Chr.7: 66396275-66396275 on Build GRCh38
rs139443483 Chr.7: 66396107-66396107 on Build GRCh38
rs1056826252 Chr.7: 66396316-66396316 on Build GRCh38
rs543714293 Chr.7: 66396243-66396243 on Build GRCh38
rs1027396149 Chr.7: 66396144-66396144 on Build GRCh38
rs932817725 Chr.7: 66396059-66396059 on Build GRCh38
rs542400721 Chr.7: 66396317-66396317 on Build GRCh38
rs368233011 Chr.7: 66396341-66396341 on Build GRCh38
rs749799228 Chr.7: 66396188-66396188 on Build GRCh38
rs781529876 Chr.7: 66396352-66396353 on Build GRCh38
rs189458130 Chr.7: 66396190-66396190 on Build GRCh38
rs1003529328 Chr.7: 66396296-66396296 on Build GRCh38
rs756494657 Chr.7: 66396135-66396135 on Build GRCh38
rs1027406155 Chr.7: 66396366-66396366 on Build GRCh38
rs959493909 Chr.7: 66396130-66396130 on Build GRCh38
rs893895076 Chr.7: 66396273-66396273 on Build GRCh38
rs1020926560 Chr.7: 66396084-66396084 on Build GRCh38
rs901365348 Chr.7: 66396049-66396049 on Build GRCh38
rs954279255 Chr.7: 66396409-66396411 on Build GRCh38
rs749961248 Chr.7: 66396067-66396067 on Build GRCh38
rs769461153 Chr.7: 66396320-66396322 on Build GRCh38
rs553390281 Chr.7: 66396085-66396085 on Build GRCh38
rs61020682 Chr.7: 66396353-66396372 on Build GRCh38
rs180985682 Chr.7: 66395946-66395946 on Build GRCh38
rs757849865 Chr.7: 66396073-66396073 on Build GRCh38
rs930351887 Chr.7: 66396009-66396009 on Build GRCh38
rs922851207 Chr.7: 66396406-66396406 on Build GRCh38
rs62465371 Chr.7: 66396385-66396385 on Build GRCh38
rs149694535 Chr.7: 66396189-66396189 on Build GRCh38
rs536348431 Chr.7: 66396075-66396075 on Build GRCh38
rs551961627 Chr.7: 66396290-66396290 on Build GRCh38
rs759474840 Chr.7: 66396151-66396151 on Build GRCh38
rs922876516 Chr.7: 66396239-66396239 on Build GRCh38
rs902444591 Chr.7: 66396116-66396116 on Build GRCh38
rs990668302 Chr.7: 66396133-66396133 on Build GRCh38
rs762533751 Chr.7: 66396386-66396387 on Build GRCh38
rs6460294 Chr.7: 66396369-66396369 on Build GRCh38
rs537281977 Chr.7: 66395920-66395920 on Build GRCh38
rs985643711 Chr.7: 66396410-66396410 on Build GRCh38
rs766170512 Chr.7: 66396030-66396030 on Build GRCh38
rs893885515 Chr.7: 66396069-66396069 on Build GRCh38
rs908807570 Chr.7: 66396023-66396023 on Build GRCh38
rs754366006 Chr.7: 66396353-66396374 on Build GRCh38
rs181808802 Chr.7: 66396262-66396262 on Build GRCh38
rs1014656719 Chr.7: 66396374-66396385 on Build GRCh38
rs975765926 Chr.7: 66396386-66396389 on Build GRCh38
rs959166946 Chr.7: 66395919-66395919 on Build GRCh38
rs1004617030 Chr.7: 66396372-66396385 on Build GRCh38
rs1041229109 Chr.7: 66396048-66396048 on Build GRCh38
rs1003842884 Chr.7: 66396117-66396117 on Build GRCh38

View Full Product Details