Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 35861422-35861932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LINC00950
Entrez Gene ID
92973
Gene Name
long intergenic non-protein coding RNA 950
Gene Aliases
FP588
UniGene
Hs.12509
Gene Chromosome Location
Chr.9: 35860274-35865518 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

Gene Symbol
TMEM8B
Entrez Gene ID
51754
Gene Name
transmembrane protein 8B
Gene Aliases
C9orf127, NAG-5, NAG5, NGX6
UniGene
Hs.493808
Gene Chromosome Location
Chr.9: 35825817-35865515 on Build GRCh38
Transcripts
NR_024006.1 XM_011517916.2

SNP Detail

SNP ID Chromosome Location
rs1039201126 Chr.9: 35861552-35861552 on Build GRCh38
rs965141133 Chr.9: 35861786-35861786 on Build GRCh38
rs759369759 Chr.9: 35861721-35861721 on Build GRCh38
rs1016221600 Chr.9: 35861735-35861735 on Build GRCh38
rs886073254 Chr.9: 35861579-35861579 on Build GRCh38
rs923396267 Chr.9: 35861851-35861851 on Build GRCh38
rs943587353 Chr.9: 35861532-35861532 on Build GRCh38
rs550755295 Chr.9: 35861472-35861472 on Build GRCh38
rs554451125 Chr.9: 35861740-35861740 on Build GRCh38
rs1042531618 Chr.9: 35861841-35861841 on Build GRCh38
rs770995496 Chr.9: 35861501-35861501 on Build GRCh38
rs974656391 Chr.9: 35861758-35861758 on Build GRCh38
rs898409115 Chr.9: 35861614-35861614 on Build GRCh38
rs118072291 Chr.9: 35861583-35861583 on Build GRCh38
rs987315086 Chr.9: 35861792-35861792 on Build GRCh38
rs148153186 Chr.9: 35861571-35861571 on Build GRCh38
rs983269527 Chr.9: 35861728-35861728 on Build GRCh38
rs933538688 Chr.9: 35861564-35861564 on Build GRCh38
rs967199798 Chr.9: 35861473-35861473 on Build GRCh38
rs77714661 Chr.9: 35861738-35861738 on Build GRCh38
rs552205530 Chr.9: 35861703-35861703 on Build GRCh38
rs923543517 Chr.9: 35861562-35861562 on Build GRCh38
rs1020918144 Chr.9: 35861722-35861722 on Build GRCh38
rs538203834 Chr.9: 35861714-35861714 on Build GRCh38
rs936926693 Chr.9: 35861867-35861867 on Build GRCh38
rs1013764826 Chr.9: 35861650-35861650 on Build GRCh38
rs868058602 Chr.9: 35861765-35861765 on Build GRCh38
rs185529646 Chr.9: 35861661-35861661 on Build GRCh38
rs979505342 Chr.9: 35861514-35861514 on Build GRCh38
rs945583873 Chr.9: 35861822-35861822 on Build GRCh38
rs959965905 Chr.9: 35861707-35861707 on Build GRCh38
rs945531484 Chr.9: 35861525-35861525 on Build GRCh38
rs566460559 Chr.9: 35861590-35861590 on Build GRCh38
rs775151521 Chr.9: 35861835-35861835 on Build GRCh38
rs1055353289 Chr.9: 35861873-35861873 on Build GRCh38
rs894775664 Chr.9: 35861622-35861622 on Build GRCh38
rs561565962 Chr.9: 35861517-35861517 on Build GRCh38
rs954049539 Chr.9: 35861779-35861779 on Build GRCh38
rs933570075 Chr.9: 35861899-35861899 on Build GRCh38
rs925412202 Chr.9: 35861742-35861742 on Build GRCh38
rs1047433354 Chr.9: 35861578-35861578 on Build GRCh38
rs995770537 Chr.9: 35861621-35861621 on Build GRCh38
rs776670152 Chr.9: 35861612-35861612 on Build GRCh38
rs925416116 Chr.9: 35861860-35861860 on Build GRCh38
rs921279201 Chr.9: 35861760-35861760 on Build GRCh38
rs912634957 Chr.9: 35861813-35861813 on Build GRCh38
rs974918592 Chr.9: 35861811-35861811 on Build GRCh38
rs932738179 Chr.9: 35861465-35861465 on Build GRCh38
rs376092312 Chr.9: 35861581-35861581 on Build GRCh38

View Full Product Details