Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 136726669-136727163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNHG7
Entrez Gene ID
84973
Gene Name
small nucleolar RNA host gene 7
Gene Aliases
NCRNA00061
UniGene
Hs.495480
Gene Chromosome Location
Chr.9: 136724594-136728184 on Build GRCh38
Transcripts
NR_003672.2 NR_024542.1 NR_024543.1 NR_002958.1

Gene Symbol
SNORA17A
Entrez Gene ID
677804
Gene Name
small nucleolar RNA, H/ACA box 17A
Gene Aliases
ACA17, SNORA17
UniGene
Hs.495480
Gene Chromosome Location
Chr.9: 136726747-136726879 on Build GRCh38
Transcripts
NR_003672.2 NR_024542.1 NR_024543.1 NR_002958.1

SNP Detail

SNP ID Chromosome Location
rs906088468 Chr.9: 136727101-136727101 on Build GRCh38
rs557173919 Chr.9: 136726985-136726985 on Build GRCh38
rs762178150 Chr.9: 136726817-136726817 on Build GRCh38
rs1017482264 Chr.9: 136727010-136727010 on Build GRCh38
rs967896677 Chr.9: 136726999-136726999 on Build GRCh38
rs764305851 Chr.9: 136726811-136726811 on Build GRCh38
rs915080632 Chr.9: 136727041-136727041 on Build GRCh38
rs770769461 Chr.9: 136726879-136726879 on Build GRCh38
rs766015992 Chr.9: 136726982-136726982 on Build GRCh38
rs552262958 Chr.9: 136726855-136726857 on Build GRCh38
rs375545347 Chr.9: 136726887-136726887 on Build GRCh38
rs751815700 Chr.9: 136726727-136726727 on Build GRCh38
rs781729321 Chr.9: 136726754-136726754 on Build GRCh38
rs879242545 Chr.9: 136727033-136727033 on Build GRCh38
rs921173078 Chr.9: 136727023-136727023 on Build GRCh38
rs760661842 Chr.9: 136726703-136726703 on Build GRCh38
rs761055665 Chr.9: 136726903-136726903 on Build GRCh38
rs370970833 Chr.9: 136726901-136726901 on Build GRCh38
rs1016822366 Chr.9: 136726933-136726933 on Build GRCh38
rs746358429 Chr.9: 136726755-136726755 on Build GRCh38
rs948973894 Chr.9: 136727137-136727137 on Build GRCh38
rs1026390748 Chr.9: 136726952-136726952 on Build GRCh38
rs184256568 Chr.9: 136726733-136726733 on Build GRCh38
rs759579674 Chr.9: 136726886-136726886 on Build GRCh38
rs565955807 Chr.9: 136726829-136726829 on Build GRCh38
rs771749445 Chr.9: 136726867-136726867 on Build GRCh38
rs915009047 Chr.9: 136726837-136726837 on Build GRCh38
rs746127853 Chr.9: 136726728-136726729 on Build GRCh38
rs1051063451 Chr.9: 136727040-136727040 on Build GRCh38
rs899363143 Chr.9: 136727141-136727141 on Build GRCh38
rs769870642 Chr.9: 136726791-136726791 on Build GRCh38
rs913701980 Chr.9: 136726993-136726993 on Build GRCh38
rs945214572 Chr.9: 136727046-136727046 on Build GRCh38
rs774066898 Chr.9: 136726714-136726714 on Build GRCh38
rs561720175 Chr.9: 136726690-136726690 on Build GRCh38
rs777432266 Chr.9: 136726872-136726872 on Build GRCh38
rs773807431 Chr.9: 136726776-136726776 on Build GRCh38
rs898674333 Chr.9: 136726918-136726918 on Build GRCh38
rs773127419 Chr.9: 136726702-136726702 on Build GRCh38
rs987850820 Chr.9: 136726986-136726986 on Build GRCh38
rs774260386 Chr.9: 136727133-136727134 on Build GRCh38
rs931480800 Chr.9: 136727093-136727095 on Build GRCh38
rs899313695 Chr.9: 136726934-136726934 on Build GRCh38
rs755555575 Chr.9: 136726926-136726926 on Build GRCh38
rs1037944683 Chr.9: 136727140-136727140 on Build GRCh38
rs762878594 Chr.9: 136726810-136726810 on Build GRCh38
rs747862546 Chr.9: 136726780-136726780 on Build GRCh38
rs370511505 Chr.9: 136726847-136726847 on Build GRCh38
rs143886682 Chr.9: 136727122-136727122 on Build GRCh38
rs115595451 Chr.9: 136726891-136726891 on Build GRCh38
rs943207398 Chr.9: 136726807-136726807 on Build GRCh38
rs757968367 Chr.9: 136727031-136727031 on Build GRCh38
rs756569427 Chr.9: 136726832-136726832 on Build GRCh38
rs964569098 Chr.9: 136727017-136727017 on Build GRCh38
rs769944531 Chr.9: 136727097-136727100 on Build GRCh38
rs760572069 Chr.9: 136726792-136726792 on Build GRCh38
rs532950511 Chr.9: 136726715-136726715 on Build GRCh38
rs780644032 Chr.9: 136726760-136726760 on Build GRCh38
rs754287075 Chr.9: 136726925-136726925 on Build GRCh38
rs200959197 Chr.9: 136726784-136726784 on Build GRCh38
rs954747756 Chr.9: 136726953-136726953 on Build GRCh38
rs374308919 Chr.9: 136726699-136726699 on Build GRCh38
rs776649549 Chr.9: 136726798-136726798 on Build GRCh38
rs939009085 Chr.9: 136727085-136727085 on Build GRCh38
rs2275159 Chr.9: 136726874-136726874 on Build GRCh38
rs751720389 Chr.9: 136726814-136726814 on Build GRCh38
rs200581664 Chr.9: 136726884-136726884 on Build GRCh38
rs774616295 Chr.9: 136726888-136726888 on Build GRCh38
rs994018155 Chr.9: 136726937-136726937 on Build GRCh38
rs748698225 Chr.9: 136726766-136726766 on Build GRCh38
rs768225828 Chr.9: 136726769-136726769 on Build GRCh38
rs780367789 Chr.9: 136726846-136726846 on Build GRCh38
rs757695258 Chr.9: 136726739-136726739 on Build GRCh38
rs376053295 Chr.9: 136726709-136726709 on Build GRCh38
rs547073627 Chr.9: 136727061-136727061 on Build GRCh38
rs778541354 Chr.9: 136726848-136726848 on Build GRCh38
rs766833140 Chr.9: 136726913-136726913 on Build GRCh38
rs373978138 Chr.9: 136726789-136726789 on Build GRCh38
rs768658762 Chr.9: 136726893-136726917 on Build GRCh38
rs201632813 Chr.9: 136726820-136726820 on Build GRCh38
rs927699513 Chr.9: 136727081-136727081 on Build GRCh38
rs1054767865 Chr.9: 136727114-136727114 on Build GRCh38
rs139828681 Chr.9: 136727103-136727103 on Build GRCh38
rs563443386 Chr.9: 136726725-136726725 on Build GRCh38
rs750687664 Chr.9: 136726902-136726902 on Build GRCh38
rs776589709 Chr.9: 136726986-136726987 on Build GRCh38
rs1037178028 Chr.9: 136726917-136726917 on Build GRCh38
rs756727155 Chr.9: 136726757-136726757 on Build GRCh38
rs770730013 Chr.9: 136726797-136726797 on Build GRCh38
rs567789309 Chr.9: 136727059-136727059 on Build GRCh38
rs747775425 Chr.9: 136726864-136726864 on Build GRCh38
rs775754328 Chr.9: 136726808-136726809 on Build GRCh38
rs2275160 Chr.9: 136726716-136726716 on Build GRCh38

View Full Product Details