Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 12903689-12904200 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TLR8-AS1
Entrez Gene ID
349408
Gene Name
TLR8 antisense RNA 1
Gene Aliases
GS1-324M7.6
UniGene
Hs.685035
Gene Chromosome Location
Chr.X: 12902817-12943300 on Build GRCh38
Transcripts
NR_030727.1

SNP Detail

SNP ID Chromosome Location
rs866847197 Chr.X: 12904169-12904169 on Build GRCh38
rs1003901678 Chr.X: 12903920-12903920 on Build GRCh38
rs940842347 Chr.X: 12904120-12904120 on Build GRCh38
rs932371516 Chr.X: 12903870-12903870 on Build GRCh38
rs917807137 Chr.X: 12903895-12903895 on Build GRCh38
rs746128734 Chr.X: 12903783-12903783 on Build GRCh38
rs1029749063 Chr.X: 12904079-12904079 on Build GRCh38
rs3761623 Chr.X: 12903983-12903983 on Build GRCh38
rs1000184135 Chr.X: 12904000-12904000 on Build GRCh38
rs767182072 Chr.X: 12903994-12903994 on Build GRCh38
rs178993 Chr.X: 12903758-12903758 on Build GRCh38
rs1031115517 Chr.X: 12903838-12903838 on Build GRCh38
rs774756530 Chr.X: 12903976-12903976 on Build GRCh38
rs956514116 Chr.X: 12903828-12903828 on Build GRCh38
rs1046782802 Chr.X: 12904033-12904033 on Build GRCh38
rs1049473634 Chr.X: 12903873-12903873 on Build GRCh38
rs1032129775 Chr.X: 12904009-12904009 on Build GRCh38
rs1000943932 Chr.X: 12903836-12903836 on Build GRCh38
rs1037127034 Chr.X: 12903885-12903885 on Build GRCh38
rs1040951852 Chr.X: 12904155-12904155 on Build GRCh38
rs1012981062 Chr.X: 12903981-12903981 on Build GRCh38
rs954231486 Chr.X: 12904110-12904110 on Build GRCh38
rs941052031 Chr.X: 12903880-12903880 on Build GRCh38
rs909529503 Chr.X: 12903875-12903875 on Build GRCh38
rs760468684 Chr.X: 12904121-12904121 on Build GRCh38
rs969711219 Chr.X: 12903896-12903896 on Build GRCh38
rs894983464 Chr.X: 12903970-12903970 on Build GRCh38
rs751995420 Chr.X: 12903852-12903852 on Build GRCh38
rs143125955 Chr.X: 12904035-12904035 on Build GRCh38
rs976944107 Chr.X: 12903844-12903844 on Build GRCh38
rs979394231 Chr.X: 12903763-12903763 on Build GRCh38
rs756971420 Chr.X: 12903999-12903999 on Build GRCh38
rs3761621 Chr.X: 12903891-12903891 on Build GRCh38
rs192205596 Chr.X: 12903826-12903826 on Build GRCh38
rs985149311 Chr.X: 12904162-12904162 on Build GRCh38
rs745387096 Chr.X: 12904071-12904071 on Build GRCh38
rs766883373 Chr.X: 12903822-12903822 on Build GRCh38
rs3761622 Chr.X: 12903932-12903932 on Build GRCh38
rs922807952 Chr.X: 12903861-12903861 on Build GRCh38
rs757917854 Chr.X: 12904088-12904088 on Build GRCh38
rs184955146 Chr.X: 12903905-12903905 on Build GRCh38

View Full Product Details