Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 73827305-73827800 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSIX
Entrez Gene ID
9383
Gene Name
TSIX transcript, XIST antisense RNA
Gene Aliases
LINC00013, NCRNA00013, XIST-AS, XIST-AS1, XISTAS
UniGene
Hs.529901
Gene Chromosome Location
Chr.X: 73792205-73829231 on Build GRCh38
Transcripts
NR_003255.2 NR_001564.2

Gene Symbol
XIST
Entrez Gene ID
7503
Gene Name
X inactive specific transcript (non-protein coding)
Gene Aliases
DXS1089, DXS399E, LINC00001, NCRNA00001, SXI1, swd66
UniGene
Hs.529901
Gene Chromosome Location
Chr.X: 73820651-73852753 on Build GRCh38
Transcripts
NR_003255.2 NR_001564.2

SNP Detail

SNP ID Chromosome Location
rs760757083 Chr.X: 73827476-73827476 on Build GRCh38
rs767152670 Chr.X: 73827367-73827367 on Build GRCh38
rs751370249 Chr.X: 73827363-73827363 on Build GRCh38
rs757090413 Chr.X: 73827366-73827366 on Build GRCh38
rs775052213 Chr.X: 73827426-73827426 on Build GRCh38
rs773402438 Chr.X: 73827417-73827417 on Build GRCh38
rs758714784 Chr.X: 73827752-73827752 on Build GRCh38
rs759499507 Chr.X: 73827691-73827691 on Build GRCh38
rs758528014 Chr.X: 73827552-73827552 on Build GRCh38
rs781675944 Chr.X: 73827589-73827589 on Build GRCh38
rs1030538660 Chr.X: 73827758-73827758 on Build GRCh38
rs747368943 Chr.X: 73827587-73827587 on Build GRCh38
rs771489927 Chr.X: 73827588-73827588 on Build GRCh38
rs1023532712 Chr.X: 73827568-73827569 on Build GRCh38
rs747601658 Chr.X: 73827420-73827420 on Build GRCh38
rs774060263 Chr.X: 73827443-73827443 on Build GRCh38
rs752988454 Chr.X: 73827504-73827504 on Build GRCh38
rs773145037 Chr.X: 73827710-73827710 on Build GRCh38
rs761320981 Chr.X: 73827444-73827444 on Build GRCh38
rs780317832 Chr.X: 73827397-73827397 on Build GRCh38
rs552073275 Chr.X: 73827629-73827629 on Build GRCh38
rs760704037 Chr.X: 73827595-73827596 on Build GRCh38
rs768643046 Chr.X: 73827621-73827621 on Build GRCh38
rs778240837 Chr.X: 73827569-73827569 on Build GRCh38
rs749025718 Chr.X: 73827724-73827724 on Build GRCh38
rs192417232 Chr.X: 73827457-73827457 on Build GRCh38
rs763540281 Chr.X: 73827436-73827436 on Build GRCh38
rs73486505 Chr.X: 73827734-73827734 on Build GRCh38
rs767391387 Chr.X: 73827445-73827445 on Build GRCh38
rs746310444 Chr.X: 73827602-73827602 on Build GRCh38
rs779921149 Chr.X: 73827685-73827685 on Build GRCh38
rs763732059 Chr.X: 73827361-73827361 on Build GRCh38
rs749370295 Chr.X: 73827402-73827402 on Build GRCh38
rs755112264 Chr.X: 73827498-73827498 on Build GRCh38
rs771397792 Chr.X: 73827422-73827422 on Build GRCh38
rs1057464 Chr.X: 73827751-73827751 on Build GRCh38
rs369143730 Chr.X: 73827408-73827408 on Build GRCh38
rs749328226 Chr.X: 73827383-73827383 on Build GRCh38
rs762440675 Chr.X: 73827435-73827435 on Build GRCh38
rs920244259 Chr.X: 73827759-73827759 on Build GRCh38
rs372996682 Chr.X: 73827410-73827410 on Build GRCh38
rs745409675 Chr.X: 73827400-73827404 on Build GRCh38
rs945314533 Chr.X: 73827356-73827356 on Build GRCh38
rs1053692984 Chr.X: 73827437-73827437 on Build GRCh38
rs113288112 Chr.X: 73827616-73827616 on Build GRCh38
rs544056508 Chr.X: 73827449-73827449 on Build GRCh38
rs377405460 Chr.X: 73827338-73827338 on Build GRCh38
rs750393927 Chr.X: 73827370-73827370 on Build GRCh38
rs937656087 Chr.X: 73827474-73827474 on Build GRCh38
rs766209213 Chr.X: 73827634-73827634 on Build GRCh38
rs147047435 Chr.X: 73827492-73827492 on Build GRCh38
rs768188002 Chr.X: 73827604-73827604 on Build GRCh38
rs778816650 Chr.X: 73827500-73827500 on Build GRCh38
rs973161200 Chr.X: 73827635-73827635 on Build GRCh38
rs751546187 Chr.X: 73827560-73827560 on Build GRCh38
rs766405635 Chr.X: 73827491-73827491 on Build GRCh38
rs1012106872 Chr.X: 73827494-73827494 on Build GRCh38
rs762638094 Chr.X: 73827339-73827339 on Build GRCh38
rs755308671 Chr.X: 73827406-73827406 on Build GRCh38
rs761049284 Chr.X: 73827336-73827336 on Build GRCh38
rs761563798 Chr.X: 73827617-73827617 on Build GRCh38
rs753855509 Chr.X: 73827684-73827684 on Build GRCh38

View Full Product Details