Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 132668074-132668590 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HS6ST2
Entrez Gene ID
90161
Gene Name
heparan sulfate 6-O-sulfotransferase 2
Gene Aliases
-
UniGene
Hs.385956
Gene Chromosome Location
Chr.X: 132626010-132961395 on Build GRCh38
Transcripts
NM_001077188.1 NM_147175.3 XM_005262490.3 XM_005262491.2 XM_011531406.1 XM_017029944.1 XM_017029945.1 XM_017029946.1 NR_046691.1

Gene Symbol
HS6ST2-AS1
Entrez Gene ID
100874102
Gene Name
HS6ST2 antisense RNA 1
Gene Aliases
-
UniGene
Hs.619341
Gene Chromosome Location
Chr.X: 132667642-132669888 on Build GRCh38
Transcripts
NM_001077188.1 NM_147175.3 XM_005262490.3 XM_005262491.2 XM_011531406.1 XM_017029944.1 XM_017029945.1 XM_017029946.1 NR_046691.1

SNP Detail

SNP ID Chromosome Location
rs549441105 Chr.X: 132668356-132668356 on Build GRCh38
rs915792574 Chr.X: 132668430-132668430 on Build GRCh38
rs920428612 Chr.X: 132668392-132668392 on Build GRCh38
rs991401883 Chr.X: 132668424-132668424 on Build GRCh38
rs1011098147 Chr.X: 132668258-132668258 on Build GRCh38
rs892235053 Chr.X: 132668465-132668465 on Build GRCh38
rs975331756 Chr.X: 132668528-132668528 on Build GRCh38
rs953635165 Chr.X: 132668323-132668323 on Build GRCh38
rs1008486029 Chr.X: 132668329-132668329 on Build GRCh38
rs778507076 Chr.X: 132668420-132668420 on Build GRCh38
rs1046402638 Chr.X: 132668452-132668452 on Build GRCh38
rs1045174680 Chr.X: 132668362-132668362 on Build GRCh38
rs1052587478 Chr.X: 132668200-132668200 on Build GRCh38
rs5930574 Chr.X: 132668260-132668260 on Build GRCh38
rs930673779 Chr.X: 132668138-132668138 on Build GRCh38
rs893964123 Chr.X: 132668490-132668490 on Build GRCh38
rs1056890789 Chr.X: 132668347-132668347 on Build GRCh38
rs1019734436 Chr.X: 132668514-132668514 on Build GRCh38
rs1030695533 Chr.X: 132668454-132668454 on Build GRCh38
rs34860051 Chr.X: 132668159-132668160 on Build GRCh38
rs1000037374 Chr.X: 132668198-132668198 on Build GRCh38
rs243466 Chr.X: 132668311-132668311 on Build GRCh38
rs904049840 Chr.X: 132668390-132668390 on Build GRCh38
rs908716780 Chr.X: 132668485-132668485 on Build GRCh38
rs1029231502 Chr.X: 132668309-132668309 on Build GRCh38
rs952007175 Chr.X: 132668423-132668423 on Build GRCh38
rs552954659 Chr.X: 132668478-132668478 on Build GRCh38
rs758085610 Chr.X: 132668527-132668527 on Build GRCh38
rs948337730 Chr.X: 132668357-132668357 on Build GRCh38
rs756830763 Chr.X: 132668369-132668369 on Build GRCh38
rs935409360 Chr.X: 132668473-132668473 on Build GRCh38
rs756690711 Chr.X: 132668136-132668136 on Build GRCh38
rs778981820 Chr.X: 132668438-132668438 on Build GRCh38
rs984890487 Chr.X: 132668224-132668224 on Build GRCh38
rs906579507 Chr.X: 132668162-132668162 on Build GRCh38

View Full Product Details