Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 135423714-135424217 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SMIM10L2A
Entrez Gene ID
399668
Gene Name
small integral membrane protein 10 like 2A
Gene Aliases
LED, LINC00086, NCRNA00086
UniGene
Hs.374414
Gene Chromosome Location
Chr.X: 135421933-135428074 on Build GRCh38
Transcripts
NR_024359.2

SNP Detail

SNP ID Chromosome Location
rs782208768 Chr.X: 135423830-135423830 on Build GRCh38
rs782252309 Chr.X: 135424033-135424033 on Build GRCh38
rs782042124 Chr.X: 135424000-135424000 on Build GRCh38
rs782473049 Chr.X: 135423847-135423847 on Build GRCh38
rs782249543 Chr.X: 135424132-135424132 on Build GRCh38
rs370651838 Chr.X: 135424166-135424166 on Build GRCh38
rs375009189 Chr.X: 135424019-135424019 on Build GRCh38
rs45601034 Chr.X: 135424194-135424194 on Build GRCh38
rs782400363 Chr.X: 135424089-135424089 on Build GRCh38
rs782471147 Chr.X: 135423864-135423864 on Build GRCh38
rs782650904 Chr.X: 135423812-135423812 on Build GRCh38
rs147145962 Chr.X: 135424167-135424167 on Build GRCh38
rs782188517 Chr.X: 135423879-135423879 on Build GRCh38
rs6635045 Chr.X: 135423866-135423866 on Build GRCh38
rs782404292 Chr.X: 135423992-135423992 on Build GRCh38
rs782402868 Chr.X: 135423979-135423979 on Build GRCh38
rs371325430 Chr.X: 135423993-135423993 on Build GRCh38
rs567086854 Chr.X: 135424162-135424162 on Build GRCh38
rs143382907 Chr.X: 135424117-135424117 on Build GRCh38

View Full Product Details