Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.Y: 22297407-22297902 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTTY5
Entrez Gene ID
83863
Gene Name
testis-specific transcript, Y-linked 5 (non-protein coding)
Gene Aliases
LINC00126, NCRNA00126, TTY5
UniGene
Hs.121389
Gene Chromosome Location
Chr.Y: 22296798-22298876 on Build GRCh38
Transcripts
NR_001541.1

SNP Detail

SNP ID Chromosome Location
rs893499444 Chr.Y: 22297753-22297753 on Build GRCh38
rs113205557 Chr.Y: 22297705-22297705 on Build GRCh38
rs1045017758 Chr.Y: 22297819-22297819 on Build GRCh38
rs987585363 Chr.Y: 22297625-22297625 on Build GRCh38
rs868238080 Chr.Y: 22297538-22297538 on Build GRCh38
rs112061226 Chr.Y: 22297751-22297751 on Build GRCh38
rs28542474 Chr.Y: 22297689-22297689 on Build GRCh38
rs559409606 Chr.Y: 22297687-22297687 on Build GRCh38
rs923709877 Chr.Y: 22297655-22297655 on Build GRCh38
rs947928378 Chr.Y: 22297754-22297754 on Build GRCh38

View Full Product Details