Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 96871452-96872003 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100132781
Entrez Gene ID
100132781
Gene Name
cyclin Y like 1 pseudogene
Gene Aliases
-
UniGene
Hs.710921
Gene Chromosome Location
Chr.9: 96868900-96872228 on Build GRCh38
Transcripts
NR_038890.1 XM_011518315.2

Gene Symbol
ZNF782
Entrez Gene ID
158431
Gene Name
zinc finger protein 782
Gene Aliases
-
UniGene
Hs.30567
Gene Chromosome Location
Chr.9: 96815628-96876698 on Build GRCh38
Transcripts
NR_038890.1 XM_011518315.2

SNP Detail

SNP ID Chromosome Location
rs182860322 Chr.9: 96871833-96871833 on Build GRCh38
rs147459476 Chr.9: 96871804-96871804 on Build GRCh38
rs138880239 Chr.9: 96871913-96871913 on Build GRCh38
rs969115572 Chr.9: 96871892-96871892 on Build GRCh38
rs924912262 Chr.9: 96871531-96871531 on Build GRCh38
rs13289271 Chr.9: 96871895-96871895 on Build GRCh38
rs1037707904 Chr.9: 96871704-96871704 on Build GRCh38
rs899203718 Chr.9: 96871787-96871787 on Build GRCh38
rs1020714369 Chr.9: 96871548-96871548 on Build GRCh38
rs557458601 Chr.9: 96871623-96871623 on Build GRCh38
rs13283192 Chr.9: 96871888-96871888 on Build GRCh38
rs1023101573 Chr.9: 96871860-96871860 on Build GRCh38
rs376147187 Chr.9: 96871519-96871519 on Build GRCh38
rs13284878 Chr.9: 96871811-96871811 on Build GRCh38
rs988495606 Chr.9: 96871572-96871572 on Build GRCh38
rs541808036 Chr.9: 96871620-96871620 on Build GRCh38
rs564132722 Chr.9: 96871797-96871797 on Build GRCh38
rs913804119 Chr.9: 96871595-96871595 on Build GRCh38
rs896046388 Chr.9: 96871809-96871809 on Build GRCh38
rs13289155 Chr.9: 96871854-96871854 on Build GRCh38
rs751264992 Chr.9: 96871927-96871927 on Build GRCh38
rs779295428 Chr.9: 96871687-96871687 on Build GRCh38
rs1002977424 Chr.9: 96871597-96871597 on Build GRCh38
rs13284893 Chr.9: 96871857-96871857 on Build GRCh38
rs746634330 Chr.9: 96871958-96871958 on Build GRCh38
rs757745331 Chr.9: 96871624-96871624 on Build GRCh38
rs958765689 Chr.9: 96871663-96871663 on Build GRCh38
rs771701827 Chr.9: 96871906-96871906 on Build GRCh38
rs903147151 Chr.9: 96871580-96871580 on Build GRCh38
rs546242626 Chr.9: 96871796-96871796 on Build GRCh38
rs575669817 Chr.9: 96871701-96871701 on Build GRCh38
rs886708512 Chr.9: 96871700-96871700 on Build GRCh38
rs142963056 Chr.9: 96871544-96871544 on Build GRCh38
rs933171288 Chr.9: 96871615-96871615 on Build GRCh38
rs1011825970 Chr.9: 96871529-96871529 on Build GRCh38
rs969083073 Chr.9: 96871498-96871498 on Build GRCh38
rs943399756 Chr.9: 96871644-96871644 on Build GRCh38
rs528929095 Chr.9: 96871930-96871930 on Build GRCh38

View Full Product Details