Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27463013-27463559 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PPP4R3CP
Entrez Gene ID
139420
Gene Name
protein phosphatase 4 regulatory subunit 3C, pseudogene
Gene Aliases
FLFL3P, SMEK3P, smk1
UniGene
Hs.350673
Gene Chromosome Location
Chr.X: 27460211-27463341 on Build GRCh38
Transcripts
NR_002784.2

SNP Detail

SNP ID Chromosome Location
rs777808766 Chr.X: 27463225-27463225 on Build GRCh38
rs267606427 Chr.X: 27463188-27463188 on Build GRCh38
rs751741879 Chr.X: 27463035-27463035 on Build GRCh38
rs771377872 Chr.X: 27463405-27463405 on Build GRCh38
rs180952925 Chr.X: 27463330-27463330 on Build GRCh38
rs939424786 Chr.X: 27463247-27463247 on Build GRCh38
rs758292930 Chr.X: 27463299-27463299 on Build GRCh38
rs189746792 Chr.X: 27463263-27463263 on Build GRCh38
rs373625965 Chr.X: 27463487-27463487 on Build GRCh38
rs902056222 Chr.X: 27463397-27463397 on Build GRCh38
rs558686424 Chr.X: 27463150-27463150 on Build GRCh38
rs1006572594 Chr.X: 27463450-27463450 on Build GRCh38
rs967393396 Chr.X: 27463479-27463479 on Build GRCh38
rs993070524 Chr.X: 27463458-27463458 on Build GRCh38
rs1016586730 Chr.X: 27463468-27463468 on Build GRCh38
rs953779253 Chr.X: 27463165-27463165 on Build GRCh38
rs913207652 Chr.X: 27463170-27463170 on Build GRCh38
rs771484463 Chr.X: 27463290-27463290 on Build GRCh38
rs1001714154 Chr.X: 27463425-27463425 on Build GRCh38
rs1039906989 Chr.X: 27463282-27463282 on Build GRCh38
rs772153687 Chr.X: 27463173-27463173 on Build GRCh38
rs972611069 Chr.X: 27463490-27463490 on Build GRCh38
rs922398075 Chr.X: 27463502-27463502 on Build GRCh38
rs1016723694 Chr.X: 27463409-27463409 on Build GRCh38
rs775058620 Chr.X: 27463305-27463305 on Build GRCh38
rs975288896 Chr.X: 27463494-27463494 on Build GRCh38
rs766560439 Chr.X: 27463034-27463034 on Build GRCh38
rs770818992 Chr.X: 27463249-27463249 on Build GRCh38
rs762211760 Chr.X: 27463478-27463478 on Build GRCh38
rs1029439186 Chr.X: 27463441-27463441 on Build GRCh38
rs778634005 Chr.X: 27463169-27463169 on Build GRCh38
rs889521499 Chr.X: 27463447-27463447 on Build GRCh38
rs747025871 Chr.X: 27463472-27463472 on Build GRCh38
rs960295488 Chr.X: 27463446-27463446 on Build GRCh38
rs904125431 Chr.X: 27463333-27463333 on Build GRCh38
rs769839940 Chr.X: 27463323-27463323 on Build GRCh38
rs753503492 Chr.X: 27463536-27463536 on Build GRCh38
rs962592235 Chr.X: 27463190-27463190 on Build GRCh38
rs191991004 Chr.X: 27463403-27463403 on Build GRCh38
rs752026417 Chr.X: 27463451-27463451 on Build GRCh38
rs12837086 Chr.X: 27463511-27463511 on Build GRCh38
rs935559389 Chr.X: 27463345-27463345 on Build GRCh38
rs901091510 Chr.X: 27463283-27463283 on Build GRCh38
rs778727584 Chr.X: 27463255-27463255 on Build GRCh38
rs978841997 Chr.X: 27463201-27463201 on Build GRCh38
rs942581322 Chr.X: 27463232-27463232 on Build GRCh38
rs1025558314 Chr.X: 27463491-27463491 on Build GRCh38
rs184287451 Chr.X: 27463246-27463246 on Build GRCh38
rs980674437 Chr.X: 27463135-27463135 on Build GRCh38
rs1034909001 Chr.X: 27463437-27463437 on Build GRCh38
rs753149856 Chr.X: 27463140-27463140 on Build GRCh38
rs754501760 Chr.X: 27463033-27463033 on Build GRCh38
rs781276374 Chr.X: 27463057-27463057 on Build GRCh38
rs904907543 Chr.X: 27463418-27463418 on Build GRCh38
rs181556689 Chr.X: 27463110-27463110 on Build GRCh38
rs960610581 Chr.X: 27463504-27463504 on Build GRCh38
rs978727452 Chr.X: 27463481-27463481 on Build GRCh38
rs759791819 Chr.X: 27463317-27463317 on Build GRCh38
rs185944459 Chr.X: 27463364-27463364 on Build GRCh38
rs964434937 Chr.X: 27463493-27463493 on Build GRCh38
rs933870678 Chr.X: 27463326-27463326 on Build GRCh38
rs755036160 Chr.X: 27463056-27463056 on Build GRCh38
rs267606428 Chr.X: 27463189-27463189 on Build GRCh38
rs370086455 Chr.X: 27463322-27463322 on Build GRCh38
rs1034615369 Chr.X: 27463139-27463139 on Build GRCh38
rs777586977 Chr.X: 27463413-27463413 on Build GRCh38

View Full Product Details