Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 86647960-86648517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLCA3P
Entrez Gene ID
9629
Gene Name
chloride channel accessory 3, pseudogene
Gene Aliases
CLCA3
UniGene
Hs.673847
Gene Chromosome Location
Chr.1: 86634276-86655376 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

Gene Symbol
LOC105378828
Entrez Gene ID
105378828
Gene Name
uncharacterized LOC105378828
Gene Aliases
-
UniGene
Hs.175285
Gene Chromosome Location
Chr.1: 86571181-86704493 on Build GRCh38
Transcripts
NR_024604.1 NR_135837.1

SNP Detail

SNP ID Chromosome Location
rs929183520 Chr.1: 86648190-86648190 on Build GRCh38
rs61460736 Chr.1: 86648080-86648080 on Build GRCh38
rs569321183 Chr.1: 86648021-86648021 on Build GRCh38
rs1053708293 Chr.1: 86648380-86648380 on Build GRCh38
rs545050840 Chr.1: 86648336-86648336 on Build GRCh38
rs530776027 Chr.1: 86648473-86648473 on Build GRCh38
rs965407109 Chr.1: 86648339-86648339 on Build GRCh38
rs937939271 Chr.1: 86648378-86648378 on Build GRCh38
rs575056469 Chr.1: 86648357-86648357 on Build GRCh38
rs972256377 Chr.1: 86648314-86648314 on Build GRCh38
rs975993317 Chr.1: 86648148-86648148 on Build GRCh38
rs766245858 Chr.1: 86648131-86648131 on Build GRCh38
rs114096608 Chr.1: 86648238-86648238 on Build GRCh38
rs1053994336 Chr.1: 86648325-86648325 on Build GRCh38
rs1042079603 Chr.1: 86648484-86648484 on Build GRCh38
rs759248736 Chr.1: 86648348-86648348 on Build GRCh38
rs985106792 Chr.1: 86648384-86648384 on Build GRCh38
rs919192993 Chr.1: 86648172-86648172 on Build GRCh38
rs981175765 Chr.1: 86648207-86648207 on Build GRCh38
rs763531410 Chr.1: 86648464-86648464 on Build GRCh38
rs778574671 Chr.1: 86648468-86648468 on Build GRCh38
rs910066276 Chr.1: 86648222-86648222 on Build GRCh38
rs1019606529 Chr.1: 86648109-86648109 on Build GRCh38
rs984379016 Chr.1: 86648202-86648202 on Build GRCh38
rs187579303 Chr.1: 86648334-86648334 on Build GRCh38
rs1035478808 Chr.1: 86648475-86648475 on Build GRCh38
rs1013876488 Chr.1: 86648008-86648008 on Build GRCh38
rs902772852 Chr.1: 86648285-86648286 on Build GRCh38
rs965358159 Chr.1: 86648097-86648097 on Build GRCh38
rs1028311739 Chr.1: 86648471-86648471 on Build GRCh38
rs954268911 Chr.1: 86648199-86648199 on Build GRCh38
rs542849827 Chr.1: 86648326-86648326 on Build GRCh38
rs893665447 Chr.1: 86648410-86648410 on Build GRCh38
rs937811996 Chr.1: 86648209-86648209 on Build GRCh38
rs918973186 Chr.1: 86648319-86648319 on Build GRCh38
rs180838048 Chr.1: 86648329-86648329 on Build GRCh38
rs990631298 Chr.1: 86648459-86648459 on Build GRCh38
rs1001486205 Chr.1: 86648286-86648286 on Build GRCh38
rs141461415 Chr.1: 86647983-86647983 on Build GRCh38
rs755589633 Chr.1: 86648449-86648449 on Build GRCh38
rs926539899 Chr.1: 86648377-86648377 on Build GRCh38
rs1042868952 Chr.1: 86648440-86648440 on Build GRCh38
rs1026030164 Chr.1: 86648364-86648364 on Build GRCh38
rs1021201487 Chr.1: 86648028-86648028 on Build GRCh38
rs752293358 Chr.1: 86648405-86648405 on Build GRCh38
rs189608061 Chr.1: 86648206-86648206 on Build GRCh38
rs965336837 Chr.1: 86648122-86648122 on Build GRCh38
rs998103112 Chr.1: 86648108-86648108 on Build GRCh38
rs146869188 Chr.1: 86648355-86648355 on Build GRCh38
rs372419206 Chr.1: 86648208-86648208 on Build GRCh38
rs901244658 Chr.1: 86648448-86648448 on Build GRCh38
rs954156974 Chr.1: 86648106-86648106 on Build GRCh38
rs896148331 Chr.1: 86647984-86647984 on Build GRCh38
rs1042048046 Chr.1: 86648284-86648284 on Build GRCh38
rs543907575 Chr.1: 86648422-86648422 on Build GRCh38
rs1006254716 Chr.1: 86648479-86648479 on Build GRCh38
rs949051311 Chr.1: 86648265-86648265 on Build GRCh38
rs1010106405 Chr.1: 86648073-86648073 on Build GRCh38

View Full Product Details