Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 58715805-58716353 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BICC1
Entrez Gene ID
80114
Gene Name
BicC family RNA binding protein 1
Gene Aliases
BICC, CYSRD
UniGene
Hs.100261
Gene Chromosome Location
Chr.10: 58512970-58831437 on Build GRCh38
Transcripts
NM_001080512.2 XM_005270169.4 XM_011540185.2 XM_011540188.2 XM_011540189.2 XM_011540190.2 XM_017016677.1 XM_017016678.1 NR_027508.1

Gene Symbol
FAM133CP
Entrez Gene ID
728640
Gene Name
family with sequence similarity 133, member A pseudogene
Gene Aliases
-
UniGene
Hs.729209
Gene Chromosome Location
Chr.10: 58715015-58717534 on Build GRCh38
Transcripts
NM_001080512.2 XM_005270169.4 XM_011540185.2 XM_011540188.2 XM_011540189.2 XM_011540190.2 XM_017016677.1 XM_017016678.1 NR_027508.1

SNP Detail

SNP ID Chromosome Location
rs140119318 Chr.10: 58716194-58716194 on Build GRCh38
rs982286864 Chr.10: 58715889-58715889 on Build GRCh38
rs528608501 Chr.10: 58715964-58715964 on Build GRCh38
rs534556969 Chr.10: 58716200-58716200 on Build GRCh38
rs1015017521 Chr.10: 58716169-58716169 on Build GRCh38
rs911965807 Chr.10: 58716327-58716327 on Build GRCh38
rs986560424 Chr.10: 58716304-58716304 on Build GRCh38
rs142136761 Chr.10: 58715924-58715924 on Build GRCh38
rs1033343778 Chr.10: 58716064-58716064 on Build GRCh38
rs1041599541 Chr.10: 58715995-58715995 on Build GRCh38
rs529922187 Chr.10: 58716020-58716020 on Build GRCh38
rs3816114 Chr.10: 58715963-58715963 on Build GRCh38
rs186973109 Chr.10: 58716078-58716078 on Build GRCh38
rs536927134 Chr.10: 58716174-58716174 on Build GRCh38
rs778789890 Chr.10: 58716262-58716262 on Build GRCh38
rs1024057697 Chr.10: 58716082-58716082 on Build GRCh38
rs542008166 Chr.10: 58715913-58715913 on Build GRCh38
rs541126851 Chr.10: 58715837-58715837 on Build GRCh38
rs182913687 Chr.10: 58715927-58715927 on Build GRCh38
rs190292184 Chr.10: 58716192-58716192 on Build GRCh38
rs991216568 Chr.10: 58715859-58715859 on Build GRCh38
rs920879383 Chr.10: 58716236-58716236 on Build GRCh38
rs867085179 Chr.10: 58716191-58716191 on Build GRCh38
rs757511523 Chr.10: 58716261-58716261 on Build GRCh38
rs770196634 Chr.10: 58716090-58716091 on Build GRCh38
rs761813901 Chr.10: 58716235-58716235 on Build GRCh38
rs903028448 Chr.10: 58716013-58716013 on Build GRCh38
rs574419056 Chr.10: 58716290-58716290 on Build GRCh38
rs569971257 Chr.10: 58716133-58716133 on Build GRCh38
rs867921892 Chr.10: 58716243-58716245 on Build GRCh38
rs755885844 Chr.10: 58716071-58716071 on Build GRCh38
rs553105627 Chr.10: 58716289-58716289 on Build GRCh38
rs916995286 Chr.10: 58715874-58715874 on Build GRCh38
rs191391408 Chr.10: 58715909-58715909 on Build GRCh38
rs148641218 Chr.10: 58715899-58715899 on Build GRCh38
rs868511553 Chr.10: 58716077-58716077 on Build GRCh38
rs971146362 Chr.10: 58716108-58716108 on Build GRCh38
rs546699148 Chr.10: 58715985-58715985 on Build GRCh38
rs962204526 Chr.10: 58716183-58716183 on Build GRCh38
rs970864318 Chr.10: 58715885-58715885 on Build GRCh38
rs761491238 Chr.10: 58716164-58716164 on Build GRCh38
rs561800448 Chr.10: 58715990-58715990 on Build GRCh38
rs536058614 Chr.10: 58716328-58716328 on Build GRCh38
rs546212588 Chr.10: 58715951-58715951 on Build GRCh38
rs751095982 Chr.10: 58715831-58715834 on Build GRCh38
rs931997943 Chr.10: 58716296-58716296 on Build GRCh38

View Full Product Details