Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9421623-9422118 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX12P
Entrez Gene ID
440081
Gene Name
DEAD/H-box helicase 12, pseudogene
Gene Aliases
CHLR2, DDX12
UniGene
Hs.447869
Gene Chromosome Location
Chr.12: 9417691-9448172 on Build GRCh38
Transcripts
NR_033399.1

SNP Detail

SNP ID Chromosome Location
rs7132464 Chr.12: 9421884-9421884 on Build GRCh38
rs762356384 Chr.12: 9422093-9422093 on Build GRCh38
rs947106875 Chr.12: 9421727-9421727 on Build GRCh38
rs891538797 Chr.12: 9421722-9421722 on Build GRCh38
rs769172931 Chr.12: 9421923-9421923 on Build GRCh38
rs773439284 Chr.12: 9421734-9421734 on Build GRCh38
rs781311835 Chr.12: 9421836-9421836 on Build GRCh38
rs370048070 Chr.12: 9421965-9421965 on Build GRCh38
rs375788846 Chr.12: 9421908-9421908 on Build GRCh38
rs201315779 Chr.12: 9422002-9422002 on Build GRCh38
rs753566679 Chr.12: 9421738-9421738 on Build GRCh38
rs1029183358 Chr.12: 9421826-9421826 on Build GRCh38
rs780826682 Chr.12: 9421709-9421709 on Build GRCh38
rs764852478 Chr.12: 9421696-9421696 on Build GRCh38
rs899140373 Chr.12: 9422049-9422049 on Build GRCh38
rs770136561 Chr.12: 9421854-9421854 on Build GRCh38
rs912244705 Chr.12: 9421684-9421684 on Build GRCh38
rs947159189 Chr.12: 9421911-9421911 on Build GRCh38
rs755167423 Chr.12: 9421838-9421838 on Build GRCh38
rs1015048047 Chr.12: 9421796-9421796 on Build GRCh38
rs994934694 Chr.12: 9422066-9422066 on Build GRCh38
rs1040929741 Chr.12: 9421699-9421699 on Build GRCh38
rs200195883 Chr.12: 9422044-9422044 on Build GRCh38
rs912862568 Chr.12: 9421888-9421888 on Build GRCh38
rs763360564 Chr.12: 9421898-9421898 on Build GRCh38
rs145849154 Chr.12: 9421725-9421725 on Build GRCh38
rs1057429876 Chr.12: 9421989-9421989 on Build GRCh38
rs932223200 Chr.12: 9421653-9421653 on Build GRCh38
rs988153247 Chr.12: 9421840-9421840 on Build GRCh38
rs763893616 Chr.12: 9421747-9421747 on Build GRCh38
rs771944374 Chr.12: 9421813-9421813 on Build GRCh38
rs749947386 Chr.12: 9421679-9421679 on Build GRCh38
rs895723531 Chr.12: 9422000-9422000 on Build GRCh38
rs748394354 Chr.12: 9421859-9421859 on Build GRCh38
rs927157174 Chr.12: 9421967-9421967 on Build GRCh38
rs4237949 Chr.12: 9421748-9421748 on Build GRCh38
rs1004880751 Chr.12: 9421778-9421778 on Build GRCh38
rs994882503 Chr.12: 9421823-9421823 on Build GRCh38
rs1042875263 Chr.12: 9421728-9421728 on Build GRCh38
rs115547270 Chr.12: 9421688-9421688 on Build GRCh38
rs367966567 Chr.12: 9421721-9421721 on Build GRCh38
rs757253758 Chr.12: 9421767-9421767 on Build GRCh38
rs1029065522 Chr.12: 9422088-9422088 on Build GRCh38
rs530206073 Chr.12: 9421642-9421642 on Build GRCh38
rs987715880 Chr.12: 9421657-9421657 on Build GRCh38
rs937170680 Chr.12: 9421987-9421987 on Build GRCh38
rs943700114 Chr.12: 9421708-9421708 on Build GRCh38
rs1035597880 Chr.12: 9421765-9421766 on Build GRCh38

View Full Product Details