Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 18838010-18838558 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A9P
Entrez Gene ID
284232
Gene Name
ankyrin repeat domain 20 family member A9, pseudogene
Gene Aliases
-
UniGene
Hs.679496
Gene Chromosome Location
Chr.13: 18834403-18871969 on Build GRCh38
Transcripts
NR_027995.1

Gene Symbol
RHOT1P3
Entrez Gene ID
100462810
Gene Name
ras homolog family member T1 pseudogene 3
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.13: 18837767-18838262 on Build GRCh38
Transcripts
NR_027995.1

SNP Detail

SNP ID Chromosome Location
rs1018445414 Chr.13: 18838137-18838137 on Build GRCh38
rs368455418 Chr.13: 18838448-18838448 on Build GRCh38
rs555078200 Chr.13: 18838207-18838207 on Build GRCh38
rs527594158 Chr.13: 18838342-18838342 on Build GRCh38
rs201389577 Chr.13: 18838388-18838388 on Build GRCh38
rs899581061 Chr.13: 18838308-18838308 on Build GRCh38
rs974928833 Chr.13: 18838044-18838044 on Build GRCh38
rs1000608065 Chr.13: 18838325-18838325 on Build GRCh38
rs982003655 Chr.13: 18838427-18838427 on Build GRCh38
rs201862525 Chr.13: 18838453-18838453 on Build GRCh38
rs140236254 Chr.13: 18838346-18838346 on Build GRCh38
rs963660400 Chr.13: 18838174-18838174 on Build GRCh38
rs981525592 Chr.13: 18838213-18838213 on Build GRCh38
rs767757772 Chr.13: 18838402-18838402 on Build GRCh38
rs116217864 Chr.13: 18838243-18838243 on Build GRCh38
rs562223671 Chr.13: 18838250-18838250 on Build GRCh38
rs1020974366 Chr.13: 18838398-18838398 on Build GRCh38
rs1038361957 Chr.13: 18838253-18838253 on Build GRCh38
rs146743998 Chr.13: 18838104-18838104 on Build GRCh38
rs571889667 Chr.13: 18838146-18838146 on Build GRCh38
rs200627073 Chr.13: 18838431-18838431 on Build GRCh38
rs78942802 Chr.13: 18838374-18838374 on Build GRCh38
rs957613559 Chr.13: 18838189-18838189 on Build GRCh38
rs549106004 Chr.13: 18838110-18838110 on Build GRCh38
rs926227373 Chr.13: 18838045-18838045 on Build GRCh38
rs536906642 Chr.13: 18838206-18838206 on Build GRCh38
rs58329008 Chr.13: 18838260-18838260 on Build GRCh38
rs117857839 Chr.13: 18838176-18838176 on Build GRCh38
rs201657853 Chr.13: 18838376-18838376 on Build GRCh38
rs927374418 Chr.13: 18838214-18838214 on Build GRCh38
rs1008499402 Chr.13: 18838263-18838263 on Build GRCh38
rs1031990480 Chr.13: 18838177-18838177 on Build GRCh38
rs532935008 Chr.13: 18838100-18838100 on Build GRCh38
rs765885937 Chr.13: 18838082-18838082 on Build GRCh38
rs561138822 Chr.13: 18838473-18838473 on Build GRCh38
rs527725008 Chr.13: 18838108-18838108 on Build GRCh38
rs776457495 Chr.13: 18838244-18838244 on Build GRCh38
rs74204215 Chr.13: 18838125-18838125 on Build GRCh38
rs562359397 Chr.13: 18838098-18838098 on Build GRCh38
rs573546879 Chr.13: 18838224-18838224 on Build GRCh38
rs537214274 Chr.13: 18838102-18838102 on Build GRCh38
rs531701422 Chr.13: 18838526-18838526 on Build GRCh38
rs577447557 Chr.13: 18838257-18838257 on Build GRCh38
rs537960722 Chr.13: 18838124-18838124 on Build GRCh38
rs199605028 Chr.13: 18838409-18838409 on Build GRCh38
rs200151111 Chr.13: 18838482-18838482 on Build GRCh38
rs572649466 Chr.13: 18838196-18838196 on Build GRCh38
rs767080433 Chr.13: 18838256-18838256 on Build GRCh38
rs554548717 Chr.13: 18838195-18838195 on Build GRCh38
rs544255290 Chr.13: 18838086-18838086 on Build GRCh38
rs919964830 Chr.13: 18838254-18838254 on Build GRCh38
rs113720754 Chr.13: 18838264-18838264 on Build GRCh38
rs551291437 Chr.13: 18838103-18838103 on Build GRCh38
rs113702118 Chr.13: 18838120-18838120 on Build GRCh38
rs1034539126 Chr.13: 18838442-18838442 on Build GRCh38
rs561870669 Chr.13: 18838081-18838081 on Build GRCh38
rs958777063 Chr.13: 18838522-18838522 on Build GRCh38
rs994109489 Chr.13: 18838111-18838111 on Build GRCh38
rs879066896 Chr.13: 18838205-18838205 on Build GRCh38
rs528985822 Chr.13: 18838085-18838085 on Build GRCh38
rs1031958279 Chr.13: 18838333-18838333 on Build GRCh38
rs974189017 Chr.13: 18838535-18838535 on Build GRCh38

View Full Product Details