Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 18841783-18842377 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD20A9P
Entrez Gene ID
284232
Gene Name
ankyrin repeat domain 20 family member A9, pseudogene
Gene Aliases
-
UniGene
Hs.679496
Gene Chromosome Location
Chr.13: 18834403-18871969 on Build GRCh38
Transcripts
NR_027995.1

SNP Detail

SNP ID Chromosome Location
rs150810239 Chr.13: 18842216-18842216 on Build GRCh38
rs2801696 Chr.13: 18841895-18841895 on Build GRCh38
rs564009621 Chr.13: 18842096-18842096 on Build GRCh38
rs200993405 Chr.13: 18842014-18842014 on Build GRCh38
rs993449099 Chr.13: 18841876-18841876 on Build GRCh38
rs989769877 Chr.13: 18842247-18842247 on Build GRCh38
rs868857805 Chr.13: 18842087-18842087 on Build GRCh38
rs1045252122 Chr.13: 18841825-18841825 on Build GRCh38
rs992492179 Chr.13: 18842068-18842068 on Build GRCh38
rs568001154 Chr.13: 18842031-18842031 on Build GRCh38
rs56186991 Chr.13: 18842080-18842080 on Build GRCh38
rs796964336 Chr.13: 18842021-18842021 on Build GRCh38
rs1038307724 Chr.13: 18842113-18842113 on Build GRCh38
rs146316569 Chr.13: 18842258-18842258 on Build GRCh38
rs142620654 Chr.13: 18842264-18842264 on Build GRCh38
rs1030325906 Chr.13: 18841923-18841923 on Build GRCh38
rs868644473 Chr.13: 18842229-18842229 on Build GRCh38
rs376322491 Chr.13: 18842324-18842324 on Build GRCh38
rs1007406408 Chr.13: 18842211-18842211 on Build GRCh38
rs905337463 Chr.13: 18842091-18842091 on Build GRCh38
rs574256306 Chr.13: 18841850-18841850 on Build GRCh38
rs1017664011 Chr.13: 18842210-18842210 on Build GRCh38
rs796664214 Chr.13: 18842216-18842216 on Build GRCh38
rs1030253765 Chr.13: 18842177-18842177 on Build GRCh38
rs149021813 Chr.13: 18842201-18842201 on Build GRCh38
rs550433883 Chr.13: 18842334-18842334 on Build GRCh38
rs545940106 Chr.13: 18842077-18842077 on Build GRCh38
rs75330701 Chr.13: 18842330-18842330 on Build GRCh38
rs544877195 Chr.13: 18841941-18841941 on Build GRCh38
rs189887243 Chr.13: 18842351-18842351 on Build GRCh38
rs376339535 Chr.13: 18841937-18841937 on Build GRCh38
rs905369361 Chr.13: 18841834-18841838 on Build GRCh38
rs942227933 Chr.13: 18842105-18842105 on Build GRCh38
rs2801695 Chr.13: 18841884-18841884 on Build GRCh38
rs897905031 Chr.13: 18842162-18842162 on Build GRCh38
rs141958605 Chr.13: 18842239-18842239 on Build GRCh38
rs934429956 Chr.13: 18842064-18842064 on Build GRCh38
rs568200112 Chr.13: 18842265-18842265 on Build GRCh38
rs2801694 Chr.13: 18841822-18841822 on Build GRCh38
rs916954552 Chr.13: 18842072-18842073 on Build GRCh38
rs765497633 Chr.13: 18842210-18842211 on Build GRCh38
rs143437651 Chr.13: 18842178-18842178 on Build GRCh38
rs146736259 Chr.13: 18842183-18842183 on Build GRCh38
rs1017352181 Chr.13: 18842000-18842000 on Build GRCh38
rs978889849 Chr.13: 18842034-18842034 on Build GRCh38
rs529167448 Chr.13: 18842277-18842277 on Build GRCh38
rs957994104 Chr.13: 18842235-18842235 on Build GRCh38
rs66871960 Chr.13: 18842234-18842235 on Build GRCh38
rs1045344823 Chr.13: 18842079-18842079 on Build GRCh38
rs978599380 Chr.13: 18842350-18842350 on Build GRCh38
rs372758938 Chr.13: 18842262-18842262 on Build GRCh38
rs200427971 Chr.13: 18842216-18842217 on Build GRCh38
rs1006680809 Chr.13: 18841836-18841836 on Build GRCh38
rs59800622 Chr.13: 18842036-18842036 on Build GRCh38
rs373220584 Chr.13: 18842028-18842028 on Build GRCh38
rs1016370158 Chr.13: 18841838-18841838 on Build GRCh38
rs4312165 Chr.13: 18842165-18842165 on Build GRCh38
rs4037790 Chr.13: 18841840-18841840 on Build GRCh38
rs986017466 Chr.13: 18841956-18841956 on Build GRCh38
rs750679738 Chr.13: 18842269-18842269 on Build GRCh38

View Full Product Details