Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 56617334-56617877 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MT1L
Entrez Gene ID
4500
Gene Name
metallothionein 1L (gene/pseudogene)
Gene Aliases
MT1, MT1R, MTF
UniGene
Hs.647358
Gene Chromosome Location
Chr.16: 56617461-56618818 on Build GRCh38
Transcripts
NR_001447.2

SNP Detail

SNP ID Chromosome Location
rs4784697 Chr.16: 56617372-56617372 on Build GRCh38
rs758279645 Chr.16: 56617850-56617850 on Build GRCh38
rs1036942786 Chr.16: 56617623-56617623 on Build GRCh38
rs909532956 Chr.16: 56617589-56617589 on Build GRCh38
rs893792756 Chr.16: 56617815-56617815 on Build GRCh38
rs1025477301 Chr.16: 56617445-56617445 on Build GRCh38
rs988332255 Chr.16: 56617467-56617467 on Build GRCh38
rs61749563 Chr.16: 56617462-56617462 on Build GRCh38
rs1032779039 Chr.16: 56617765-56617765 on Build GRCh38
rs138738594 Chr.16: 56617757-56617757 on Build GRCh38
rs969418362 Chr.16: 56617447-56617447 on Build GRCh38
rs572958057 Chr.16: 56617710-56617710 on Build GRCh38
rs761672205 Chr.16: 56617455-56617455 on Build GRCh38
rs965508179 Chr.16: 56617506-56617506 on Build GRCh38
rs369404838 Chr.16: 56617579-56617579 on Build GRCh38
rs1048317258 Chr.16: 56617353-56617353 on Build GRCh38
rs757773732 Chr.16: 56617816-56617816 on Build GRCh38
rs540355628 Chr.16: 56617714-56617714 on Build GRCh38
rs533318264 Chr.16: 56617514-56617514 on Build GRCh38
rs527816068 Chr.16: 56617530-56617530 on Build GRCh38
rs80248054 Chr.16: 56617544-56617544 on Build GRCh38
rs1009805648 Chr.16: 56617826-56617826 on Build GRCh38
rs115091016 Chr.16: 56617715-56617715 on Build GRCh38
rs1013711207 Chr.16: 56617406-56617406 on Build GRCh38
rs545985553 Chr.16: 56617367-56617367 on Build GRCh38
rs533723028 Chr.16: 56617763-56617763 on Build GRCh38
rs561699414 Chr.16: 56617448-56617448 on Build GRCh38
rs878868562 Chr.16: 56617543-56617543 on Build GRCh38
rs545257843 Chr.16: 56617774-56617774 on Build GRCh38
rs538524915 Chr.16: 56617584-56617584 on Build GRCh38
rs572939058 Chr.16: 56617388-56617388 on Build GRCh38
rs4783946 Chr.16: 56617397-56617397 on Build GRCh38
rs566843697 Chr.16: 56617520-56617520 on Build GRCh38
rs544892421 Chr.16: 56617487-56617487 on Build GRCh38
rs904773 Chr.16: 56617689-56617689 on Build GRCh38
rs539251527 Chr.16: 56617690-56617690 on Build GRCh38
rs1021106007 Chr.16: 56617476-56617476 on Build GRCh38
rs563212287 Chr.16: 56617488-56617488 on Build GRCh38
rs563036858 Chr.16: 56617759-56617759 on Build GRCh38
rs773042844 Chr.16: 56617486-56617486 on Build GRCh38
rs556939173 Chr.16: 56617648-56617648 on Build GRCh38
rs7499355 Chr.16: 56617609-56617609 on Build GRCh38
rs1032608282 Chr.16: 56617456-56617456 on Build GRCh38
rs557971043 Chr.16: 56617694-56617694 on Build GRCh38
rs888333114 Chr.16: 56617359-56617359 on Build GRCh38
rs941324238 Chr.16: 56617357-56617357 on Build GRCh38
rs146322755 Chr.16: 56617753-56617753 on Build GRCh38
rs551998174 Chr.16: 56617519-56617519 on Build GRCh38

View Full Product Details