Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 20400216-20400767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC144CP
Entrez Gene ID
348254
Gene Name
coiled-coil domain containing 144C, pseudogene
Gene Aliases
CCDC144C
UniGene
Hs.462475
Gene Chromosome Location
Chr.17: 20321174-20402191 on Build GRCh38
Transcripts
NR_023380.1

SNP Detail

SNP ID Chromosome Location
rs759170318 Chr.17: 20400597-20400597 on Build GRCh38
rs1014220537 Chr.17: 20400248-20400248 on Build GRCh38
rs115465086 Chr.17: 20400736-20400736 on Build GRCh38
rs1056288814 Chr.17: 20400492-20400492 on Build GRCh38
rs967295388 Chr.17: 20400526-20400526 on Build GRCh38
rs201756897 Chr.17: 20400514-20400514 on Build GRCh38
rs985949314 Chr.17: 20400680-20400680 on Build GRCh38
rs188209556 Chr.17: 20400734-20400734 on Build GRCh38
rs556014003 Chr.17: 20400445-20400445 on Build GRCh38
rs967242975 Chr.17: 20400274-20400274 on Build GRCh38
rs930196078 Chr.17: 20400383-20400383 on Build GRCh38
rs574397725 Chr.17: 20400500-20400500 on Build GRCh38
rs888612224 Chr.17: 20400400-20400400 on Build GRCh38
rs554129378 Chr.17: 20400544-20400544 on Build GRCh38
rs1032847232 Chr.17: 20400560-20400560 on Build GRCh38
rs572375918 Chr.17: 20400568-20400568 on Build GRCh38
rs558397939 Chr.17: 20400344-20400344 on Build GRCh38
rs753024576 Chr.17: 20400593-20400594 on Build GRCh38
rs986281836 Chr.17: 20400321-20400321 on Build GRCh38
rs183925469 Chr.17: 20400368-20400368 on Build GRCh38
rs546260841 Chr.17: 20400645-20400645 on Build GRCh38
rs533883740 Chr.17: 20400339-20400339 on Build GRCh38
rs1036001266 Chr.17: 20400381-20400381 on Build GRCh38
rs74699367 Chr.17: 20400517-20400517 on Build GRCh38
rs953167024 Chr.17: 20400303-20400303 on Build GRCh38
rs925774958 Chr.17: 20400279-20400279 on Build GRCh38
rs564584494 Chr.17: 20400715-20400715 on Build GRCh38
rs753296809 Chr.17: 20400579-20400579 on Build GRCh38
rs2694311 Chr.17: 20400242-20400242 on Build GRCh38
rs1048624621 Chr.17: 20400385-20400385 on Build GRCh38
rs895868904 Chr.17: 20400496-20400496 on Build GRCh38
rs911644767 Chr.17: 20400699-20400699 on Build GRCh38
rs191741236 Chr.17: 20400367-20400367 on Build GRCh38
rs1001661491 Chr.17: 20400470-20400470 on Build GRCh38
rs978588678 Chr.17: 20400278-20400278 on Build GRCh38
rs764820819 Chr.17: 20400608-20400608 on Build GRCh38
rs540540667 Chr.17: 20400625-20400625 on Build GRCh38
rs1020632752 Chr.17: 20400269-20400269 on Build GRCh38
rs918862060 Chr.17: 20400382-20400382 on Build GRCh38
rs535443144 Chr.17: 20400519-20400519 on Build GRCh38
rs766057101 Chr.17: 20400557-20400557 on Build GRCh38

View Full Product Details