Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 32150598-32151149 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARGFXP2
Entrez Gene ID
503640
Gene Name
arginine-fifty homeobox pseudogene 2
Gene Aliases
-
UniGene
Hs.631752
Gene Chromosome Location
Chr.17: 32150357-32151571 on Build GRCh38
Transcripts
NR_002222.1 NM_001033566.2 NM_001033567.2 NM_001033568.2 NM_001288754.1 NM_001288755.1 NM_001288758.1 NM_018307.4 NR_110083.1 XM_011524971.2 XM_011524973.2 XM_011524975.2 XM_011524976.2 XM_017024819.1 XM_017024821.1 XM_017024823.1 XM_017024824.1 XM_017024825.1

Gene Symbol
RHOT1
Entrez Gene ID
55288
Gene Name
ras homolog family member T1
Gene Aliases
ARHT1, MIRO-1, MIRO1
UniGene
Hs.655325
Gene Chromosome Location
Chr.17: 32142454-32253374 on Build GRCh38
Transcripts
NR_002222.1 NM_001033566.2 NM_001033567.2 NM_001033568.2 NM_001288754.1 NM_001288755.1 NM_001288758.1 NM_018307.4 NR_110083.1 XM_011524971.2 XM_011524973.2 XM_011524975.2 XM_011524976.2 XM_017024819.1 XM_017024821.1 XM_017024823.1 XM_017024824.1 XM_017024825.1

SNP Detail

SNP ID Chromosome Location
rs776312686 Chr.17: 32150995-32150995 on Build GRCh38
rs752805778 Chr.17: 32150838-32150838 on Build GRCh38
rs73989707 Chr.17: 32150662-32150662 on Build GRCh38
rs112682767 Chr.17: 32150959-32150960 on Build GRCh38
rs71144809 Chr.17: 32150983-32150984 on Build GRCh38
rs944789669 Chr.17: 32150867-32150867 on Build GRCh38
rs566985828 Chr.17: 32150935-32150935 on Build GRCh38
rs550953941 Chr.17: 32150804-32150804 on Build GRCh38
rs534327146 Chr.17: 32150808-32150808 on Build GRCh38
rs993587694 Chr.17: 32151025-32151025 on Build GRCh38
rs913946419 Chr.17: 32151112-32151112 on Build GRCh38
rs12940465 Chr.17: 32150826-32150826 on Build GRCh38
rs547294445 Chr.17: 32150711-32150711 on Build GRCh38
rs1041573340 Chr.17: 32150898-32150898 on Build GRCh38
rs573805129 Chr.17: 32150846-32150846 on Build GRCh38
rs1017874179 Chr.17: 32151105-32151105 on Build GRCh38
rs972218188 Chr.17: 32150809-32150809 on Build GRCh38
rs778886706 Chr.17: 32150961-32150961 on Build GRCh38
rs952301104 Chr.17: 32150814-32150814 on Build GRCh38
rs146748999 Chr.17: 32150821-32150821 on Build GRCh38
rs150449021 Chr.17: 32150963-32150964 on Build GRCh38
rs181804010 Chr.17: 32150906-32150906 on Build GRCh38
rs9906736 Chr.17: 32150845-32150845 on Build GRCh38
rs945481925 Chr.17: 32151118-32151118 on Build GRCh38
rs779830274 Chr.17: 32150904-32150904 on Build GRCh38
rs73284185 Chr.17: 32150874-32150874 on Build GRCh38
rs567020805 Chr.17: 32150796-32150796 on Build GRCh38
rs1013942507 Chr.17: 32150959-32150964 on Build GRCh38
rs1027819771 Chr.17: 32151028-32151028 on Build GRCh38
rs767912606 Chr.17: 32150716-32150716 on Build GRCh38
rs976549045 Chr.17: 32150873-32150873 on Build GRCh38
rs976114683 Chr.17: 32151111-32151111 on Build GRCh38
rs745841877 Chr.17: 32150877-32150877 on Build GRCh38
rs999824248 Chr.17: 32150630-32150630 on Build GRCh38
rs114164052 Chr.17: 32151080-32151080 on Build GRCh38
rs567765880 Chr.17: 32150813-32150813 on Build GRCh38
rs34323066 Chr.17: 32150982-32150983 on Build GRCh38
rs536791107 Chr.17: 32150879-32150879 on Build GRCh38
rs373565814 Chr.17: 32151071-32151071 on Build GRCh38
rs1023982547 Chr.17: 32150959-32150967 on Build GRCh38
rs966411410 Chr.17: 32151108-32151108 on Build GRCh38
rs755280966 Chr.17: 32150961-32150962 on Build GRCh38
rs768282155 Chr.17: 32150929-32150929 on Build GRCh38
rs71362803 Chr.17: 32150958-32150959 on Build GRCh38
rs958476065 Chr.17: 32150667-32150667 on Build GRCh38
rs765829884 Chr.17: 32150959-32150961 on Build GRCh38

View Full Product Details