Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 57046607-57046980 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF126P1
Entrez Gene ID
376412
Gene Name
ring finger protein 126 pseudogene 1
Gene Aliases
-
UniGene
Hs.132299
Gene Chromosome Location
Chr.17: 57045478-57046795 on Build GRCh38
Transcripts
NR_002818.2

SNP Detail

SNP ID Chromosome Location
rs533628278 Chr.17: 57046913-57046913 on Build GRCh38
rs184944762 Chr.17: 57046827-57046827 on Build GRCh38
rs1051854320 Chr.17: 57046787-57046787 on Build GRCh38
rs942195223 Chr.17: 57046684-57046684 on Build GRCh38
rs1036968346 Chr.17: 57046690-57046690 on Build GRCh38
rs991836771 Chr.17: 57046861-57046861 on Build GRCh38
rs189732420 Chr.17: 57046870-57046870 on Build GRCh38
rs987796579 Chr.17: 57046816-57046816 on Build GRCh38
rs1047091901 Chr.17: 57046895-57046895 on Build GRCh38
rs143250361 Chr.17: 57046757-57046757 on Build GRCh38
rs557033302 Chr.17: 57046759-57046759 on Build GRCh38
rs899231543 Chr.17: 57046718-57046718 on Build GRCh38
rs577037633 Chr.17: 57046683-57046683 on Build GRCh38
rs761872431 Chr.17: 57046878-57046878 on Build GRCh38
rs991939264 Chr.17: 57046627-57046627 on Build GRCh38
rs575734198 Chr.17: 57046825-57046825 on Build GRCh38
rs558831974 Chr.17: 57046628-57046628 on Build GRCh38
rs371003367 Chr.17: 57046916-57046916 on Build GRCh38
rs969024163 Chr.17: 57046876-57046876 on Build GRCh38
rs947731856 Chr.17: 57046635-57046635 on Build GRCh38
rs899283956 Chr.17: 57046938-57046938 on Build GRCh38
rs563050212 Chr.17: 57046802-57046802 on Build GRCh38
rs368258315 Chr.17: 57046715-57046715 on Build GRCh38
rs532653360 Chr.17: 57046932-57046932 on Build GRCh38
rs1031227327 Chr.17: 57046839-57046839 on Build GRCh38
rs1037696722 Chr.17: 57046920-57046920 on Build GRCh38
rs956569043 Chr.17: 57046852-57046852 on Build GRCh38
rs372771190 Chr.17: 57046834-57046834 on Build GRCh38
rs993579645 Chr.17: 57046945-57046945 on Build GRCh38
rs1045282265 Chr.17: 57046668-57046668 on Build GRCh38
rs914068320 Chr.17: 57046857-57046857 on Build GRCh38
rs938036530 Chr.17: 57046906-57046906 on Build GRCh38
rs140217249 Chr.17: 57046903-57046903 on Build GRCh38
rs151176209 Chr.17: 57046838-57046838 on Build GRCh38

View Full Product Details