Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 63859774-63860318 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TCAM1P
Entrez Gene ID
146771
Gene Name
testicular cell adhesion molecule 1, pseudogene
Gene Aliases
TCAM1
UniGene
Hs.629177
Gene Chromosome Location
Chr.17: 63857016-63864379 on Build GRCh38
Transcripts
NR_002947.2

SNP Detail

SNP ID Chromosome Location
rs1003764952 Chr.17: 63860039-63860039 on Build GRCh38
rs773256032 Chr.17: 63859902-63859902 on Build GRCh38
rs370325711 Chr.17: 63860228-63860228 on Build GRCh38
rs770046103 Chr.17: 63859901-63859901 on Build GRCh38
rs948508776 Chr.17: 63860246-63860246 on Build GRCh38
rs942677125 Chr.17: 63859952-63859952 on Build GRCh38
rs766865996 Chr.17: 63860063-63860063 on Build GRCh38
rs75310753 Chr.17: 63859837-63859837 on Build GRCh38
rs532502856 Chr.17: 63860226-63860226 on Build GRCh38
rs376386470 Chr.17: 63860083-63860083 on Build GRCh38
rs984330492 Chr.17: 63860284-63860284 on Build GRCh38
rs1015594180 Chr.17: 63860044-63860045 on Build GRCh38
rs73337478 Chr.17: 63860074-63860074 on Build GRCh38
rs144651940 Chr.17: 63860213-63860213 on Build GRCh38
rs966440945 Chr.17: 63860207-63860207 on Build GRCh38
rs191038457 Chr.17: 63860216-63860216 on Build GRCh38
rs761292977 Chr.17: 63860184-63860184 on Build GRCh38
rs951561917 Chr.17: 63860273-63860273 on Build GRCh38
rs2156903 Chr.17: 63859819-63859819 on Build GRCh38
rs180982878 Chr.17: 63860028-63860028 on Build GRCh38
rs992993345 Chr.17: 63859828-63859828 on Build GRCh38
rs375224660 Chr.17: 63860241-63860241 on Build GRCh38
rs897972076 Chr.17: 63860062-63860062 on Build GRCh38
rs1001147227 Chr.17: 63860208-63860208 on Build GRCh38
rs774722137 Chr.17: 63860127-63860127 on Build GRCh38
rs930151106 Chr.17: 63859860-63859860 on Build GRCh38
rs1018912356 Chr.17: 63860104-63860104 on Build GRCh38
rs539444038 Chr.17: 63860136-63860136 on Build GRCh38
rs1054102643 Chr.17: 63859890-63859890 on Build GRCh38
rs986122122 Chr.17: 63860101-63860101 on Build GRCh38
rs572567077 Chr.17: 63860288-63860288 on Build GRCh38
rs763753315 Chr.17: 63860209-63860209 on Build GRCh38
rs186445834 Chr.17: 63860158-63860158 on Build GRCh38
rs901182714 Chr.17: 63860007-63860007 on Build GRCh38
rs948373922 Chr.17: 63859943-63859943 on Build GRCh38
rs910128508 Chr.17: 63860287-63860287 on Build GRCh38
rs1021478978 Chr.17: 63860182-63860182 on Build GRCh38
rs78679940 Chr.17: 63859927-63859927 on Build GRCh38
rs904487653 Chr.17: 63860192-63860192 on Build GRCh38
rs1039761775 Chr.17: 63860002-63860002 on Build GRCh38
rs763045313 Chr.17: 63859914-63859914 on Build GRCh38

View Full Product Details