Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 2655001-2655546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CBX3P2
Entrez Gene ID
645158
Gene Name
chromobox 3 pseudogene 2
Gene Aliases
-
UniGene
Hs.712861
Gene Chromosome Location
Chr.18: 2652170-2655395 on Build GRCh38
Transcripts
NR_033754.2

SNP Detail

SNP ID Chromosome Location
rs1018946961 Chr.18: 2655489-2655489 on Build GRCh38
rs12965596 Chr.18: 2655483-2655483 on Build GRCh38
rs929661678 Chr.18: 2655128-2655128 on Build GRCh38
rs904166247 Chr.18: 2655166-2655166 on Build GRCh38
rs888195278 Chr.18: 2655252-2655252 on Build GRCh38
rs1007656113 Chr.18: 2655467-2655467 on Build GRCh38
rs916833780 Chr.18: 2655075-2655075 on Build GRCh38
rs1047192547 Chr.18: 2655250-2655250 on Build GRCh38
rs763804620 Chr.18: 2655113-2655113 on Build GRCh38
rs945923051 Chr.18: 2655265-2655265 on Build GRCh38
rs182982490 Chr.18: 2655230-2655230 on Build GRCh38
rs112937381 Chr.18: 2655208-2655208 on Build GRCh38
rs1039833522 Chr.18: 2655519-2655519 on Build GRCh38
rs888221881 Chr.18: 2655131-2655131 on Build GRCh38
rs766440556 Chr.18: 2655053-2655053 on Build GRCh38
rs994986218 Chr.18: 2655394-2655395 on Build GRCh38
rs1029574355 Chr.18: 2655316-2655316 on Build GRCh38
rs866358499 Chr.18: 2655433-2655433 on Build GRCh38
rs541062284 Chr.18: 2655493-2655493 on Build GRCh38
rs998033742 Chr.18: 2655303-2655303 on Build GRCh38
rs953430388 Chr.18: 2655129-2655129 on Build GRCh38
rs948338611 Chr.18: 2655106-2655106 on Build GRCh38
rs1046637911 Chr.18: 2655110-2655110 on Build GRCh38
rs924036397 Chr.18: 2655037-2655037 on Build GRCh38
rs749994816 Chr.18: 2655217-2655217 on Build GRCh38
rs760599197 Chr.18: 2655092-2655092 on Build GRCh38
rs930260611 Chr.18: 2655475-2655475 on Build GRCh38
rs1031451522 Chr.18: 2655361-2655361 on Build GRCh38
rs1042850014 Chr.18: 2655154-2655154 on Build GRCh38
rs186508707 Chr.18: 2655197-2655197 on Build GRCh38
rs141381487 Chr.18: 2655385-2655385 on Build GRCh38
rs958399617 Chr.18: 2655212-2655212 on Build GRCh38
rs983062639 Chr.18: 2655494-2655494 on Build GRCh38
rs929630630 Chr.18: 2655235-2655235 on Build GRCh38
rs998064724 Chr.18: 2655177-2655177 on Build GRCh38
rs28601731 Chr.18: 2655071-2655071 on Build GRCh38
rs1046780758 Chr.18: 2655130-2655130 on Build GRCh38
rs965254451 Chr.18: 2655236-2655236 on Build GRCh38
rs943954486 Chr.18: 2655136-2655136 on Build GRCh38
rs913071258 Chr.18: 2655138-2655139 on Build GRCh38
rs766301680 Chr.18: 2655129-2655130 on Build GRCh38
rs973547891 Chr.18: 2655102-2655103 on Build GRCh38
rs1040860157 Chr.18: 2655137-2655137 on Build GRCh38
rs989653988 Chr.18: 2655214-2655214 on Build GRCh38
rs976697519 Chr.18: 2655248-2655248 on Build GRCh38
rs535233869 Chr.18: 2655119-2655119 on Build GRCh38
rs1005365512 Chr.18: 2655132-2655132 on Build GRCh38
rs999532303 Chr.18: 2655352-2655352 on Build GRCh38
rs563923873 Chr.18: 2655514-2655514 on Build GRCh38
rs867418538 Chr.18: 2655226-2655226 on Build GRCh38
rs953599572 Chr.18: 2655223-2655223 on Build GRCh38
rs928200664 Chr.18: 2655413-2655413 on Build GRCh38
rs753685379 Chr.18: 2655135-2655135 on Build GRCh38
rs1029604130 Chr.18: 2655184-2655184 on Build GRCh38
rs557856390 Chr.18: 2655216-2655216 on Build GRCh38
rs549612377 Chr.18: 2655059-2655059 on Build GRCh38
rs895520658 Chr.18: 2655369-2655369 on Build GRCh38
rs989930334 Chr.18: 2655387-2655387 on Build GRCh38
rs543214635 Chr.18: 2655330-2655330 on Build GRCh38
rs189719526 Chr.18: 2655210-2655210 on Build GRCh38
rs1055407400 Chr.18: 2655173-2655173 on Build GRCh38
rs941073169 Chr.18: 2655257-2655257 on Build GRCh38
rs181775501 Chr.18: 2655164-2655164 on Build GRCh38
rs1015051576 Chr.18: 2655036-2655036 on Build GRCh38
rs186262104 Chr.18: 2655251-2655251 on Build GRCh38
rs560115734 Chr.18: 2655383-2655383 on Build GRCh38
rs867348163 Chr.18: 2655336-2655336 on Build GRCh38
rs563838572 Chr.18: 2655280-2655280 on Build GRCh38
rs982273605 Chr.18: 2655412-2655412 on Build GRCh38
rs921044840 Chr.18: 2655526-2655526 on Build GRCh38
rs969498926 Chr.18: 2655401-2655401 on Build GRCh38
rs781274887 Chr.18: 2655392-2655392 on Build GRCh38
rs945776834 Chr.18: 2655140-2655140 on Build GRCh38
rs899645662 Chr.18: 2655293-2655293 on Build GRCh38
rs982304832 Chr.18: 2655224-2655224 on Build GRCh38
rs1055355555 Chr.18: 2655344-2655344 on Build GRCh38
rs909499632 Chr.18: 2655504-2655504 on Build GRCh38
rs936844055 Chr.18: 2655038-2655038 on Build GRCh38
rs73380268 Chr.18: 2655281-2655281 on Build GRCh38
rs954666951 Chr.18: 2655194-2655194 on Build GRCh38

View Full Product Details