Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 21381468-21382016 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZNF738
Entrez Gene ID
148203
Gene Name
zinc finger protein 738
Gene Aliases
-
UniGene
Hs.124384
Gene Chromosome Location
Chr.19: 21358933-21388582 on Build GRCh38
Transcripts
NR_027130.1

SNP Detail

SNP ID Chromosome Location
rs769682568 Chr.19: 21381618-21381618 on Build GRCh38
rs569905566 Chr.19: 21381501-21381502 on Build GRCh38
rs113012822 Chr.19: 21381646-21381646 on Build GRCh38
rs560391503 Chr.19: 21381567-21381567 on Build GRCh38
rs369235226 Chr.19: 21381704-21381704 on Build GRCh38
rs568522265 Chr.19: 21381706-21381706 on Build GRCh38
rs181259075 Chr.19: 21381562-21381562 on Build GRCh38
rs1016328437 Chr.19: 21381683-21381683 on Build GRCh38
rs774436916 Chr.19: 21381961-21381961 on Build GRCh38
rs936824486 Chr.19: 21381852-21381852 on Build GRCh38
rs906487097 Chr.19: 21381895-21381895 on Build GRCh38
rs916292408 Chr.19: 21381863-21381863 on Build GRCh38
rs58140656 Chr.19: 21381696-21381696 on Build GRCh38
rs974738391 Chr.19: 21381715-21381715 on Build GRCh38
rs746465150 Chr.19: 21381516-21381516 on Build GRCh38
rs543891656 Chr.19: 21381941-21381941 on Build GRCh38
rs1053963769 Chr.19: 21381861-21381861 on Build GRCh38
rs148878410 Chr.19: 21381784-21381784 on Build GRCh38
rs527607069 Chr.19: 21381594-21381594 on Build GRCh38
rs994033638 Chr.19: 21381587-21381587 on Build GRCh38
rs762675746 Chr.19: 21381731-21381731 on Build GRCh38
rs552134849 Chr.19: 21381596-21381596 on Build GRCh38
rs554513182 Chr.19: 21381580-21381580 on Build GRCh38
rs576909853 Chr.19: 21381878-21381878 on Build GRCh38
rs746268340 Chr.19: 21381974-21381975 on Build GRCh38
rs994064692 Chr.19: 21381926-21381926 on Build GRCh38
rs902311865 Chr.19: 21381488-21381488 on Build GRCh38
rs947718191 Chr.19: 21381873-21381873 on Build GRCh38
rs1006702067 Chr.19: 21381619-21381619 on Build GRCh38
rs554118295 Chr.19: 21381768-21381768 on Build GRCh38
rs371066169 Chr.19: 21381643-21381643 on Build GRCh38
rs535539780 Chr.19: 21381625-21381640 on Build GRCh38
rs894817039 Chr.19: 21381512-21381512 on Build GRCh38
rs572756378 Chr.19: 21381782-21381782 on Build GRCh38
rs188230335 Chr.19: 21381534-21381534 on Build GRCh38
rs779606903 Chr.19: 21381513-21381513 on Build GRCh38
rs190236894 Chr.19: 21381821-21381821 on Build GRCh38
rs537923820 Chr.19: 21381649-21381649 on Build GRCh38
rs184899336 Chr.19: 21381728-21381728 on Build GRCh38
rs888301638 Chr.19: 21381968-21381969 on Build GRCh38
rs936406398 Chr.19: 21381494-21381494 on Build GRCh38
rs866521212 Chr.19: 21381770-21381770 on Build GRCh38
rs570524901 Chr.19: 21381632-21381632 on Build GRCh38

View Full Product Details