Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 218060957-218061501 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CXCR2P1
Entrez Gene ID
3580
Gene Name
C-X-C motif chemokine receptor 2 pseudogene 1
Gene Aliases
CXCR2P, IL8RBP
UniGene
Hs.647858
Gene Chromosome Location
Chr.2: 218059155-218061290 on Build GRCh38
Transcripts
NR_002712.1 NR_034176.1 XM_011511014.2 XM_017003895.1 XM_017003896.1 XM_017003897.1

Gene Symbol
RUFY4
Entrez Gene ID
285180
Gene Name
RUN and FYVE domain containing 4
Gene Aliases
ZFYVE31
UniGene
Hs.570069
Gene Chromosome Location
Chr.2: 218034934-218090584 on Build GRCh38
Transcripts
NR_002712.1 NR_034176.1 XM_011511014.2 XM_017003895.1 XM_017003896.1 XM_017003897.1

SNP Detail

SNP ID Chromosome Location
rs756757463 Chr.2: 218061003-218061003 on Build GRCh38
rs984898879 Chr.2: 218061094-218061094 on Build GRCh38
rs553037813 Chr.2: 218061232-218061232 on Build GRCh38
rs994777057 Chr.2: 218061140-218061140 on Build GRCh38
rs141412182 Chr.2: 218061448-218061448 on Build GRCh38
rs532801003 Chr.2: 218061447-218061447 on Build GRCh38
rs1025187014 Chr.2: 218061344-218061344 on Build GRCh38
rs767609446 Chr.2: 218061022-218061022 on Build GRCh38
rs569679142 Chr.2: 218061176-218061176 on Build GRCh38
rs1036020728 Chr.2: 218061362-218061362 on Build GRCh38
rs77326308 Chr.2: 218061409-218061409 on Build GRCh38
rs537169858 Chr.2: 218061203-218061203 on Build GRCh38
rs949048417 Chr.2: 218061184-218061184 on Build GRCh38
rs148287403 Chr.2: 218061231-218061231 on Build GRCh38
rs962307589 Chr.2: 218061331-218061331 on Build GRCh38
rs989354172 Chr.2: 218061442-218061442 on Build GRCh38
rs1025608054 Chr.2: 218061087-218061087 on Build GRCh38
rs114610787 Chr.2: 218061023-218061023 on Build GRCh38
rs767688153 Chr.2: 218061039-218061039 on Build GRCh38
rs541725884 Chr.2: 218061299-218061299 on Build GRCh38
rs936389272 Chr.2: 218061419-218061419 on Build GRCh38
rs757490803 Chr.2: 218061211-218061211 on Build GRCh38
rs749614412 Chr.2: 218061027-218061027 on Build GRCh38
rs751376569 Chr.2: 218061011-218061011 on Build GRCh38
rs762017012 Chr.2: 218061037-218061037 on Build GRCh38
rs746283633 Chr.2: 218061308-218061308 on Build GRCh38
rs752504503 Chr.2: 218060986-218060986 on Build GRCh38
rs186314785 Chr.2: 218061219-218061219 on Build GRCh38
rs533596333 Chr.2: 218061212-218061212 on Build GRCh38
rs776130649 Chr.2: 218060994-218060994 on Build GRCh38
rs773319303 Chr.2: 218061047-218061047 on Build GRCh38
rs545926019 Chr.2: 218061349-218061349 on Build GRCh38
rs758296040 Chr.2: 218060996-218060996 on Build GRCh38
rs777514690 Chr.2: 218060998-218060998 on Build GRCh38
rs966194364 Chr.2: 218061017-218061017 on Build GRCh38
rs949717152 Chr.2: 218061345-218061346 on Build GRCh38
rs1016367810 Chr.2: 218061323-218061323 on Build GRCh38
rs1055444101 Chr.2: 218061319-218061319 on Build GRCh38
rs564910044 Chr.2: 218061239-218061239 on Build GRCh38
rs1050372556 Chr.2: 218061472-218061472 on Build GRCh38
rs370341653 Chr.2: 218061342-218061342 on Build GRCh38
rs994411135 Chr.2: 218061435-218061435 on Build GRCh38
rs970819040 Chr.2: 218061399-218061399 on Build GRCh38
rs566361217 Chr.2: 218061028-218061028 on Build GRCh38
rs758264520 Chr.2: 218061015-218061015 on Build GRCh38
rs1029049821 Chr.2: 218061286-218061286 on Build GRCh38
rs563320489 Chr.2: 218061328-218061328 on Build GRCh38
rs1027169897 Chr.2: 218061438-218061439 on Build GRCh38
rs889222222 Chr.2: 218061476-218061476 on Build GRCh38
rs937071961 Chr.2: 218061024-218061024 on Build GRCh38
rs1045218553 Chr.2: 218061201-218061201 on Build GRCh38
rs559828556 Chr.2: 218061462-218061462 on Build GRCh38
rs780752445 Chr.2: 218061004-218061004 on Build GRCh38
rs1041360626 Chr.2: 218061452-218061452 on Build GRCh38
rs772380255 Chr.2: 218061378-218061378 on Build GRCh38
rs776009330 Chr.2: 218061393-218061393 on Build GRCh38
rs980435178 Chr.2: 218061406-218061406 on Build GRCh38
rs558580586 Chr.2: 218061208-218061208 on Build GRCh38
rs536279922 Chr.2: 218061052-218061052 on Build GRCh38
rs997486248 Chr.2: 218061285-218061285 on Build GRCh38
rs945193895 Chr.2: 218061449-218061449 on Build GRCh38
rs962255309 Chr.2: 218061134-218061134 on Build GRCh38
rs777642642 Chr.2: 218061088-218061088 on Build GRCh38
rs925773319 Chr.2: 218061254-218061254 on Build GRCh38
rs754786558 Chr.2: 218061019-218061019 on Build GRCh38
rs544647712 Chr.2: 218061141-218061141 on Build GRCh38
rs181756261 Chr.2: 218061170-218061170 on Build GRCh38
rs765136225 Chr.2: 218060979-218060979 on Build GRCh38
rs889169774 Chr.2: 218061301-218061301 on Build GRCh38
rs909481795 Chr.2: 218061115-218061115 on Build GRCh38
rs78231757 Chr.2: 218061467-218061467 on Build GRCh38
rs774500217 Chr.2: 218061035-218061035 on Build GRCh38
rs556537062 Chr.2: 218061002-218061002 on Build GRCh38

View Full Product Details