Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 16827788-16828352 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSFY1P1
Entrez Gene ID
27437
Gene Name
heat shock transcription factor, Y-linked 1 pseudogene 1
Gene Aliases
CECR8, HSFYL1, HSFYP1, NCRNA00016
UniGene
Hs.143955
Gene Chromosome Location
Chr.22: 16827474-16829335 on Build GRCh38
Transcripts
NR_003607.1

SNP Detail

SNP ID Chromosome Location
rs774636116 Chr.22: 16828066-16828066 on Build GRCh38
rs549216212 Chr.22: 16828151-16828151 on Build GRCh38
rs567661653 Chr.22: 16828321-16828321 on Build GRCh38
rs868275611 Chr.22: 16828192-16828192 on Build GRCh38
rs762267402 Chr.22: 16828238-16828238 on Build GRCh38
rs534887995 Chr.22: 16828325-16828325 on Build GRCh38
rs775697548 Chr.22: 16828247-16828247 on Build GRCh38
rs946906562 Chr.22: 16828230-16828230 on Build GRCh38
rs575565750 Chr.22: 16827857-16827857 on Build GRCh38
rs577940993 Chr.22: 16827945-16827945 on Build GRCh38
rs529677009 Chr.22: 16828136-16828136 on Build GRCh38
rs568797742 Chr.22: 16828076-16828076 on Build GRCh38
rs1026062143 Chr.22: 16828002-16828002 on Build GRCh38
rs964059136 Chr.22: 16828118-16828118 on Build GRCh38
rs561170886 Chr.22: 16827964-16827964 on Build GRCh38
rs914047361 Chr.22: 16828189-16828189 on Build GRCh38
rs1043967116 Chr.22: 16828235-16828235 on Build GRCh38
rs796810509 Chr.22: 16828029-16828029 on Build GRCh38
rs988243345 Chr.22: 16828163-16828163 on Build GRCh38
rs550287791 Chr.22: 16828047-16828047 on Build GRCh38
rs1006108463 Chr.22: 16828065-16828065 on Build GRCh38
rs187426985 Chr.22: 16827980-16827980 on Build GRCh38
rs922607403 Chr.22: 16828129-16828129 on Build GRCh38
rs867195345 Chr.22: 16828248-16828248 on Build GRCh38
rs1017967346 Chr.22: 16828101-16828101 on Build GRCh38
rs564826158 Chr.22: 16828031-16828031 on Build GRCh38
rs546856203 Chr.22: 16827983-16827983 on Build GRCh38
rs975446217 Chr.22: 16828124-16828124 on Build GRCh38
rs763156973 Chr.22: 16827937-16827937 on Build GRCh38
rs748888687 Chr.22: 16827912-16827912 on Build GRCh38
rs35746111 Chr.22: 16828296-16828297 on Build GRCh38
rs532261945 Chr.22: 16828035-16828035 on Build GRCh38

View Full Product Details