Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 38361518-38362038 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC400927
Entrez Gene ID
400927
Gene Name
TPTE and PTEN homologous inositol lipid phosphatase pseudogene
Gene Aliases
-
UniGene
Hs.720223
Gene Chromosome Location
Chr.22: 38344665-38398926 on Build GRCh38
Transcripts
NR_002821.2 NM_001289912.1

Gene Symbol
LOC400927-CSNK1E
Entrez Gene ID
102800317
Gene Name
LOC400927-CSNK1E readthrough
Gene Aliases
-
UniGene
Hs.720223
Gene Chromosome Location
Chr.22: 38290691-38398522 on Build GRCh38
Transcripts
NR_002821.2 NM_001289912.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
86345 c.? p.? Chr.22: 38361518-38361518 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs926576253 Chr.22: 38361910-38361912 on Build GRCh38
rs1038661779 Chr.22: 38361727-38361727 on Build GRCh38
rs926747986 Chr.22: 38361613-38361613 on Build GRCh38
rs78969742 Chr.22: 38361575-38361575 on Build GRCh38
rs890888585 Chr.22: 38361777-38361777 on Build GRCh38
rs62230193 Chr.22: 38361685-38361685 on Build GRCh38
rs981164682 Chr.22: 38361613-38361614 on Build GRCh38
rs71197108 Chr.22: 38361600-38361615 on Build GRCh38
rs908614210 Chr.22: 38361979-38361979 on Build GRCh38
rs575233453 Chr.22: 38361557-38361557 on Build GRCh38
rs996544090 Chr.22: 38361759-38361759 on Build GRCh38
rs201375900 Chr.22: 38361579-38361579 on Build GRCh38
rs898723996 Chr.22: 38361730-38361730 on Build GRCh38
rs572514632 Chr.22: 38361720-38361720 on Build GRCh38
rs62230192 Chr.22: 38361571-38361571 on Build GRCh38
rs970358662 Chr.22: 38361872-38361873 on Build GRCh38
rs908748053 Chr.22: 38361704-38361704 on Build GRCh38
rs1007893894 Chr.22: 38361807-38361807 on Build GRCh38
rs879012230 Chr.22: 38361583-38361583 on Build GRCh38
rs533272740 Chr.22: 38361984-38361984 on Build GRCh38
rs992276645 Chr.22: 38361957-38361957 on Build GRCh38
rs974623002 Chr.22: 38361988-38361988 on Build GRCh38
rs968106675 Chr.22: 38361612-38361612 on Build GRCh38
rs201398545 Chr.22: 38361564-38361579 on Build GRCh38
rs914467567 Chr.22: 38361612-38361613 on Build GRCh38
rs1031382224 Chr.22: 38361818-38361818 on Build GRCh38
rs57483179 Chr.22: 38361762-38361762 on Build GRCh38
rs1021568306 Chr.22: 38361854-38361854 on Build GRCh38
rs960693017 Chr.22: 38361935-38361937 on Build GRCh38
rs62230191 Chr.22: 38361567-38361567 on Build GRCh38
rs554218014 Chr.22: 38361636-38361636 on Build GRCh38
rs146040089 Chr.22: 38361909-38361910 on Build GRCh38
rs1011352930 Chr.22: 38361852-38361852 on Build GRCh38
rs368108707 Chr.22: 38361717-38361717 on Build GRCh38
rs62230190 Chr.22: 38361542-38361542 on Build GRCh38
rs1056646575 Chr.22: 38361643-38361643 on Build GRCh38
rs955908679 Chr.22: 38361833-38361833 on Build GRCh38
rs370697858 Chr.22: 38361563-38361563 on Build GRCh38

View Full Product Details