Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 21586823-21587420 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUSBP1
Entrez Gene ID
728411
Gene Name
glucuronidase, beta pseudogene 1
Gene Aliases
-
UniGene
Hs.88181
Gene Chromosome Location
Chr.5: 21459480-21589372 on Build GRCh38
Transcripts
NR_027026.1

SNP Detail

SNP ID Chromosome Location
rs913076025 Chr.5: 21586984-21586984 on Build GRCh38
rs145898972 Chr.5: 21587382-21587382 on Build GRCh38
rs965857863 Chr.5: 21586900-21586900 on Build GRCh38
rs1017739216 Chr.5: 21586883-21586883 on Build GRCh38
rs1020516242 Chr.5: 21587314-21587314 on Build GRCh38
rs968367132 Chr.5: 21587300-21587300 on Build GRCh38
rs1055422137 Chr.5: 21587156-21587156 on Build GRCh38
rs534189183 Chr.5: 21587393-21587393 on Build GRCh38
rs1045268974 Chr.5: 21587068-21587068 on Build GRCh38
rs1017167027 Chr.5: 21587246-21587246 on Build GRCh38
rs116372584 Chr.5: 21587258-21587258 on Build GRCh38
rs988258941 Chr.5: 21587302-21587302 on Build GRCh38
rs17683739 Chr.5: 21586874-21586874 on Build GRCh38
rs531997333 Chr.5: 21587047-21587047 on Build GRCh38
rs199552132 Chr.5: 21587334-21587334 on Build GRCh38
rs779062699 Chr.5: 21587329-21587329 on Build GRCh38
rs887700736 Chr.5: 21587213-21587214 on Build GRCh38
rs73059053 Chr.5: 21586921-21586921 on Build GRCh38
rs925897489 Chr.5: 21587121-21587121 on Build GRCh38
rs62350445 Chr.5: 21587053-21587053 on Build GRCh38
rs992220837 Chr.5: 21587335-21587335 on Build GRCh38
rs975434586 Chr.5: 21586880-21586880 on Build GRCh38
rs955473146 Chr.5: 21586914-21586914 on Build GRCh38
rs561678798 Chr.5: 21587103-21587103 on Build GRCh38
rs569331065 Chr.5: 21587325-21587325 on Build GRCh38
rs925866244 Chr.5: 21587333-21587333 on Build GRCh38
rs1031398617 Chr.5: 21587264-21587264 on Build GRCh38
rs900111881 Chr.5: 21587257-21587257 on Build GRCh38
rs192706771 Chr.5: 21587176-21587176 on Build GRCh38
rs976151100 Chr.5: 21586905-21586905 on Build GRCh38
rs536427330 Chr.5: 21587096-21587130 on Build GRCh38
rs146654628 Chr.5: 21587181-21587181 on Build GRCh38
rs955684363 Chr.5: 21587272-21587272 on Build GRCh38
rs754962588 Chr.5: 21587298-21587298 on Build GRCh38
rs1007797872 Chr.5: 21587223-21587223 on Build GRCh38
rs1013961798 Chr.5: 21587194-21587194 on Build GRCh38
rs531483714 Chr.5: 21587085-21587085 on Build GRCh38
rs956941177 Chr.5: 21586923-21586923 on Build GRCh38
rs1042998457 Chr.5: 21587075-21587075 on Build GRCh38
rs1048967657 Chr.5: 21587187-21587187 on Build GRCh38
rs922665998 Chr.5: 21586906-21586906 on Build GRCh38
rs756517672 Chr.5: 21586913-21586913 on Build GRCh38
rs915513457 Chr.5: 21586941-21586941 on Build GRCh38
rs1006192508 Chr.5: 21587238-21587238 on Build GRCh38
rs546115799 Chr.5: 21586932-21586932 on Build GRCh38
rs939471955 Chr.5: 21587128-21587128 on Build GRCh38
rs530920813 Chr.5: 21586853-21586854 on Build GRCh38
rs936996503 Chr.5: 21587135-21587135 on Build GRCh38
rs919342811 Chr.5: 21587339-21587339 on Build GRCh38
rs549735754 Chr.5: 21587318-21587318 on Build GRCh38
rs1029705473 Chr.5: 21587297-21587297 on Build GRCh38
rs146877238 Chr.5: 21587170-21587170 on Build GRCh38
rs1056852105 Chr.5: 21587166-21587168 on Build GRCh38
rs887296326 Chr.5: 21587232-21587232 on Build GRCh38
rs115386957 Chr.5: 21587368-21587368 on Build GRCh38
rs1047640310 Chr.5: 21587220-21587220 on Build GRCh38
rs527539324 Chr.5: 21587152-21587152 on Build GRCh38
rs548201438 Chr.5: 21587101-21587101 on Build GRCh38
rs569166906 Chr.5: 21587024-21587024 on Build GRCh38
rs567061845 Chr.5: 21587334-21587334 on Build GRCh38
rs140737757 Chr.5: 21587379-21587379 on Build GRCh38
rs988325801 Chr.5: 21586928-21586928 on Build GRCh38

View Full Product Details