Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 30263177-30263734 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HCG17
Entrez Gene ID
414778
Gene Name
HLA complex group 17 (non-protein coding)
Gene Aliases
HCG18, LINC00046, NCRNA00046
UniGene
Hs.485041
Gene Chromosome Location
Chr.6: 30234039-30326134 on Build GRCh38
Transcripts
NR_052012.1 NR_027822.1

Gene Symbol
HLA-L
Entrez Gene ID
3139
Gene Name
major histocompatibility complex, class I, L (pseudogene)
Gene Aliases
HLA-92, HLA92, HLAL
UniGene
Hs.656020
Gene Chromosome Location
Chr.6: 30259562-30266951 on Build GRCh38
Transcripts
NR_052012.1 NR_027822.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6677946 c.551G>A p.G184E Chr.6: 30263330-30263330 on Build GRCh38
6677945 c.647_648delAA p.*216fs? Chr.6: 30263426-30263427 on Build GRCh38
5466797 c.574G>A p.A192T Chr.6: 30263353-30263353 on Build GRCh38
5870034 c.584delT p.P197fs*>19 Chr.6: 30263363-30263363 on Build GRCh38
5870033 c.520G>A p.G174S Chr.6: 30263299-30263299 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772832759 Chr.6: 30263388-30263388 on Build GRCh38
rs771771507 Chr.6: 30263367-30263367 on Build GRCh38
rs145231932 Chr.6: 30263299-30263299 on Build GRCh38
rs754264056 Chr.6: 30263428-30263428 on Build GRCh38
rs757165579 Chr.6: 30263654-30263654 on Build GRCh38
rs942181167 Chr.6: 30263437-30263437 on Build GRCh38
rs761188600 Chr.6: 30263435-30263436 on Build GRCh38
rs761387755 Chr.6: 30263334-30263334 on Build GRCh38
rs529941501 Chr.6: 30263274-30263274 on Build GRCh38
rs747299344 Chr.6: 30263308-30263308 on Build GRCh38
rs147993495 Chr.6: 30263313-30263313 on Build GRCh38
rs757499849 Chr.6: 30263452-30263452 on Build GRCh38
rs544040815 Chr.6: 30263229-30263229 on Build GRCh38
rs1051283210 Chr.6: 30263655-30263655 on Build GRCh38
rs935784476 Chr.6: 30263620-30263620 on Build GRCh38
rs775243124 Chr.6: 30263383-30263383 on Build GRCh38
rs750025551 Chr.6: 30263236-30263236 on Build GRCh38
rs781341159 Chr.6: 30263456-30263456 on Build GRCh38
rs374848887 Chr.6: 30263443-30263443 on Build GRCh38
rs777889566 Chr.6: 30263444-30263444 on Build GRCh38
rs3130401 Chr.6: 30263496-30263496 on Build GRCh38
rs768659219 Chr.6: 30263328-30263328 on Build GRCh38
rs760450423 Chr.6: 30263412-30263412 on Build GRCh38
rs561156261 Chr.6: 30263242-30263242 on Build GRCh38
rs3094073 Chr.6: 30263447-30263447 on Build GRCh38
rs200574417 Chr.6: 30263510-30263510 on Build GRCh38
rs766906144 Chr.6: 30263266-30263266 on Build GRCh38
rs3132659 Chr.6: 30263553-30263553 on Build GRCh38
rs9278630 Chr.6: 30263509-30263509 on Build GRCh38
rs764763656 Chr.6: 30263691-30263691 on Build GRCh38
rs927095518 Chr.6: 30263215-30263215 on Build GRCh38
rs750121871 Chr.6: 30263702-30263702 on Build GRCh38
rs2516699 Chr.6: 30263331-30263331 on Build GRCh38
rs2516700 Chr.6: 30263294-30263294 on Build GRCh38
rs748286346 Chr.6: 30263297-30263297 on Build GRCh38
rs190298933 Chr.6: 30263295-30263295 on Build GRCh38
rs748994811 Chr.6: 30263325-30263325 on Build GRCh38
rs749121399 Chr.6: 30263460-30263460 on Build GRCh38
rs1006932233 Chr.6: 30263326-30263326 on Build GRCh38
rs755551541 Chr.6: 30263289-30263289 on Build GRCh38
rs766101839 Chr.6: 30263418-30263418 on Build GRCh38
rs773714730 Chr.6: 30263315-30263316 on Build GRCh38
rs373851852 Chr.6: 30263228-30263228 on Build GRCh38
rs898582550 Chr.6: 30263362-30263362 on Build GRCh38
rs373399670 Chr.6: 30263441-30263441 on Build GRCh38
rs370516808 Chr.6: 30263510-30263510 on Build GRCh38
rs777977377 Chr.6: 30263307-30263307 on Build GRCh38
rs865813029 Chr.6: 30263293-30263293 on Build GRCh38
rs536489956 Chr.6: 30263639-30263639 on Build GRCh38
rs759773846 Chr.6: 30263436-30263436 on Build GRCh38
rs141674432 Chr.6: 30263320-30263320 on Build GRCh38
rs764969535 Chr.6: 30263454-30263454 on Build GRCh38
rs895813379 Chr.6: 30263636-30263636 on Build GRCh38
rs770735220 Chr.6: 30263387-30263387 on Build GRCh38
rs990695500 Chr.6: 30263623-30263623 on Build GRCh38
rs748092032 Chr.6: 30263232-30263232 on Build GRCh38
rs772265288 Chr.6: 30263299-30263299 on Build GRCh38

View Full Product Details