Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32006286-32006834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP21A1P
Entrez Gene ID
1590
Gene Name
cytochrome P450 family 21 subfamily A member 1, pseudogene
Gene Aliases
CYP21A, CYP21P, P450c21A
UniGene
Hs.654479
Gene Chromosome Location
Chr.6: 32005636-32008909 on Build GRCh38
Transcripts
NR_040090.1

SNP Detail

SNP ID Chromosome Location
rs778763057 Chr.6: 32006318-32006318 on Build GRCh38
rs769383737 Chr.6: 32006436-32006436 on Build GRCh38
rs546382810 Chr.6: 32006669-32006669 on Build GRCh38
rs958912857 Chr.6: 32006331-32006331 on Build GRCh38
rs541161602 Chr.6: 32006706-32006706 on Build GRCh38
rs770542536 Chr.6: 32006345-32006345 on Build GRCh38
rs769465406 Chr.6: 32006589-32006589 on Build GRCh38
rs200147745 Chr.6: 32006462-32006462 on Build GRCh38
rs575901843 Chr.6: 32006620-32006620 on Build GRCh38
rs547386048 Chr.6: 32006336-32006336 on Build GRCh38
rs183512826 Chr.6: 32006381-32006381 on Build GRCh38
rs201094127 Chr.6: 32006550-32006550 on Build GRCh38
rs1015668839 Chr.6: 32006808-32006810 on Build GRCh38
rs755091925 Chr.6: 32006385-32006385 on Build GRCh38
rs183831026 Chr.6: 32006795-32006795 on Build GRCh38
rs763333539 Chr.6: 32006444-32006444 on Build GRCh38
rs755020999 Chr.6: 32006687-32006687 on Build GRCh38
rs544758585 Chr.6: 32006682-32006682 on Build GRCh38
rs758255784 Chr.6: 32006553-32006553 on Build GRCh38
rs750514817 Chr.6: 32006397-32006397 on Build GRCh38
rs770505386 Chr.6: 32006426-32006426 on Build GRCh38
rs551963712 Chr.6: 32006439-32006439 on Build GRCh38
rs776359700 Chr.6: 32006430-32006430 on Build GRCh38
rs773553083 Chr.6: 32006756-32006756 on Build GRCh38
rs552928784 Chr.6: 32006488-32006488 on Build GRCh38
rs756119190 Chr.6: 32006399-32006399 on Build GRCh38
rs757054481 Chr.6: 32006698-32006698 on Build GRCh38
rs772997765 Chr.6: 32006765-32006765 on Build GRCh38
rs781017188 Chr.6: 32006583-32006583 on Build GRCh38
rs758469000 Chr.6: 32006317-32006318 on Build GRCh38
rs147487168 Chr.6: 32006690-32006690 on Build GRCh38
rs763428582 Chr.6: 32006365-32006365 on Build GRCh38
rs1004184331 Chr.6: 32006783-32006783 on Build GRCh38
rs751468144 Chr.6: 32006691-32006691 on Build GRCh38
rs991146568 Chr.6: 32006357-32006357 on Build GRCh38
rs775993702 Chr.6: 32006351-32006351 on Build GRCh38
rs762114303 Chr.6: 32006638-32006638 on Build GRCh38
rs186792335 Chr.6: 32006408-32006408 on Build GRCh38
rs753929542 Chr.6: 32006684-32006684 on Build GRCh38
rs200662100 Chr.6: 32006353-32006353 on Build GRCh38
rs774852583 Chr.6: 32006637-32006637 on Build GRCh38
rs765341153 Chr.6: 32006792-32006792 on Build GRCh38
rs771723635 Chr.6: 32006329-32006329 on Build GRCh38
rs28641793 Chr.6: 32006578-32006578 on Build GRCh38
rs745405653 Chr.6: 32006433-32006433 on Build GRCh38
rs771148605 Chr.6: 32006760-32006760 on Build GRCh38
rs570578547 Chr.6: 32006463-32006463 on Build GRCh38
rs779309787 Chr.6: 32006738-32006738 on Build GRCh38
rs559291124 Chr.6: 32006707-32006707 on Build GRCh38
rs529701523 Chr.6: 32006759-32006759 on Build GRCh38
rs900077807 Chr.6: 32006739-32006739 on Build GRCh38
rs749287586 Chr.6: 32006314-32006314 on Build GRCh38
rs778852956 Chr.6: 32006400-32006400 on Build GRCh38
rs757414795 Chr.6: 32006391-32006391 on Build GRCh38
rs761834506 Chr.6: 32006802-32006802 on Build GRCh38
rs756208796 Chr.6: 32006539-32006539 on Build GRCh38
rs747789065 Chr.6: 32006390-32006390 on Build GRCh38
rs56155986 Chr.6: 32006368-32006368 on Build GRCh38
rs192097200 Chr.6: 32006422-32006422 on Build GRCh38
rs559415863 Chr.6: 32006343-32006343 on Build GRCh38
rs749862295 Chr.6: 32006621-32006621 on Build GRCh38
rs767821242 Chr.6: 32006649-32006649 on Build GRCh38
rs534813636 Chr.6: 32006481-32006481 on Build GRCh38
rs750602869 Chr.6: 32006516-32006516 on Build GRCh38
rs748751282 Chr.6: 32006727-32006727 on Build GRCh38
rs574173238 Chr.6: 32006579-32006579 on Build GRCh38
rs747941297 Chr.6: 32006328-32006328 on Build GRCh38
rs759559800 Chr.6: 32006768-32006768 on Build GRCh38
rs532258592 Chr.6: 32006317-32006317 on Build GRCh38
rs776179786 Chr.6: 32006344-32006344 on Build GRCh38
rs529984739 Chr.6: 32006358-32006358 on Build GRCh38
rs1079995 Chr.6: 32006551-32006551 on Build GRCh38
rs777710341 Chr.6: 32006571-32006571 on Build GRCh38
rs745488806 Chr.6: 32006588-32006588 on Build GRCh38
rs748666629 Chr.6: 32006743-32006743 on Build GRCh38
rs764583707 Chr.6: 32006456-32006456 on Build GRCh38
rs72502600 Chr.6: 32006812-32006812 on Build GRCh38
rs772492579 Chr.6: 32006745-32006745 on Build GRCh38
rs769054250 Chr.6: 32006635-32006635 on Build GRCh38

View Full Product Details