Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 7079294-7079884 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC100131257
Entrez Gene ID
100131257
Gene Name
zinc finger protein 655 pseudogene
Gene Aliases
-
UniGene
Hs.551110
Gene Chromosome Location
Chr.7: 7075770-7096786 on Build GRCh38
Transcripts
NR_034022.1

SNP Detail

SNP ID Chromosome Location
rs774093502 Chr.7: 7079704-7079704 on Build GRCh38
rs959901721 Chr.7: 7079397-7079397 on Build GRCh38
rs71533347 Chr.7: 7079607-7079607 on Build GRCh38
rs147199112 Chr.7: 7079548-7079548 on Build GRCh38
rs989071162 Chr.7: 7079752-7079752 on Build GRCh38
rs191591024 Chr.7: 7079448-7079448 on Build GRCh38
rs183666106 Chr.7: 7079366-7079366 on Build GRCh38
rs947302173 Chr.7: 7079474-7079474 on Build GRCh38
rs942711525 Chr.7: 7079786-7079786 on Build GRCh38
rs1015887946 Chr.7: 7079606-7079606 on Build GRCh38
rs578060319 Chr.7: 7079481-7079481 on Build GRCh38
rs570263143 Chr.7: 7079732-7079732 on Build GRCh38
rs779197073 Chr.7: 7079735-7079735 on Build GRCh38
rs549621467 Chr.7: 7079784-7079784 on Build GRCh38
rs760313181 Chr.7: 7079547-7079549 on Build GRCh38
rs144674895 Chr.7: 7079423-7079423 on Build GRCh38
rs190214289 Chr.7: 7079564-7079564 on Build GRCh38
rs2280918 Chr.7: 7079838-7079838 on Build GRCh38
rs748748764 Chr.7: 7079689-7079689 on Build GRCh38
rs1035434908 Chr.7: 7079642-7079642 on Build GRCh38
rs148116669 Chr.7: 7079518-7079518 on Build GRCh38
rs768249747 Chr.7: 7079596-7079596 on Build GRCh38
rs573979299 Chr.7: 7079449-7079449 on Build GRCh38
rs755662882 Chr.7: 7079595-7079595 on Build GRCh38
rs978637074 Chr.7: 7079765-7079765 on Build GRCh38
rs543259899 Chr.7: 7079560-7079560 on Build GRCh38
rs532769536 Chr.7: 7079647-7079647 on Build GRCh38
rs181917220 Chr.7: 7079618-7079618 on Build GRCh38
rs61742262 Chr.7: 7079369-7079369 on Build GRCh38
rs189511952 Chr.7: 7079757-7079757 on Build GRCh38
rs747590239 Chr.7: 7079816-7079816 on Build GRCh38
rs141869178 Chr.7: 7079537-7079537 on Build GRCh38
rs776265139 Chr.7: 7079792-7079792 on Build GRCh38
rs746676307 Chr.7: 7079764-7079764 on Build GRCh38
rs12113977 Chr.7: 7079628-7079628 on Build GRCh38
rs551771236 Chr.7: 7079354-7079354 on Build GRCh38
rs185508924 Chr.7: 7079684-7079684 on Build GRCh38
rs768191300 Chr.7: 7079773-7079773 on Build GRCh38
rs911192612 Chr.7: 7079487-7079487 on Build GRCh38
rs770935255 Chr.7: 7079695-7079695 on Build GRCh38
rs901249864 Chr.7: 7079528-7079528 on Build GRCh38
rs184915832 Chr.7: 7079469-7079469 on Build GRCh38
rs988650148 Chr.7: 7079419-7079419 on Build GRCh38
rs888713183 Chr.7: 7079558-7079558 on Build GRCh38
rs942765369 Chr.7: 7079507-7079507 on Build GRCh38

View Full Product Details