Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 44028785-44029327 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RASA4CP
Entrez Gene ID
401331
Gene Name
RAS p21 protein activator 4C, pseudogene
Gene Aliases
RASA4P
UniGene
Hs.530089
Gene Chromosome Location
Chr.7: 44028887-44040623 on Build GRCh38
Transcripts
NR_024116.2

SNP Detail

SNP ID Chromosome Location
rs1023393409 Chr.7: 44029245-44029247 on Build GRCh38
rs890398657 Chr.7: 44029134-44029134 on Build GRCh38
rs1031145303 Chr.7: 44028957-44028957 on Build GRCh38
rs59250451 Chr.7: 44029279-44029279 on Build GRCh38
rs980261229 Chr.7: 44028834-44028835 on Build GRCh38
rs368759821 Chr.7: 44029164-44029164 on Build GRCh38
rs1030207874 Chr.7: 44028932-44028932 on Build GRCh38
rs2595662 Chr.7: 44029282-44029282 on Build GRCh38
rs938824581 Chr.7: 44029017-44029017 on Build GRCh38
rs1052306647 Chr.7: 44029215-44029215 on Build GRCh38
rs879894375 Chr.7: 44029226-44029226 on Build GRCh38
rs989179672 Chr.7: 44028807-44028807 on Build GRCh38
rs900419967 Chr.7: 44028911-44028911 on Build GRCh38
rs1010701819 Chr.7: 44029234-44029234 on Build GRCh38
rs973424853 Chr.7: 44029291-44029291 on Build GRCh38
rs960345893 Chr.7: 44029270-44029271 on Build GRCh38
rs980313156 Chr.7: 44028979-44028979 on Build GRCh38
rs931991863 Chr.7: 44029108-44029108 on Build GRCh38
rs1037692505 Chr.7: 44028896-44028896 on Build GRCh38
rs1003227445 Chr.7: 44029255-44029255 on Build GRCh38
rs970296249 Chr.7: 44028975-44028975 on Build GRCh38
rs983659329 Chr.7: 44029026-44029026 on Build GRCh38
rs574858903 Chr.7: 44028921-44028921 on Build GRCh38
rs564512829 Chr.7: 44029107-44029107 on Build GRCh38
rs2595663 Chr.7: 44029064-44029064 on Build GRCh38
rs989135551 Chr.7: 44028962-44028962 on Build GRCh38
rs999749237 Chr.7: 44029202-44029202 on Build GRCh38
rs533832547 Chr.7: 44028818-44028818 on Build GRCh38
rs963515530 Chr.7: 44029285-44029285 on Build GRCh38
rs761128791 Chr.7: 44029304-44029304 on Build GRCh38
rs576392299 Chr.7: 44028841-44028841 on Build GRCh38
rs113779039 Chr.7: 44029109-44029109 on Build GRCh38
rs890252726 Chr.7: 44028935-44028935 on Build GRCh38
rs192769047 Chr.7: 44029028-44029028 on Build GRCh38
rs1023342652 Chr.7: 44028963-44028963 on Build GRCh38
rs112549799 Chr.7: 44029133-44029133 on Build GRCh38
rs938867735 Chr.7: 44028842-44028842 on Build GRCh38
rs1048245625 Chr.7: 44028864-44028864 on Build GRCh38
rs914809483 Chr.7: 44028819-44028819 on Build GRCh38
rs922034059 Chr.7: 44029305-44029305 on Build GRCh38
rs2595664 Chr.7: 44028895-44028895 on Build GRCh38
rs1036332916 Chr.7: 44029262-44029262 on Build GRCh38
rs542260294 Chr.7: 44028945-44028945 on Build GRCh38
rs927491466 Chr.7: 44028995-44028995 on Build GRCh38
rs528823418 Chr.7: 44029287-44029287 on Build GRCh38
rs928883498 Chr.7: 44029016-44029016 on Build GRCh38
rs528717835 Chr.7: 44029254-44029254 on Build GRCh38
rs543889784 Chr.7: 44028958-44028958 on Build GRCh38
rs886542339 Chr.7: 44028882-44028882 on Build GRCh38
rs948980302 Chr.7: 44028829-44028829 on Build GRCh38
rs546324196 Chr.7: 44029034-44029034 on Build GRCh38
rs762699126 Chr.7: 44029049-44029049 on Build GRCh38
rs942005269 Chr.7: 44028883-44028883 on Build GRCh38

View Full Product Details