Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 129126016-129126572 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LOC407835
Entrez Gene ID
407835
Gene Name
mitogen-activated protein kinase kinase 2 pseudogene
Gene Aliases
-
UniGene
Hs.592273
Gene Chromosome Location
Chr.7: 129126271-129128007 on Build GRCh38
Transcripts
NR_002144.1

SNP Detail

SNP ID Chromosome Location
rs574490294 Chr.7: 129126253-129126253 on Build GRCh38
rs191933253 Chr.7: 129126312-129126312 on Build GRCh38
rs796900127 Chr.7: 129126307-129126307 on Build GRCh38
rs10464695 Chr.7: 129126409-129126409 on Build GRCh38
rs952866391 Chr.7: 129126255-129126255 on Build GRCh38
rs765719416 Chr.7: 129126476-129126476 on Build GRCh38
rs3745986 Chr.7: 129126411-129126411 on Build GRCh38
rs10464697 Chr.7: 129126474-129126474 on Build GRCh38
rs904192249 Chr.7: 129126339-129126339 on Build GRCh38
rs10464694 Chr.7: 129126111-129126111 on Build GRCh38
rs908675173 Chr.7: 129126298-129126298 on Build GRCh38
rs187139581 Chr.7: 129126093-129126093 on Build GRCh38
rs904297921 Chr.7: 129126103-129126103 on Build GRCh38
rs1012604730 Chr.7: 129126099-129126099 on Build GRCh38
rs975414193 Chr.7: 129126244-129126244 on Build GRCh38
rs530144266 Chr.7: 129126527-129126527 on Build GRCh38
rs996307465 Chr.7: 129126242-129126242 on Build GRCh38
rs927696079 Chr.7: 129126051-129126051 on Build GRCh38
rs774443199 Chr.7: 129126497-129126499 on Build GRCh38
rs776804624 Chr.7: 129126510-129126510 on Build GRCh38
rs1018805336 Chr.7: 129126423-129126423 on Build GRCh38
rs747839526 Chr.7: 129126501-129126501 on Build GRCh38
rs768886029 Chr.7: 129126473-129126473 on Build GRCh38
rs1008710045 Chr.7: 129126412-129126412 on Build GRCh38
rs541969955 Chr.7: 129126283-129126283 on Build GRCh38
rs1028284676 Chr.7: 129126432-129126432 on Build GRCh38
rs1053294790 Chr.7: 129126386-129126386 on Build GRCh38
rs1029123652 Chr.7: 129126259-129126259 on Build GRCh38
rs952830940 Chr.7: 129126437-129126437 on Build GRCh38
rs780653626 Chr.7: 129126484-129126484 on Build GRCh38
rs753275324 Chr.7: 129126529-129126529 on Build GRCh38
rs755711913 Chr.7: 129126525-129126525 on Build GRCh38
rs773372091 Chr.7: 129126506-129126506 on Build GRCh38
rs1044132296 Chr.7: 129126152-129126152 on Build GRCh38
rs10464698 Chr.7: 129126479-129126479 on Build GRCh38
rs940215862 Chr.7: 129126058-129126058 on Build GRCh38
rs553032653 Chr.7: 129126200-129126200 on Build GRCh38
rs935733011 Chr.7: 129126365-129126365 on Build GRCh38
rs900461106 Chr.7: 129126426-129126426 on Build GRCh38
rs948352192 Chr.7: 129126323-129126323 on Build GRCh38
rs1044100493 Chr.7: 129126333-129126333 on Build GRCh38
rs537764474 Chr.7: 129126131-129126131 on Build GRCh38
rs566154337 Chr.7: 129126517-129126517 on Build GRCh38
rs939202259 Chr.7: 129126090-129126090 on Build GRCh38
rs984614741 Chr.7: 129126271-129126271 on Build GRCh38
rs754438804 Chr.7: 129126475-129126475 on Build GRCh38
rs753123556 Chr.7: 129126516-129126516 on Build GRCh38
rs761898829 Chr.7: 129126526-129126527 on Build GRCh38
rs745523204 Chr.7: 129126492-129126492 on Build GRCh38
rs917527297 Chr.7: 129126320-129126320 on Build GRCh38
rs1008741442 Chr.7: 129126157-129126160 on Build GRCh38
rs901926827 Chr.7: 129126217-129126217 on Build GRCh38
rs530686127 Chr.7: 129126319-129126319 on Build GRCh38
rs144539404 Chr.7: 129126149-129126149 on Build GRCh38
rs1057398083 Chr.7: 129126074-129126074 on Build GRCh38
rs540701726 Chr.7: 129126186-129126186 on Build GRCh38
rs758210352 Chr.7: 129126245-129126245 on Build GRCh38
rs997252377 Chr.7: 129126428-129126428 on Build GRCh38
rs771885520 Chr.7: 129126503-129126503 on Build GRCh38
rs779515070 Chr.7: 129126495-129126495 on Build GRCh38
rs1019024287 Chr.7: 129126208-129126208 on Build GRCh38
rs762125455 Chr.7: 129126471-129126471 on Build GRCh38
rs6952057 Chr.7: 129126451-129126451 on Build GRCh38
rs755617164 Chr.7: 129126493-129126493 on Build GRCh38
rs760152062 Chr.7: 129126514-129126514 on Build GRCh38
rs1044518587 Chr.7: 129126100-129126100 on Build GRCh38
rs902539047 Chr.7: 129126157-129126157 on Build GRCh38
rs1000436481 Chr.7: 129126106-129126106 on Build GRCh38
rs766617100 Chr.7: 129126509-129126509 on Build GRCh38
rs760921088 Chr.7: 129126507-129126507 on Build GRCh38
rs767567766 Chr.7: 129126473-129126473 on Build GRCh38
rs558554150 Chr.7: 129126147-129126147 on Build GRCh38
rs749242379 Chr.7: 129126496-129126496 on Build GRCh38
rs750089706 Chr.7: 129126521-129126521 on Build GRCh38
rs764518395 Chr.7: 129126520-129126520 on Build GRCh38
rs754921429 Chr.7: 129126531-129126531 on Build GRCh38

View Full Product Details