Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 53788951-53789314 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NDC1
Entrez Gene ID
55706
Gene Name
NDC1 transmembrane nucleoporin
Gene Aliases
NET3, TMEM48
UniGene
Hs.476525
Gene Chromosome Location
Chr.1: 53765460-53838552 on Build GRCh38
Transcripts
NM_001168551.1 NM_018087.4 NR_033142.1 XM_011541766.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3490780 c.1649C>T p.S550F Chr.1: 53789183-53789183 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs368922094 Chr.1: 53789208-53789208 on Build GRCh38
rs754591838 Chr.1: 53789177-53789177 on Build GRCh38
rs890847281 Chr.1: 53789022-53789022 on Build GRCh38
rs750676104 Chr.1: 53789244-53789244 on Build GRCh38
rs1007976625 Chr.1: 53789024-53789024 on Build GRCh38
rs769374376 Chr.1: 53789137-53789137 on Build GRCh38
rs189347848 Chr.1: 53789028-53789028 on Build GRCh38
rs1050736024 Chr.1: 53788997-53788997 on Build GRCh38
rs147077553 Chr.1: 53789188-53789188 on Build GRCh38
rs201585736 Chr.1: 53789267-53789267 on Build GRCh38
rs377046272 Chr.1: 53789127-53789127 on Build GRCh38
rs201816655 Chr.1: 53789267-53789267 on Build GRCh38
rs755485919 Chr.1: 53789106-53789106 on Build GRCh38
rs748590393 Chr.1: 53789113-53789113 on Build GRCh38
rs372408315 Chr.1: 53789111-53789111 on Build GRCh38
rs776398313 Chr.1: 53789018-53789018 on Build GRCh38
rs552255946 Chr.1: 53789027-53789027 on Build GRCh38
rs777687072 Chr.1: 53789211-53789213 on Build GRCh38
rs370200244 Chr.1: 53789119-53789119 on Build GRCh38
rs775239207 Chr.1: 53789174-53789174 on Build GRCh38
rs982897957 Chr.1: 53789172-53789172 on Build GRCh38
rs116539849 Chr.1: 53789207-53789207 on Build GRCh38
rs752645808 Chr.1: 53789098-53789099 on Build GRCh38
rs944551775 Chr.1: 53789278-53789278 on Build GRCh38
rs767660252 Chr.1: 53789218-53789218 on Build GRCh38
rs559567972 Chr.1: 53789050-53789050 on Build GRCh38
rs180846077 Chr.1: 53789062-53789062 on Build GRCh38
rs911806073 Chr.1: 53789255-53789255 on Build GRCh38
rs533206467 Chr.1: 53789222-53789222 on Build GRCh38
rs936825317 Chr.1: 53789283-53789283 on Build GRCh38
rs944518977 Chr.1: 53789019-53789019 on Build GRCh38
rs756102709 Chr.1: 53789117-53789118 on Build GRCh38
rs986103271 Chr.1: 53789228-53789228 on Build GRCh38
rs754392980 Chr.1: 53789083-53789083 on Build GRCh38
rs1002469906 Chr.1: 53789072-53789072 on Build GRCh38
rs753537046 Chr.1: 53789091-53789091 on Build GRCh38
rs766459039 Chr.1: 53789245-53789245 on Build GRCh38
rs376992599 Chr.1: 53789118-53789118 on Build GRCh38
rs755952976 Chr.1: 53789010-53789010 on Build GRCh38
rs113270967 Chr.1: 53789054-53789054 on Build GRCh38

View Full Product Details