Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 14126016-14126305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ASF1B
Entrez Gene ID
55723
Gene Name
anti-silencing function 1B histone chaperone
Gene Aliases
CIA-II
UniGene
Hs.26516
Gene Chromosome Location
Chr.19: 14119509-14136628 on Build GRCh38
Transcripts
NM_018154.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
268715 c.128T>G p.I43S Chr.19: 14126219-14126219 on Build GRCh38
991824 c.112C>T p.L38L Chr.19: 14126235-14126235 on Build GRCh38
1750682 c.110-1G>A p.? Chr.19: 14126238-14126238 on Build GRCh38
6539071 c.145G>C p.E49Q Chr.19: 14126202-14126202 on Build GRCh38
5797332 c.204G>T p.G68G Chr.19: 14126143-14126143 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755843923 Chr.19: 14126146-14126146 on Build GRCh38
rs373277929 Chr.19: 14126191-14126191 on Build GRCh38
rs767856727 Chr.19: 14126255-14126255 on Build GRCh38
rs769492764 Chr.19: 14126135-14126136 on Build GRCh38
rs772162700 Chr.19: 14126092-14126092 on Build GRCh38
rs368856106 Chr.19: 14126170-14126170 on Build GRCh38
rs759509266 Chr.19: 14126272-14126272 on Build GRCh38
rs747288070 Chr.19: 14126103-14126103 on Build GRCh38
rs779012633 Chr.19: 14126164-14126164 on Build GRCh38
rs767209457 Chr.19: 14126256-14126256 on Build GRCh38
rs559670040 Chr.19: 14126197-14126197 on Build GRCh38
rs565115248 Chr.19: 14126141-14126141 on Build GRCh38
rs761571770 Chr.19: 14126135-14126135 on Build GRCh38
rs760314299 Chr.19: 14126267-14126267 on Build GRCh38
rs137874538 Chr.19: 14126281-14126281 on Build GRCh38
rs773715421 Chr.19: 14126118-14126118 on Build GRCh38
rs758989967 Chr.19: 14126093-14126094 on Build GRCh38
rs748460085 Chr.19: 14126086-14126086 on Build GRCh38
rs566307477 Chr.19: 14126045-14126045 on Build GRCh38
rs773938801 Chr.19: 14126252-14126252 on Build GRCh38
rs965023038 Chr.19: 14126204-14126204 on Build GRCh38
rs918502510 Chr.19: 14126249-14126249 on Build GRCh38
rs148060285 Chr.19: 14126179-14126179 on Build GRCh38
rs746430062 Chr.19: 14126236-14126236 on Build GRCh38
rs751749975 Chr.19: 14126206-14126206 on Build GRCh38
rs772671019 Chr.19: 14126265-14126265 on Build GRCh38
rs199895808 Chr.19: 14126157-14126157 on Build GRCh38
rs143580212 Chr.19: 14126152-14126152 on Build GRCh38
rs747406217 Chr.19: 14126202-14126202 on Build GRCh38
rs762215946 Chr.19: 14126209-14126209 on Build GRCh38
rs754982565 Chr.19: 14126084-14126084 on Build GRCh38
rs951966563 Chr.19: 14126257-14126257 on Build GRCh38
rs777126137 Chr.19: 14126215-14126215 on Build GRCh38
rs770397947 Chr.19: 14126238-14126238 on Build GRCh38
rs754895590 Chr.19: 14126159-14126159 on Build GRCh38
rs536826941 Chr.19: 14126046-14126046 on Build GRCh38
rs376931561 Chr.19: 14126271-14126271 on Build GRCh38
rs773656807 Chr.19: 14126096-14126096 on Build GRCh38
rs753837490 Chr.19: 14126158-14126158 on Build GRCh38
rs767033850 Chr.19: 14126140-14126140 on Build GRCh38
rs760118996 Chr.19: 14126108-14126108 on Build GRCh38
rs766218422 Chr.19: 14126270-14126270 on Build GRCh38
rs57092832 Chr.19: 14126106-14126106 on Build GRCh38
rs764338336 Chr.19: 14126151-14126151 on Build GRCh38
rs749381131 Chr.19: 14126083-14126083 on Build GRCh38
rs543610465 Chr.19: 14126113-14126113 on Build GRCh38

View Full Product Details