Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 123347981-123348400 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATAD2
Entrez Gene ID
29028
Gene Name
ATPase family, AAA domain containing 2
Gene Aliases
ANCCA, CT137, PRO2000
UniGene
Hs.370834
Gene Chromosome Location
Chr.8: 123319850-123396992 on Build GRCh38
Transcripts
NM_014109.3 XM_011516994.2 XM_011516995.2 XM_011516996.2 NR_037516.1 NR_030382.1

Gene Symbol
MIR548AA1
Entrez Gene ID
100500863
Gene Name
microRNA 548aa-1
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.8: 123348034-123348130 on Build GRCh38
Transcripts
NM_014109.3 XM_011516994.2 XM_011516995.2 XM_011516996.2 NR_037516.1 NR_030382.1

Gene Symbol
MIR548D1
Entrez Gene ID
693130
Gene Name
microRNA 548d-1
Gene Aliases
MIRN548D1
UniGene
Gene Chromosome Location
Chr.8: 123348034-123348130 on Build GRCh38
Transcripts
NM_014109.3 XM_011516994.2 XM_011516995.2 XM_011516996.2 NR_037516.1 NR_030382.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
453961 c.1857A>G p.P619P Chr.8: 123348223-123348223 on Build GRCh38
1095761 c.1810C>T p.R604* Chr.8: 123348270-123348270 on Build GRCh38
1193668 c.1816G>C p.E606Q Chr.8: 123348264-123348264 on Build GRCh38
4531915 c.1839G>A p.R613R Chr.8: 123348241-123348241 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs370140867 Chr.8: 123348025-123348025 on Build GRCh38
rs139092616 Chr.8: 123348186-123348186 on Build GRCh38
rs757332901 Chr.8: 123348060-123348061 on Build GRCh38
rs758344972 Chr.8: 123348033-123348033 on Build GRCh38
rs750212451 Chr.8: 123348255-123348255 on Build GRCh38
rs754130715 Chr.8: 123348294-123348294 on Build GRCh38
rs751509912 Chr.8: 123348243-123348243 on Build GRCh38
rs963554784 Chr.8: 123348298-123348298 on Build GRCh38
rs564189536 Chr.8: 123348009-123348009 on Build GRCh38
rs748695953 Chr.8: 123348150-123348150 on Build GRCh38
rs935904524 Chr.8: 123348061-123348061 on Build GRCh38
rs751604390 Chr.8: 123348050-123348050 on Build GRCh38
rs77106954 Chr.8: 123348247-123348247 on Build GRCh38
rs756289684 Chr.8: 123348269-123348269 on Build GRCh38
rs374379681 Chr.8: 123348030-123348030 on Build GRCh38
rs986429735 Chr.8: 123348151-123348151 on Build GRCh38
rs767291207 Chr.8: 123348250-123348250 on Build GRCh38
rs372010968 Chr.8: 123348199-123348199 on Build GRCh38
rs963181415 Chr.8: 123348043-123348043 on Build GRCh38
rs77695560 Chr.8: 123348328-123348328 on Build GRCh38
rs945838795 Chr.8: 123348231-123348231 on Build GRCh38
rs996605039 Chr.8: 123348130-123348130 on Build GRCh38
rs756308753 Chr.8: 123348103-123348103 on Build GRCh38
rs760071061 Chr.8: 123348196-123348196 on Build GRCh38
rs753807695 Chr.8: 123348153-123348157 on Build GRCh38
rs774837640 Chr.8: 123348146-123348146 on Build GRCh38
rs200851511 Chr.8: 123348036-123348036 on Build GRCh38
rs763762878 Chr.8: 123348238-123348238 on Build GRCh38
rs780888544 Chr.8: 123348077-123348077 on Build GRCh38
rs772462004 Chr.8: 123348163-123348163 on Build GRCh38
rs773472317 Chr.8: 123348166-123348166 on Build GRCh38
rs1039680238 Chr.8: 123348083-123348083 on Build GRCh38
rs759162581 Chr.8: 123348023-123348023 on Build GRCh38
rs546313806 Chr.8: 123348107-123348107 on Build GRCh38
rs781196581 Chr.8: 123348116-123348120 on Build GRCh38
rs1054802786 Chr.8: 123348211-123348211 on Build GRCh38
rs547418666 Chr.8: 123348332-123348332 on Build GRCh38
rs371444010 Chr.8: 123348026-123348026 on Build GRCh38
rs746078453 Chr.8: 123348084-123348084 on Build GRCh38
rs757069877 Chr.8: 123348068-123348068 on Build GRCh38
rs1035009357 Chr.8: 123348321-123348321 on Build GRCh38
rs755021622 Chr.8: 123348302-123348302 on Build GRCh38
rs749387291 Chr.8: 123348115-123348115 on Build GRCh38
rs368426598 Chr.8: 123348184-123348184 on Build GRCh38
rs762539572 Chr.8: 123348220-123348220 on Build GRCh38
rs1012272925 Chr.8: 123348350-123348350 on Build GRCh38
rs890551988 Chr.8: 123348157-123348157 on Build GRCh38
rs1041557770 Chr.8: 123348240-123348240 on Build GRCh38
rs370815347 Chr.8: 123348136-123348136 on Build GRCh38
rs945791405 Chr.8: 123348012-123348012 on Build GRCh38
rs780245795 Chr.8: 123348288-123348288 on Build GRCh38
rs779016467 Chr.8: 123348320-123348320 on Build GRCh38
rs374503725 Chr.8: 123348214-123348214 on Build GRCh38

View Full Product Details