Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 71837340-71837721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFB108B
Entrez Gene ID
245911
Gene Name
defensin beta 108B
Gene Aliases
DEFB-8, hBD-8
UniGene
Hs.555032
Gene Chromosome Location
Chr.11: 71833200-71837562 on Build GRCh38
Transcripts
NM_001002035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5839527 c.208C>T p.P70S Chr.11: 71837548-71837548 on Build GRCh38
6642180 c.212A>C p.K71T Chr.11: 71837552-71837552 on Build GRCh38
6642181 c.69A>C p.K23N Chr.11: 71837409-71837409 on Build GRCh38
232151 c.158A>G p.Q53R Chr.11: 71837498-71837498 on Build GRCh38
467375 c.72C>G p.F24L Chr.11: 71837412-71837412 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs531858025 Chr.11: 71837541-71837541 on Build GRCh38
rs201854901 Chr.11: 71837519-71837519 on Build GRCh38
rs773781729 Chr.11: 71837540-71837540 on Build GRCh38
rs991221017 Chr.11: 71837662-71837662 on Build GRCh38
rs199595023 Chr.11: 71837477-71837477 on Build GRCh38
rs367553368 Chr.11: 71837436-71837436 on Build GRCh38
rs777312251 Chr.11: 71837520-71837520 on Build GRCh38
rs767328417 Chr.11: 71837410-71837412 on Build GRCh38
rs757126254 Chr.11: 71837480-71837480 on Build GRCh38
rs866145014 Chr.11: 71837410-71837410 on Build GRCh38
rs373789469 Chr.11: 71837456-71837456 on Build GRCh38
rs746190682 Chr.11: 71837521-71837521 on Build GRCh38
rs772022019 Chr.11: 71837437-71837437 on Build GRCh38
rs1022738820 Chr.11: 71837667-71837667 on Build GRCh38
rs775985368 Chr.11: 71837528-71837528 on Build GRCh38
rs529069743 Chr.11: 71837663-71837663 on Build GRCh38
rs764272809 Chr.11: 71837615-71837622 on Build GRCh38
rs367546709 Chr.11: 71837497-71837497 on Build GRCh38
rs764047296 Chr.11: 71837400-71837400 on Build GRCh38
rs151229532 Chr.11: 71837428-71837428 on Build GRCh38
rs769986675 Chr.11: 71837653-71837653 on Build GRCh38
rs531662353 Chr.11: 71837534-71837534 on Build GRCh38
rs971237882 Chr.11: 71837690-71837690 on Build GRCh38
rs750337604 Chr.11: 71837545-71837545 on Build GRCh38
rs967106082 Chr.11: 71837412-71837412 on Build GRCh38
rs766083888 Chr.11: 71837556-71837556 on Build GRCh38
rs759426240 Chr.11: 71837402-71837402 on Build GRCh38
rs1050566419 Chr.11: 71837509-71837509 on Build GRCh38
rs770036762 Chr.11: 71837524-71837524 on Build GRCh38
rs767445344 Chr.11: 71837486-71837486 on Build GRCh38
rs998542685 Chr.11: 71837601-71837601 on Build GRCh38
rs758714905 Chr.11: 71837503-71837503 on Build GRCh38
rs12793731 Chr.11: 71837446-71837446 on Build GRCh38
rs758493122 Chr.11: 71837581-71837581 on Build GRCh38
rs751125854 Chr.11: 71837478-71837478 on Build GRCh38
rs757881672 Chr.11: 71837398-71837398 on Build GRCh38
rs12285495 Chr.11: 71837474-71837474 on Build GRCh38
rs766820225 Chr.11: 71837382-71837382 on Build GRCh38
rs370343685 Chr.11: 71837429-71837429 on Build GRCh38
rs754322529 Chr.11: 71837384-71837384 on Build GRCh38
rs58527722 Chr.11: 71837600-71837600 on Build GRCh38
rs773613245 Chr.11: 71837376-71837376 on Build GRCh38
rs747515157 Chr.11: 71837444-71837444 on Build GRCh38
rs772590345 Chr.11: 71837369-71837369 on Build GRCh38
rs757438969 Chr.11: 71837593-71837593 on Build GRCh38
rs375089195 Chr.11: 71837373-71837373 on Build GRCh38
rs775030808 Chr.11: 71837458-71837458 on Build GRCh38
rs760706013 Chr.11: 71837549-71837549 on Build GRCh38
rs761313598 Chr.11: 71837378-71837378 on Build GRCh38
rs561772986 Chr.11: 71837619-71837619 on Build GRCh38
rs779033335 Chr.11: 71837432-71837432 on Build GRCh38
rs762496329 Chr.11: 71837530-71837530 on Build GRCh38
rs372248720 Chr.11: 71837543-71837543 on Build GRCh38
rs748287572 Chr.11: 71837502-71837502 on Build GRCh38
rs1018610025 Chr.11: 71837411-71837411 on Build GRCh38
rs747100615 Chr.11: 71837508-71837508 on Build GRCh38
rs374723902 Chr.11: 71837576-71837576 on Build GRCh38
rs751651595 Chr.11: 71837592-71837592 on Build GRCh38
rs777703579 Chr.11: 71837505-71837505 on Build GRCh38
rs12285436 Chr.11: 71837419-71837419 on Build GRCh38
rs949950538 Chr.11: 71837448-71837448 on Build GRCh38
rs760777502 Chr.11: 71837414-71837416 on Build GRCh38
rs200656078 Chr.11: 71837447-71837447 on Build GRCh38
rs775589583 Chr.11: 71837412-71837413 on Build GRCh38
rs780949189 Chr.11: 71837420-71837420 on Build GRCh38
rs751417471 Chr.11: 71837405-71837405 on Build GRCh38
rs750178072 Chr.11: 71837489-71837489 on Build GRCh38
rs202154400 Chr.11: 71837366-71837366 on Build GRCh38
rs181751318 Chr.11: 71837442-71837442 on Build GRCh38
rs1033144924 Chr.11: 71837615-71837615 on Build GRCh38
rs778147372 Chr.11: 71837583-71837583 on Build GRCh38
rs7120658 Chr.11: 71837498-71837498 on Build GRCh38

View Full Product Details