Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 109330481-109330796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPPA4
Entrez Gene ID
55211
Gene Name
developmental pluripotency associated 4
Gene Aliases
2410091M23Rik
UniGene
Hs.317659
Gene Chromosome Location
Chr.3: 109326141-109337572 on Build GRCh38
Transcripts
NM_018189.3 XM_011512954.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204510 c.577G>T p.V193L Chr.3: 109330626-109330626 on Build GRCh38
4112121 c.421A>G p.I141V Chr.3: 109330782-109330782 on Build GRCh38
341918 c.620C>A p.A207E Chr.3: 109330583-109330583 on Build GRCh38
7234825 c.467C>A p.T156K Chr.3: 109330736-109330736 on Build GRCh38
4112119 c.600G>T p.E200D Chr.3: 109330603-109330603 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs886124178 Chr.3: 109330758-109330758 on Build GRCh38
rs1048273008 Chr.3: 109330707-109330707 on Build GRCh38
rs947201148 Chr.3: 109330602-109330602 on Build GRCh38
rs201448153 Chr.3: 109330583-109330583 on Build GRCh38
rs747488047 Chr.3: 109330514-109330514 on Build GRCh38
rs765696593 Chr.3: 109330758-109330759 on Build GRCh38
rs745469049 Chr.3: 109330517-109330517 on Build GRCh38
rs768800699 Chr.3: 109330732-109330732 on Build GRCh38
rs772033541 Chr.3: 109330656-109330656 on Build GRCh38
rs13059848 Chr.3: 109330537-109330537 on Build GRCh38
rs774853142 Chr.3: 109330733-109330733 on Build GRCh38
rs556329682 Chr.3: 109330676-109330676 on Build GRCh38
rs199804229 Chr.3: 109330684-109330684 on Build GRCh38
rs370176490 Chr.3: 109330511-109330511 on Build GRCh38
rs780828761 Chr.3: 109330591-109330591 on Build GRCh38
rs368294457 Chr.3: 109330713-109330713 on Build GRCh38
rs768647140 Chr.3: 109330621-109330623 on Build GRCh38
rs748894803 Chr.3: 109330640-109330640 on Build GRCh38
rs990657669 Chr.3: 109330658-109330658 on Build GRCh38
rs749174334 Chr.3: 109330530-109330530 on Build GRCh38
rs745411606 Chr.3: 109330616-109330616 on Build GRCh38
rs532069241 Chr.3: 109330582-109330582 on Build GRCh38
rs200697559 Chr.3: 109330682-109330682 on Build GRCh38
rs777627521 Chr.3: 109330704-109330704 on Build GRCh38
rs934658424 Chr.3: 109330647-109330647 on Build GRCh38
rs769774536 Chr.3: 109330665-109330665 on Build GRCh38
rs367909790 Chr.3: 109330534-109330534 on Build GRCh38
rs3905734 Chr.3: 109330526-109330526 on Build GRCh38
rs746955852 Chr.3: 109330708-109330708 on Build GRCh38
rs765508136 Chr.3: 109330565-109330565 on Build GRCh38
rs201703878 Chr.3: 109330736-109330736 on Build GRCh38
rs537248080 Chr.3: 109330735-109330735 on Build GRCh38
rs375086646 Chr.3: 109330626-109330626 on Build GRCh38
rs1006358421 Chr.3: 109330739-109330739 on Build GRCh38
rs754650082 Chr.3: 109330748-109330748 on Build GRCh38
rs762470455 Chr.3: 109330759-109330759 on Build GRCh38
rs768456586 Chr.3: 109330644-109330644 on Build GRCh38
rs759666609 Chr.3: 109330662-109330662 on Build GRCh38
rs769619300 Chr.3: 109330518-109330518 on Build GRCh38
rs759719517 Chr.3: 109330549-109330549 on Build GRCh38
rs114878637 Chr.3: 109330738-109330738 on Build GRCh38
rs774037203 Chr.3: 109330651-109330651 on Build GRCh38
rs763118338 Chr.3: 109330669-109330669 on Build GRCh38
rs902012074 Chr.3: 109330698-109330698 on Build GRCh38
rs933603168 Chr.3: 109330737-109330737 on Build GRCh38
rs977994190 Chr.3: 109330590-109330590 on Build GRCh38
rs978574038 Chr.3: 109330631-109330631 on Build GRCh38
rs200311182 Chr.3: 109330536-109330536 on Build GRCh38
rs752019060 Chr.3: 109330673-109330673 on Build GRCh38
rs775458371 Chr.3: 109330717-109330717 on Build GRCh38
rs543557352 Chr.3: 109330538-109330538 on Build GRCh38
rs754636421 Chr.3: 109330686-109330686 on Build GRCh38
rs373512697 Chr.3: 109330507-109330507 on Build GRCh38
rs763219306 Chr.3: 109330724-109330724 on Build GRCh38
rs775717386 Chr.3: 109330667-109330667 on Build GRCh38
rs554506484 Chr.3: 109330759-109330759 on Build GRCh38
rs777175856 Chr.3: 109330679-109330680 on Build GRCh38
rs16834356 Chr.3: 109330660-109330660 on Build GRCh38
rs775118512 Chr.3: 109330528-109330528 on Build GRCh38
rs778483710 Chr.3: 109330766-109330766 on Build GRCh38
rs181240950 Chr.3: 109330672-109330672 on Build GRCh38
rs142452442 Chr.3: 109330515-109330515 on Build GRCh38
rs143598626 Chr.3: 109330588-109330588 on Build GRCh38
rs199540692 Chr.3: 109330564-109330564 on Build GRCh38
rs748055827 Chr.3: 109330655-109330655 on Build GRCh38
rs748000660 Chr.3: 109330689-109330689 on Build GRCh38
rs988953410 Chr.3: 109330574-109330574 on Build GRCh38
rs771980680 Chr.3: 109330697-109330697 on Build GRCh38
rs753329314 Chr.3: 109330741-109330741 on Build GRCh38
rs776736639 Chr.3: 109330543-109330543 on Build GRCh38

View Full Product Details