Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 39268663-39269114 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNL2
Entrez Gene ID
282616
Gene Name
interferon lambda 2
Gene Aliases
IL-28A, IL28A
UniGene
Hs.567792
Gene Chromosome Location
Chr.19: 39268517-39270092 on Build GRCh38
Transcripts
NM_172138.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4581010 c.95A>G p.H32R Chr.19: 39268761-39268761 on Build GRCh38
3722163 c.37C>T p.L13L Chr.19: 39268703-39268703 on Build GRCh38
4563583 c.97G>A p.G33R Chr.19: 39268763-39268763 on Build GRCh38
7371816 c.17C>G p.T6S Chr.19: 39268683-39268683 on Build GRCh38
389293 c.85G>T p.A29S Chr.19: 39268751-39268751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199501344 Chr.19: 39268773-39268773 on Build GRCh38
rs759745962 Chr.19: 39268878-39268878 on Build GRCh38
rs754931534 Chr.19: 39268734-39268734 on Build GRCh38
rs535520944 Chr.19: 39268842-39268842 on Build GRCh38
rs1000104618 Chr.19: 39268913-39268913 on Build GRCh38
rs200675991 Chr.19: 39268864-39268864 on Build GRCh38
rs532926776 Chr.19: 39269067-39269067 on Build GRCh38
rs746675668 Chr.19: 39268746-39268746 on Build GRCh38
rs557053526 Chr.19: 39268860-39268860 on Build GRCh38
rs746621773 Chr.19: 39268685-39268685 on Build GRCh38
rs946389369 Chr.19: 39268937-39268937 on Build GRCh38
rs992450663 Chr.19: 39268940-39268940 on Build GRCh38
rs777634754 Chr.19: 39268811-39268811 on Build GRCh38
rs532766892 Chr.19: 39268993-39268993 on Build GRCh38
rs373005397 Chr.19: 39268810-39268810 on Build GRCh38
rs776222534 Chr.19: 39268760-39268760 on Build GRCh38
rs968687384 Chr.19: 39268689-39268689 on Build GRCh38
rs1031218741 Chr.19: 39268921-39268921 on Build GRCh38
rs200471919 Chr.19: 39268693-39268693 on Build GRCh38
rs550650355 Chr.19: 39269030-39269030 on Build GRCh38
rs565459671 Chr.19: 39268900-39268900 on Build GRCh38
rs553114 Chr.19: 39268902-39268902 on Build GRCh38
rs553088 Chr.19: 39268912-39268912 on Build GRCh38
rs397826845 Chr.19: 39268996-39268996 on Build GRCh38
rs770429697 Chr.19: 39268750-39268750 on Build GRCh38
rs762308663 Chr.19: 39268706-39268706 on Build GRCh38
rs200755331 Chr.19: 39268714-39268714 on Build GRCh38
rs371778513 Chr.19: 39268696-39268696 on Build GRCh38
rs554971 Chr.19: 39268695-39268695 on Build GRCh38
rs1042064784 Chr.19: 39268951-39268951 on Build GRCh38
rs753950573 Chr.19: 39268775-39268775 on Build GRCh38
rs114035390 Chr.19: 39269020-39269020 on Build GRCh38
rs972987698 Chr.19: 39269028-39269028 on Build GRCh38
rs369387979 Chr.19: 39268794-39268794 on Build GRCh38
rs561634632 Chr.19: 39269000-39269000 on Build GRCh38
rs369868159 Chr.19: 39268884-39268884 on Build GRCh38
rs199818417 Chr.19: 39268684-39268684 on Build GRCh38
rs773990908 Chr.19: 39268867-39268867 on Build GRCh38
rs779618556 Chr.19: 39268751-39268751 on Build GRCh38
rs979790736 Chr.19: 39268994-39268994 on Build GRCh38
rs762668168 Chr.19: 39268713-39268714 on Build GRCh38
rs748670339 Chr.19: 39268852-39268852 on Build GRCh38
rs780745473 Chr.19: 39268826-39268826 on Build GRCh38
rs918504543 Chr.19: 39269029-39269029 on Build GRCh38
rs1051076891 Chr.19: 39269049-39269049 on Build GRCh38
rs753803573 Chr.19: 39268730-39268730 on Build GRCh38
rs201014057 Chr.19: 39268703-39268703 on Build GRCh38
rs769396116 Chr.19: 39268762-39268762 on Build GRCh38
rs757050162 Chr.19: 39268903-39268903 on Build GRCh38
rs751259853 Chr.19: 39268813-39268813 on Build GRCh38
rs552927033 Chr.19: 39268820-39268820 on Build GRCh38
rs771298925 Chr.19: 39268871-39268871 on Build GRCh38
rs768460141 Chr.19: 39268787-39268787 on Build GRCh38
rs767990657 Chr.19: 39268769-39268769 on Build GRCh38
rs1022459163 Chr.19: 39268911-39268911 on Build GRCh38
rs372103073 Chr.19: 39268737-39268737 on Build GRCh38
rs765282881 Chr.19: 39268801-39268801 on Build GRCh38
rs758240436 Chr.19: 39268742-39268742 on Build GRCh38
rs574525470 Chr.19: 39268828-39268828 on Build GRCh38
rs752713056 Chr.19: 39268891-39268891 on Build GRCh38
rs551550242 Chr.19: 39269081-39269081 on Build GRCh38
rs369655847 Chr.19: 39268718-39268718 on Build GRCh38
rs955692886 Chr.19: 39268700-39268700 on Build GRCh38
rs376288905 Chr.19: 39268774-39268774 on Build GRCh38
rs778499103 Chr.19: 39269002-39269002 on Build GRCh38
rs376999742 Chr.19: 39268874-39268874 on Build GRCh38
rs961282087 Chr.19: 39268933-39268933 on Build GRCh38
rs138027659 Chr.19: 39268761-39268761 on Build GRCh38
rs777420552 Chr.19: 39268743-39268743 on Build GRCh38
rs562612532 Chr.19: 39269056-39269056 on Build GRCh38
rs540965298 Chr.19: 39268948-39268948 on Build GRCh38
rs969764779 Chr.19: 39268950-39268950 on Build GRCh38
rs368918145 Chr.19: 39268763-39268763 on Build GRCh38

View Full Product Details