Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15425808-15426173 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PIR
Entrez Gene ID
8544
Gene Name
pirin
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15384799-15493588 on Build GRCh38
Transcripts
NM_001018109.2 NM_003662.3 NR_037859.2

Gene Symbol
PIR-FIGF
Entrez Gene ID
100532742
Gene Name
PIR-FIGF readthrough
Gene Aliases
-
UniGene
Hs.495728
Gene Chromosome Location
Chr.X: 15345591-15491309 on Build GRCh38
Transcripts
NM_001018109.2 NM_003662.3 NR_037859.2

SNP Detail

SNP ID Chromosome Location
rs774124755 Chr.X: 15426066-15426066 on Build GRCh38
rs771437546 Chr.X: 15425864-15425864 on Build GRCh38
rs950081140 Chr.X: 15426006-15426006 on Build GRCh38
rs772251328 Chr.X: 15425989-15425989 on Build GRCh38
rs111780638 Chr.X: 15426094-15426094 on Build GRCh38
rs200390589 Chr.X: 15425879-15425881 on Build GRCh38
rs760741248 Chr.X: 15425999-15425999 on Build GRCh38
rs368014883 Chr.X: 15425921-15425921 on Build GRCh38
rs996991565 Chr.X: 15425868-15425868 on Build GRCh38
rs770715122 Chr.X: 15425903-15425903 on Build GRCh38
rs953093600 Chr.X: 15425971-15425971 on Build GRCh38
rs779413343 Chr.X: 15425935-15425935 on Build GRCh38
rs746613574 Chr.X: 15425870-15425870 on Build GRCh38
rs368608154 Chr.X: 15425875-15425875 on Build GRCh38
rs987103941 Chr.X: 15425861-15425861 on Build GRCh38
rs753891187 Chr.X: 15426124-15426124 on Build GRCh38
rs764238685 Chr.X: 15426012-15426012 on Build GRCh38
rs746739175 Chr.X: 15425945-15425945 on Build GRCh38
rs780763035 Chr.X: 15425954-15425954 on Build GRCh38
rs754259162 Chr.X: 15425915-15425915 on Build GRCh38
rs183177987 Chr.X: 15426100-15426100 on Build GRCh38
rs768548608 Chr.X: 15425871-15425871 on Build GRCh38
rs761528281 Chr.X: 15425876-15425876 on Build GRCh38
rs779589221 Chr.X: 15425922-15425922 on Build GRCh38
rs750346122 Chr.X: 15425860-15425861 on Build GRCh38
rs779792525 Chr.X: 15425869-15425869 on Build GRCh38
rs759125882 Chr.X: 15425860-15425860 on Build GRCh38
rs954431759 Chr.X: 15425838-15425838 on Build GRCh38
rs762777791 Chr.X: 15425975-15425975 on Build GRCh38
rs773172117 Chr.X: 15425974-15425974 on Build GRCh38
rs776946260 Chr.X: 15426017-15426017 on Build GRCh38
rs187855264 Chr.X: 15426125-15426125 on Build GRCh38
rs1044407956 Chr.X: 15426037-15426037 on Build GRCh38
rs769537322 Chr.X: 15425970-15425970 on Build GRCh38
rs781277264 Chr.X: 15425859-15425860 on Build GRCh38
rs371272133 Chr.X: 15425887-15425887 on Build GRCh38
rs762223686 Chr.X: 15425893-15425893 on Build GRCh38
rs779692852 Chr.X: 15426036-15426037 on Build GRCh38
rs962956251 Chr.X: 15425858-15425859 on Build GRCh38
rs376827172 Chr.X: 15426015-15426015 on Build GRCh38
rs753011154 Chr.X: 15425907-15425907 on Build GRCh38
rs1017569191 Chr.X: 15425837-15425837 on Build GRCh38
rs747955981 Chr.X: 15425957-15425957 on Build GRCh38
rs372186417 Chr.X: 15425998-15425998 on Build GRCh38
rs750894736 Chr.X: 15426049-15426049 on Build GRCh38
rs748258826 Chr.X: 15425900-15425900 on Build GRCh38
rs750922769 Chr.X: 15425927-15425927 on Build GRCh38
rs929148493 Chr.X: 15425936-15425936 on Build GRCh38
rs374143293 Chr.X: 15425891-15425891 on Build GRCh38
rs760795372 Chr.X: 15425911-15425911 on Build GRCh38
rs768193675 Chr.X: 15425953-15425953 on Build GRCh38
rs375030091 Chr.X: 15425844-15425844 on Build GRCh38
rs764587964 Chr.X: 15425912-15425912 on Build GRCh38
rs1000195695 Chr.X: 15426085-15426085 on Build GRCh38
rs757971715 Chr.X: 15425932-15425932 on Build GRCh38

View Full Product Details