Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8142319-8142493 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PER1
Entrez Gene ID
5187
Gene Name
period circadian clock 1
Gene Aliases
PER, RIGUI, hPER
UniGene
Hs.445534
Gene Chromosome Location
Chr.17: 8140470-8156405 on Build GRCh38
Transcripts
NM_002616.2 XM_005256689.1 XM_005256690.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
249071 c.3355G>A p.D1119N Chr.17: 8142363-8142363 on Build GRCh38
6387714 c.3338G>T p.G1113V Chr.17: 8142380-8142380 on Build GRCh38
6002949 c.3321G>A p.E1107E Chr.17: 8142397-8142397 on Build GRCh38
6366368 c.3367A>G p.K1123E Chr.17: 8142351-8142351 on Build GRCh38
3523871 c.3315G>A p.E1105E Chr.17: 8142403-8142403 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761827422 Chr.17: 8142351-8142351 on Build GRCh38
rs750719734 Chr.17: 8142467-8142467 on Build GRCh38
rs529303485 Chr.17: 8142382-8142382 on Build GRCh38
rs368035422 Chr.17: 8142384-8142384 on Build GRCh38
rs762847470 Chr.17: 8142415-8142415 on Build GRCh38
rs768367754 Chr.17: 8142471-8142471 on Build GRCh38
rs202048579 Chr.17: 8142379-8142379 on Build GRCh38
rs750301840 Chr.17: 8142358-8142358 on Build GRCh38
rs112743620 Chr.17: 8142396-8142396 on Build GRCh38
rs752793680 Chr.17: 8142378-8142378 on Build GRCh38
rs201102174 Chr.17: 8142468-8142468 on Build GRCh38
rs371070132 Chr.17: 8142414-8142414 on Build GRCh38
rs114068540 Chr.17: 8142439-8142439 on Build GRCh38
rs780669269 Chr.17: 8142473-8142473 on Build GRCh38
rs768486009 Chr.17: 8142468-8142468 on Build GRCh38
rs371186640 Chr.17: 8142405-8142405 on Build GRCh38
rs540838050 Chr.17: 8142393-8142393 on Build GRCh38
rs375691411 Chr.17: 8142424-8142424 on Build GRCh38
rs779718056 Chr.17: 8142387-8142387 on Build GRCh38
rs746690728 Chr.17: 8142390-8142390 on Build GRCh38
rs757689443 Chr.17: 8142455-8142455 on Build GRCh38
rs149653091 Chr.17: 8142369-8142369 on Build GRCh38
rs752918242 Chr.17: 8142437-8142437 on Build GRCh38
rs771598776 Chr.17: 8142385-8142385 on Build GRCh38
rs368543052 Chr.17: 8142464-8142464 on Build GRCh38
rs760607110 Chr.17: 8142370-8142396 on Build GRCh38
rs374121023 Chr.17: 8142383-8142383 on Build GRCh38
rs147163947 Chr.17: 8142448-8142448 on Build GRCh38
rs778020756 Chr.17: 8142381-8142381 on Build GRCh38
rs764201177 Chr.17: 8142442-8142442 on Build GRCh38
rs147413427 Chr.17: 8142346-8142346 on Build GRCh38
rs746600933 Chr.17: 8142469-8142469 on Build GRCh38
rs1008816205 Chr.17: 8142406-8142406 on Build GRCh38
rs954121684 Chr.17: 8142376-8142376 on Build GRCh38
rs755852172 Chr.17: 8142352-8142352 on Build GRCh38
rs769829455 Chr.17: 8142412-8142412 on Build GRCh38
rs755144407 Chr.17: 8142361-8142361 on Build GRCh38
rs775278263 Chr.17: 8142363-8142389 on Build GRCh38

View Full Product Details