Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 42287400-42287551 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CIC
Entrez Gene ID
23152
Gene Name
capicua transcriptional repressor
Gene Aliases
-
UniGene
Hs.388236
Gene Chromosome Location
Chr.19: 42268537-42295796 on Build GRCh38
Transcripts
NM_001304815.1 NM_015125.4 XM_005258673.2 XM_005258674.2 XM_005258675.2 XM_011526660.2 XM_011526661.2 XM_011526662.2 XM_011526663.2 XM_011526664.2 XM_011526665.2 XM_011526666.2 XM_011526667.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1304659 c.556G>C p.E186Q Chr.19: 42287423-42287423 on Build GRCh38
7155188 c.543G>A p.R181R Chr.19: 42287410-42287410 on Build GRCh38
132835 c.579delC p.N193fs*12 Chr.19: 42287446-42287446 on Build GRCh38
997303 c.566G>A p.G189E Chr.19: 42287433-42287433 on Build GRCh38
997304 c.568C>T p.R190C Chr.19: 42287435-42287435 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202120202 Chr.19: 42287419-42287419 on Build GRCh38
rs771011282 Chr.19: 42287506-42287506 on Build GRCh38
rs766543605 Chr.19: 42287522-42287522 on Build GRCh38
rs371508603 Chr.19: 42287491-42287491 on Build GRCh38
rs770380344 Chr.19: 42287421-42287421 on Build GRCh38
rs752629455 Chr.19: 42287436-42287436 on Build GRCh38
rs775459264 Chr.19: 42287488-42287488 on Build GRCh38
rs757154610 Chr.19: 42287500-42287501 on Build GRCh38
rs773981943 Chr.19: 42287478-42287478 on Build GRCh38
rs921491201 Chr.19: 42287475-42287475 on Build GRCh38
rs771542960 Chr.19: 42287483-42287483 on Build GRCh38
rs748898613 Chr.19: 42287465-42287465 on Build GRCh38
rs189233154 Chr.19: 42287492-42287492 on Build GRCh38
rs750667375 Chr.19: 42287517-42287517 on Build GRCh38
rs375561179 Chr.19: 42287496-42287496 on Build GRCh38
rs765610879 Chr.19: 42287507-42287507 on Build GRCh38
rs763930907 Chr.19: 42287440-42287440 on Build GRCh38
rs768949914 Chr.19: 42287461-42287461 on Build GRCh38
rs760430429 Chr.19: 42287438-42287438 on Build GRCh38
rs150792335 Chr.19: 42287450-42287450 on Build GRCh38
rs373664149 Chr.19: 42287459-42287459 on Build GRCh38
rs539878308 Chr.19: 42287515-42287515 on Build GRCh38
rs761952423 Chr.19: 42287504-42287504 on Build GRCh38
rs144903246 Chr.19: 42287449-42287449 on Build GRCh38
rs368685259 Chr.19: 42287521-42287521 on Build GRCh38
rs368084583 Chr.19: 42287460-42287460 on Build GRCh38
rs759484594 Chr.19: 42287431-42287431 on Build GRCh38
rs746989941 Chr.19: 42287437-42287437 on Build GRCh38
rs745452217 Chr.19: 42287482-42287482 on Build GRCh38
rs200883150 Chr.19: 42287445-42287445 on Build GRCh38
rs781571434 Chr.19: 42287464-42287464 on Build GRCh38
rs370245391 Chr.19: 42287456-42287456 on Build GRCh38
rs529525339 Chr.19: 42287470-42287470 on Build GRCh38
rs773785693 Chr.19: 42287428-42287428 on Build GRCh38
rs767502801 Chr.19: 42287435-42287435 on Build GRCh38
rs939764861 Chr.19: 42287514-42287514 on Build GRCh38
rs767494967 Chr.19: 42287516-42287517 on Build GRCh38
rs754033796 Chr.19: 42287442-42287442 on Build GRCh38

View Full Product Details