Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14148489-14148663 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XPC
Entrez Gene ID
7508
Gene Name
XPC complex subunit, DNA damage recognition and repair factor
Gene Aliases
RAD4, XP3, XPCC, p125
UniGene
Hs.475538
Gene Chromosome Location
Chr.3: 14145147-14178672 on Build GRCh38
Transcripts
NM_004628.4 NR_027299.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5863657 c.2344G>A p.V782M Chr.3: 14148638-14148638 on Build GRCh38
3427161 c.2365G>T p.D789Y Chr.3: 14148617-14148617 on Build GRCh38
7221909 c.2335C>G p.L779V Chr.3: 14148647-14148647 on Build GRCh38
1039299 c.2419G>A p.V807M Chr.3: 14148563-14148563 on Build GRCh38
6345823 c.2348C>A p.A783D Chr.3: 14148634-14148634 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768821703 Chr.3: 14148550-14148550 on Build GRCh38
rs756757380 Chr.3: 14148632-14148632 on Build GRCh38
rs770292125 Chr.3: 14148586-14148586 on Build GRCh38
rs747213990 Chr.3: 14148576-14148576 on Build GRCh38
rs763252584 Chr.3: 14148603-14148603 on Build GRCh38
rs747041300 Chr.3: 14148527-14148528 on Build GRCh38
rs373971774 Chr.3: 14148527-14148527 on Build GRCh38
rs199828421 Chr.3: 14148564-14148564 on Build GRCh38
rs761877857 Chr.3: 14148581-14148581 on Build GRCh38
rs778987248 Chr.3: 14148560-14148560 on Build GRCh38
rs200148127 Chr.3: 14148578-14148578 on Build GRCh38
rs755615146 Chr.3: 14148545-14148545 on Build GRCh38
rs115309145 Chr.3: 14148520-14148520 on Build GRCh38
rs753463306 Chr.3: 14148631-14148631 on Build GRCh38
rs1045962488 Chr.3: 14148627-14148627 on Build GRCh38
rs999397100 Chr.3: 14148515-14148515 on Build GRCh38
rs375378876 Chr.3: 14148528-14148528 on Build GRCh38
rs777385996 Chr.3: 14148550-14148550 on Build GRCh38
rs767093217 Chr.3: 14148529-14148529 on Build GRCh38
rs762357025 Chr.3: 14148514-14148514 on Build GRCh38
rs757316607 Chr.3: 14148559-14148559 on Build GRCh38
rs370753746 Chr.3: 14148638-14148638 on Build GRCh38
rs767045808 Chr.3: 14148620-14148620 on Build GRCh38
rs774552303 Chr.3: 14148609-14148609 on Build GRCh38
rs745660242 Chr.3: 14148563-14148563 on Build GRCh38
rs546461471 Chr.3: 14148639-14148639 on Build GRCh38
rs982648261 Chr.3: 14148617-14148617 on Build GRCh38
rs746768609 Chr.3: 14148641-14148641 on Build GRCh38
rs775734716 Chr.3: 14148519-14148519 on Build GRCh38
rs369028638 Chr.3: 14148544-14148544 on Build GRCh38
rs773652980 Chr.3: 14148599-14148599 on Build GRCh38
rs577415509 Chr.3: 14148640-14148640 on Build GRCh38
rs755294791 Chr.3: 14148642-14148642 on Build GRCh38
rs760421156 Chr.3: 14148613-14148613 on Build GRCh38
rs752120059 Chr.3: 14148570-14148570 on Build GRCh38
rs753373269 Chr.3: 14148558-14148558 on Build GRCh38
rs766730833 Chr.3: 14148605-14148605 on Build GRCh38
rs928148829 Chr.3: 14148634-14148634 on Build GRCh38

View Full Product Details