Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 155187313-155187482 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MUC1
Entrez Gene ID
4582
Gene Name
mucin 1, cell surface associated
Gene Aliases
ADMCKD, ADMCKD1, CA 15-3, CD227, EMA, H23AG, KL-6, MAM6, MCD, MCKD, MCKD1, MUC-1, MUC-1/SEC, MUC-1/X, MUC1/ZD, PEM, PEMT, PUM
UniGene
Hs.89603
Gene Chromosome Location
Chr.1: 155185824-155192915 on Build GRCh38
Transcripts
NM_001018016.2 NM_001018017.2 NM_001044390.2 NM_001044391.2 NM_001044392.2 NM_001044393.2 NM_001204285.1 NM_001204286.1 NM_001204287.1 NM_001204288.1 NM_001204289.1 NM_001204290.1 NM_001204291.1 NM_001204292.1 NM_001204293.1 NM_001204294.1 NM_001204295.1 NM_001204296.1 NM_001204297.1 NM_002456.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6717167 c.577_580delTGTC p.C193fs*22 Chr.1: 155187365-155187368 on Build GRCh38
94738 c.581T>C p.V194A Chr.1: 155187471-155187471 on Build GRCh38
3705037 c.646C>T p.P216S Chr.1: 155187326-155187326 on Build GRCh38
6717155 c.587G>A p.R196H Chr.1: 155187358-155187358 on Build GRCh38
2210020 c.590G>A p.R197Q Chr.1: 155187355-155187355 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199768496 Chr.1: 155187456-155187456 on Build GRCh38
rs758224990 Chr.1: 155187394-155187394 on Build GRCh38
rs769527913 Chr.1: 155187420-155187420 on Build GRCh38
rs746266997 Chr.1: 155187430-155187430 on Build GRCh38
rs1611771 Chr.1: 155187419-155187419 on Build GRCh38
rs988044105 Chr.1: 155187372-155187372 on Build GRCh38
rs749456681 Chr.1: 155187421-155187421 on Build GRCh38
rs371436760 Chr.1: 155187431-155187431 on Build GRCh38
rs367561217 Chr.1: 155187420-155187420 on Build GRCh38
rs369318053 Chr.1: 155187442-155187442 on Build GRCh38
rs747704526 Chr.1: 155187365-155187368 on Build GRCh38
rs750236420 Chr.1: 155187383-155187383 on Build GRCh38
rs913567944 Chr.1: 155187386-155187386 on Build GRCh38
rs977872999 Chr.1: 155187399-155187404 on Build GRCh38
rs878946562 Chr.1: 155187377-155187377 on Build GRCh38
rs982193787 Chr.1: 155187403-155187403 on Build GRCh38
rs879028475 Chr.1: 155187376-155187376 on Build GRCh38
rs761508564 Chr.1: 155187433-155187433 on Build GRCh38
rs753535070 Chr.1: 155187350-155187350 on Build GRCh38
rs774814129 Chr.1: 155187344-155187344 on Build GRCh38
rs957938526 Chr.1: 155187355-155187355 on Build GRCh38
rs374941067 Chr.1: 155187438-155187438 on Build GRCh38
rs747587046 Chr.1: 155187435-155187435 on Build GRCh38
rs773917931 Chr.1: 155187345-155187345 on Build GRCh38
rs751544953 Chr.1: 155187405-155187405 on Build GRCh38
rs757509583 Chr.1: 155187425-155187425 on Build GRCh38
rs756151925 Chr.1: 155187412-155187412 on Build GRCh38
rs764882391 Chr.1: 155187358-155187358 on Build GRCh38
rs763738651 Chr.1: 155187348-155187348 on Build GRCh38
rs780480243 Chr.1: 155187351-155187352 on Build GRCh38
rs929525354 Chr.1: 155187444-155187444 on Build GRCh38
rs1032150970 Chr.1: 155187351-155187351 on Build GRCh38
rs780058580 Chr.1: 155187404-155187404 on Build GRCh38
rs779110864 Chr.1: 155187426-155187426 on Build GRCh38
rs761613959 Chr.1: 155187356-155187356 on Build GRCh38

View Full Product Details