Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 43529453-43529608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DHX8
Entrez Gene ID
1659
Gene Name
DEAH-box helicase 8
Gene Aliases
DDX8, HRH1, PRP22, PRPF22
UniGene
Hs.463105
Gene Chromosome Location
Chr.17: 43483865-43544787 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001261439.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Gene Symbol
ETV4
Entrez Gene ID
2118
Gene Name
ETS variant 4
Gene Aliases
E1A-F, E1AF, PEA3, PEAS3
UniGene
Hs.434059
Gene Chromosome Location
Chr.17: 43527843-43546432 on Build GRCh38
Transcripts
NM_001322219.1 NM_001079675.2 NM_001261437.1 NM_001261438.1 NM_001261439.1 NM_001986.2 XM_011524514.1 XM_017024349.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4130116 c.1050G>T p.L350F Chr.17: 43529582-43529582 on Build GRCh38
1302873 c.1104G>A p.E368E Chr.17: 43529528-43529528 on Build GRCh38
122908 c.1094G>A p.R365Q Chr.17: 43529538-43529538 on Build GRCh38
7416499 c.1121C>A p.P374H Chr.17: 43529511-43529511 on Build GRCh38
6839421 c.1081G>A p.A361T Chr.17: 43529551-43529551 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756584324 Chr.17: 43529586-43529586 on Build GRCh38
rs776142667 Chr.17: 43529539-43529539 on Build GRCh38
rs376288935 Chr.17: 43529544-43529544 on Build GRCh38
rs750774903 Chr.17: 43529584-43529584 on Build GRCh38
rs374306817 Chr.17: 43529538-43529538 on Build GRCh38
rs1009668412 Chr.17: 43529572-43529572 on Build GRCh38
rs1002680213 Chr.17: 43529547-43529547 on Build GRCh38
rs760615534 Chr.17: 43529531-43529531 on Build GRCh38
rs772037381 Chr.17: 43529516-43529516 on Build GRCh38
rs946119546 Chr.17: 43529578-43529578 on Build GRCh38
rs201929762 Chr.17: 43529524-43529524 on Build GRCh38
rs767829041 Chr.17: 43529581-43529581 on Build GRCh38
rs781628034 Chr.17: 43529479-43529479 on Build GRCh38
rs749388064 Chr.17: 43529493-43529493 on Build GRCh38
rs950004591 Chr.17: 43529520-43529520 on Build GRCh38
rs377565878 Chr.17: 43529512-43529512 on Build GRCh38
rs780428673 Chr.17: 43529587-43529587 on Build GRCh38
rs768920741 Chr.17: 43529506-43529506 on Build GRCh38
rs199811549 Chr.17: 43529543-43529543 on Build GRCh38
rs149219456 Chr.17: 43529554-43529554 on Build GRCh38
rs925179572 Chr.17: 43529517-43529517 on Build GRCh38
rs570875728 Chr.17: 43529476-43529476 on Build GRCh38
rs762134193 Chr.17: 43529568-43529568 on Build GRCh38
rs150661130 Chr.17: 43529492-43529492 on Build GRCh38

View Full Product Details