Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 104772845-104773005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AKT1
Entrez Gene ID
207
Gene Name
AKT serine/threonine kinase 1
Gene Aliases
AKT, CWS6, PKB, PKB-ALPHA, PRKBA, RAC, RAC-ALPHA
UniGene
Hs.525622
Gene Chromosome Location
Chr.14: 104769349-104795743 on Build GRCh38
Transcripts
NM_001014431.1 NM_001014432.1 NM_005163.2 XM_005267401.1 XM_017021075.1 XM_017021076.1 XM_017021077.1 XM_017021078.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5044339 c.1069C>T p.L357F Chr.14: 104772981-104772981 on Build GRCh38
6981608 c.1061A>T p.H354L Chr.14: 104772989-104772989 on Build GRCh38
6979438 c.1106C>T p.P369L Chr.14: 104772944-104772944 on Build GRCh38
6924776 c.1142C>T p.S381L Chr.14: 104772908-104772908 on Build GRCh38
1235769 c.1099C>T p.R367C Chr.14: 104772951-104772951 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372502847 Chr.14: 104772963-104772963 on Build GRCh38
rs767207905 Chr.14: 104772901-104772901 on Build GRCh38
rs746272761 Chr.14: 104772941-104772941 on Build GRCh38
rs763680129 Chr.14: 104772871-104772871 on Build GRCh38
rs952698197 Chr.14: 104772872-104772872 on Build GRCh38
rs755174756 Chr.14: 104772904-104772904 on Build GRCh38
rs373253729 Chr.14: 104772927-104772927 on Build GRCh38
rs200702576 Chr.14: 104772868-104772868 on Build GRCh38
rs889182971 Chr.14: 104772932-104772932 on Build GRCh38
rs369527283 Chr.14: 104772916-104772916 on Build GRCh38
rs781388586 Chr.14: 104772938-104772938 on Build GRCh38
rs758624702 Chr.14: 104772892-104772894 on Build GRCh38
rs761810870 Chr.14: 104772892-104772892 on Build GRCh38
rs1050565251 Chr.14: 104772924-104772924 on Build GRCh38
rs773975370 Chr.14: 104772958-104772958 on Build GRCh38
rs11555432 Chr.14: 104772980-104772980 on Build GRCh38
rs150399268 Chr.14: 104772943-104772943 on Build GRCh38
rs529348439 Chr.14: 104772869-104772869 on Build GRCh38
rs747408700 Chr.14: 104772937-104772937 on Build GRCh38
rs11555431 Chr.14: 104772888-104772888 on Build GRCh38
rs751392710 Chr.14: 104772873-104772873 on Build GRCh38
rs549370342 Chr.14: 104772942-104772942 on Build GRCh38
rs369520527 Chr.14: 104772950-104772950 on Build GRCh38
rs374865919 Chr.14: 104772928-104772928 on Build GRCh38

View Full Product Details