Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 150578137-150578278 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MCL1
Entrez Gene ID
4170
Gene Name
BCL2 family apoptosis regulator
Gene Aliases
BCL2L3, EAT, MCL1-ES, MCL1L, MCL1S, Mcl-1, TM, bcl2-L-3, mcl1/EAT
UniGene
Hs.632486
Gene Chromosome Location
Chr.1: 150574551-150579738 on Build GRCh38
Transcripts
NM_001197320.1 NM_021960.4 NM_182763.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5038403 c.936+2T>G p.? Chr.1: 150578242-150578242 on Build GRCh38
6976395 c.932G>A p.G311D Chr.1: 150578248-150578248 on Build GRCh38
5947076 c.933C>T p.G311G Chr.1: 150578247-150578247 on Build GRCh38
4022042 c.913T>G p.W305G Chr.1: 150578267-150578267 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1001856748 Chr.1: 150578164-150578164 on Build GRCh38
rs749306667 Chr.1: 150578249-150578249 on Build GRCh38
rs779312384 Chr.1: 150578255-150578255 on Build GRCh38
rs753670749 Chr.1: 150578227-150578227 on Build GRCh38
rs766888806 Chr.1: 150578203-150578203 on Build GRCh38
rs999919956 Chr.1: 150578158-150578164 on Build GRCh38
rs587650071 Chr.1: 150578182-150578183 on Build GRCh38
rs202220481 Chr.1: 150578204-150578204 on Build GRCh38
rs1024008111 Chr.1: 150578191-150578191 on Build GRCh38
rs755934201 Chr.1: 150578213-150578213 on Build GRCh38
rs587718092 Chr.1: 150578183-150578183 on Build GRCh38
rs960288801 Chr.1: 150578170-150578170 on Build GRCh38
rs777774413 Chr.1: 150578217-150578217 on Build GRCh38
rs867437014 Chr.1: 150578225-150578242 on Build GRCh38
rs1035031712 Chr.1: 150578166-150578166 on Build GRCh38
rs74848639 Chr.1: 150578242-150578242 on Build GRCh38
rs868269964 Chr.1: 150578180-150578180 on Build GRCh38
rs1014326833 Chr.1: 150578183-150578183 on Build GRCh38
rs201581549 Chr.1: 150578198-150578198 on Build GRCh38
rs780734052 Chr.1: 150578241-150578261 on Build GRCh38

View Full Product Details