Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 207555666-207555794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CREB1
Entrez Gene ID
1385
Gene Name
cAMP responsive element binding protein 1
Gene Aliases
CREB, CREB-1
UniGene
Hs.516646
Gene Chromosome Location
Chr.2: 207529892-207605989 on Build GRCh38
Transcripts
NM_001320793.1 NM_004379.4 NM_134442.4 NR_135473.1 XM_011510645.1 XM_011510646.2 XM_011510647.2 XM_011510648.2 XM_011510649.2 XM_017003399.1 XM_017003400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1015518 c.79G>A p.V27I Chr.2: 207555714-207555714 on Build GRCh38
2714427 c.45A>T p.A15A Chr.2: 207555680-207555680 on Build GRCh38
4090624 c.91C>T p.P31S Chr.2: 207555726-207555726 on Build GRCh38
2714428 c.66C>T p.N22N Chr.2: 207555701-207555701 on Build GRCh38
4881775 c.37G>A p.G13R Chr.2: 207555672-207555672 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs778482192 Chr.2: 207555751-207555752 on Build GRCh38
rs771018333 Chr.2: 207555715-207555715 on Build GRCh38
rs56292136 Chr.2: 207555702-207555702 on Build GRCh38
rs763981369 Chr.2: 207555696-207555696 on Build GRCh38
rs114514847 Chr.2: 207555738-207555738 on Build GRCh38
rs762778822 Chr.2: 207555693-207555693 on Build GRCh38
rs984795660 Chr.2: 207555741-207555741 on Build GRCh38
rs755767566 Chr.2: 207555733-207555733 on Build GRCh38
rs761256671 Chr.2: 207555703-207555703 on Build GRCh38
rs370037055 Chr.2: 207555710-207555710 on Build GRCh38

View Full Product Details