Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147615815-147615954 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BCL9
Entrez Gene ID
607
Gene Name
B-cell CLL/lymphoma 9
Gene Aliases
LGS
UniGene
Hs.415209
Gene Chromosome Location
Chr.1: 147541394-147626219 on Build GRCh38
Transcripts
NM_004326.3 XM_005272971.4 XM_005277417.3 XM_005277418.3 XM_017002026.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1747707 c.579G>A p.L193L Chr.1: 147615821-147615821 on Build GRCh38
6032045 c.660+1G>A p.? Chr.1: 147615903-147615903 on Build GRCh38
3473190 c.598A>G p.I200V Chr.1: 147615840-147615840 on Build GRCh38
1333658 c.660+1G>T p.? Chr.1: 147615903-147615903 on Build GRCh38
6582033 c.658C>G p.L220V Chr.1: 147615900-147615900 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs375569438 Chr.1: 147615884-147615884 on Build GRCh38
rs782239671 Chr.1: 147615868-147615868 on Build GRCh38
rs139276872 Chr.1: 147615840-147615840 on Build GRCh38
rs368566731 Chr.1: 147615900-147615900 on Build GRCh38
rs782322083 Chr.1: 147615863-147615863 on Build GRCh38
rs782275886 Chr.1: 147615862-147615862 on Build GRCh38
rs781910017 Chr.1: 147615927-147615927 on Build GRCh38
rs140111701 Chr.1: 147615869-147615869 on Build GRCh38
rs782599827 Chr.1: 147615843-147615843 on Build GRCh38
rs782515483 Chr.1: 147615852-147615852 on Build GRCh38
rs781872210 Chr.1: 147615851-147615851 on Build GRCh38
rs372542307 Chr.1: 147615914-147615914 on Build GRCh38
rs782349883 Chr.1: 147615874-147615874 on Build GRCh38
rs782485224 Chr.1: 147615842-147615842 on Build GRCh38
rs782042986 Chr.1: 147615897-147615897 on Build GRCh38
rs142730791 Chr.1: 147615895-147615895 on Build GRCh38
rs782577576 Chr.1: 147615864-147615864 on Build GRCh38
rs3737843 Chr.1: 147615921-147615921 on Build GRCh38
rs375695240 Chr.1: 147615860-147615860 on Build GRCh38
rs143819239 Chr.1: 147615896-147615896 on Build GRCh38
rs782009263 Chr.1: 147615880-147615880 on Build GRCh38

View Full Product Details