Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 91969580-91969747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRIP11
Entrez Gene ID
9321
Gene Name
thyroid hormone receptor interactor 11
Gene Aliases
ACG1A, CEV14, GMAP-210, GMAP210, TRIP-11, TRIP230
UniGene
Hs.632339
Gene Chromosome Location
Chr.14: 91965991-92040140 on Build GRCh38
Transcripts
NM_001321851.1 NM_004239.4 XM_017021787.1 XM_017021788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
958815 c.5934G>T p.K1978N Chr.14: 91969679-91969679 on Build GRCh38
5519279 c.5892A>G p.L1964L Chr.14: 91969721-91969721 on Build GRCh38
95410 c.5909G>A p.G1970E Chr.14: 91969704-91969704 on Build GRCh38
4885209 c.5940G>T p.*1980Y Chr.14: 91969673-91969673 on Build GRCh38
5848712 c.5930delT p.L1977fs*1 Chr.14: 91969683-91969683 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754053786 Chr.14: 91969717-91969717 on Build GRCh38
rs113605039 Chr.14: 91969713-91969713 on Build GRCh38
rs373269085 Chr.14: 91969664-91969664 on Build GRCh38
rs775880564 Chr.14: 91969659-91969659 on Build GRCh38
rs541929543 Chr.14: 91969629-91969629 on Build GRCh38
rs571487642 Chr.14: 91969614-91969614 on Build GRCh38
rs142075650 Chr.14: 91969724-91969724 on Build GRCh38
rs772345820 Chr.14: 91969680-91969680 on Build GRCh38
rs61742059 Chr.14: 91969726-91969726 on Build GRCh38
rs780662900 Chr.14: 91969632-91969632 on Build GRCh38
rs761029164 Chr.14: 91969710-91969710 on Build GRCh38
rs745585138 Chr.14: 91969637-91969637 on Build GRCh38
rs148788333 Chr.14: 91969609-91969609 on Build GRCh38
rs769402471 Chr.14: 91969640-91969640 on Build GRCh38
rs762368847 Chr.14: 91969676-91969676 on Build GRCh38
rs35979920 Chr.14: 91969639-91969640 on Build GRCh38
rs201626889 Chr.14: 91969666-91969666 on Build GRCh38
rs151317471 Chr.14: 91969691-91969691 on Build GRCh38

View Full Product Details